Sigma-Aldrich
Life Science > Stem Cell Biology > Embryonic and Induced Pluripotent Stem Cells > Induced Pluripotent Stem Cell FAQs
Cell Culture
Induced Pluripotent Stem Cell FAQs
Frequently Asked Questions
Glossary – definitions of stem cell terms
Protocols
iPS Products
Need technical help? Ask a Scientist
How are the induced pluripotent stem (iPS) cells produced?
Currently, iPS cells are produced by inserting copies of four stem cell-associated genes; Oct 3/4, Sox 2, Klf4, and c-Myc (or Oct 3/4, Sox 2, Nanog, and LIN28) into specialized cells using viral vectors. Shinya Yamanaka produced the first iPS cells from mouse cells in 2006. This was followed in 2007 by the generation of iPS cells from human somatic cells by both the Yamanaka lab and the lab of James Thomson.
What are the advantages of iPS cells over embryonic stem cells?
The advantage of iPS cells is that they are not derived from human embryos, which is the ethical concern in this field. By removing the bioethical issues, the scientists are more likely to obtain more federal funding and support. Another significant benefit of iPS cell technology would permit for creation of cell lines that are genetically tailored to a patient. This could eliminate the concern of immune rejection, where the body’s immune system identifies implanted cells or tissues as unknown and attacks them.
What are the risks associated with iPS cell use in humans?
The retroviruses used in the generation of iPSc are associated with cancer because they insert DNA anywhere in a cell's genome, which could potentially trigger the expression of cancer-causing genes. Another risk associated with iPS cell technology applied to humans is the fact that c-Myc, which is one of the genes used in reprogramming, is a known oncogene whose overexpression could also cause cancer.
How are iPS cells similar to ES cells?
iPS cells are similar to ES cells in morphology, teratoma formation, proliferation, as well as their ability to differentiate along a given lineage. They also express cell surface markers and genes that characterize ES cells.
Does iPS cell technology eliminate the need for embryonic stem cell research?
Recent advances do not eliminate the need for ES cell research since it is not yet quite clear whether iPS cells differ extensively from the embryonic stem cells. To bring stem cell research to clinical realization, it is necessary to investigate all the aspects in this field.
What are disease specific iPS cells?
Disease specific iPS cells are iPS cells generated from subjects with a genetic disease.
Is there an iPS cell bank and where?
The University of Wisconsin and James Thomson at the WiCell Research Institute started the first iPS cell bank. There, the scientist grow, test, store and distribute iPS cell lines.
Which media should be used for culturing iPS cells?
The recipe for media for culturing iPS cells was acquired from WiCell MEF-based iPS Cell Culture Protocols and Sigma's catalog numbers have been included for your convenience;
IPS Cell Culture Medium (250 ml)
  • 200 ml — DMEM-F12 (D8900, D2906)
  • 50 ml — Knockout Serum Replacement
  • 2.5 ml — 100 mM L-Glutamine (G8540) + BME Solution (73 mg of L-Glutamine + 5 ml of PBS (P5493) + 3.5 ml 2-Mercatoethanol (M7522))
  • 2.5 ml — Non-Essential Amino Acids (M7145)
  • 0.5 ml — 50 mg/ml Basic FGF solution (100 mg of bFGF (F0291, F9786) + 2 ml of 0.1% BSA (A2153) in PBS (P5368)).