Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Stemgent Reprogramming Lentiviruses
Reprogramming Lentiviruses for Mouse Cells
Stemgent Dox Lentivirus Set: mOKSM (Product code ST000021)
The Stemgent Dox Lentivirus Set: mOKSM provides an affordable inducible method for generating iPS cells. The set includes mOct4, mSox2, mc-Myc, and mKlf4 transcription factors along with reverse tetracycline transcriptional activator (rtTA) and GFP, prepackaged as lentivirus particles tested and proven to reprogram mouse somatic cells. Each unique virus is provided as a VSV-G pseudotyped virus stock capable of infecting both dividing and non-dividing cells. The expression of the four transcription factors, in conjunction with rtTA, has been shown to direct a variety of differentiated cell types to become reprogrammed iPS cells1.
References
  1. Brambrink, T., Foreman, R., Welstead, G.G., Lengner, C.J., Wernig, M., Suh, H., and Jaenisch, R. (2008) Sequential expression of pluripotency markers during direct reprogramming of mouse somatic cells. Cell Stem Cell 2, 151-159.
  2. Wernig, M., Lengner, C.J., Hanna, J., Lodato, M.A., Steine, E., Foreman, R., Staerk, J., Markoulaki, S., and Jaenisch, R. (2008) A drug-inducible transgenic system for direct reprogramming of multiple somatic cell types. Nat Biotechnol 26,916-924.
  3. Szymczak, A.L., Workman, C.J., Wang, Y., Vignali, K.M., Dilioglou, S., Vanin, E.F., and Vignali, D.A. (2004) Correction of multi-gene defi ciency in vivo using a single 'self-cleaving' 2A peptide-based retroviral vector. Nat Biotechnol 22, 589-594.
  4. Carey, B.W., Markoulaki, S., Hanna, J., Saha, K., Gao, Q., Mitalipova, M., and Jaenisch, R. (2009) Reprogramming of murine and human somatic cells using a single polycistronic vector. Proc Natl Acad Sci USA 106, 157-162.