Your Favorite Gene - Powered By Ingenuity
www.sigmaaldrich.com/stemcell
Sigma-Aldrich
Your Favorite Gene - Powered By Ingenuity
Find Activin-A Products
Gene INHBA; INHBA_HUMAN
Inhibin, beta A (activin A, activin AB alpha polypeptide)
NCBI/Entrez 3624
HGNC 6066
UniProt/Swiss-Prot/ UniProt/TrEMBL P08476 Q5HYA2 Q53H39
Ensembl ENSG00000122641
OMIM NA
GeneCards GC07M041695
Synonyms: Activin beta-A chain, EDF, Erythroid differentiation protein, FRP, Inhibin beta A chain precursor
Activin beta-A, Activin-A;  Homo sapiens
Activin-A/EDF, a TGF-beta family member, is the homeodimer of the INHBA gene product inhibin/activin beta-A chain. The human gene INHBA (map locus 7p15-P13; Ensembl 7p14.1) gene product is a 426 AA long (47.4 kDa) precursor protein that contains a 20 AA single sequence (1-20), a 290 AA propeptide sequence (21 to 310) and a 116 AA inhibin/activin subunit (311 to 426). The subunit contains four potential disulfide intra-chain bridges (314-322; 321-391; 350-423; 354-425) and a potential interchain disulfide bridge at Cys390. Activin A and TGF-beta 1 are structurally related disulfide-linked homodimers synthesized as large precursors. The pro-regions are required for proper folding, disulfide bonding and export of the homodimers, Gray AM and Mason AJ. (1990).
Activin-A participates in a diverse array of functions that include; hypothalamic/pituitary/gonadal hormone secretion, insulin secretion; cell growth, differentiation and survival (apoptosis); embryonic patterning and development; wound healing; and inflammation/immune response. It was initially identified as a follicle stimulating hormone/FSH-releasing protein (gonadal hormone), Ling N, et al. (1986) and erythroid differentiation factor (EDF), Schwall R, et al. (1989). The FSH activity is linked to an increase in the population of pituitary gonadotrophs by Katayama T, et al. (1990). In addition to gonadotrophs, activin-A also modifies somatotrophs and lactotrophs, Kitaoka, et al. (1988).
Activin-A stimulates glycogenolysis in isolated rat hepatocytes, Mine T ET. al. (1989) and elevates insulin release from rat pancreatic islets, Verspohl EJ et al. (1993). Activin-A stimulates insulin secretion in rat pancreatic islets, Totsuka Y, et al. (1988).
Follistatin, an activin-binding protein, is a principle regulator of activin activity, de Winter JP, et al. (1996); however during embryogenesis, Cripto is an important noncompetitive activin antagonist that facilitates Nodal signaling, Kelber JA, et al. (2008).
A role for activin-A as a regulator of cell proliferation was recognized by Gonzalez-Manchon C and Vale W. (1989); wherein Act-A inhibited the growth of and induced morphological changes in CHO-KI cells in culture, in a way similar to but less potent than TGF-beta.
Activin A is involved with the entire process of embryo development from germ cells thru embryonic development to adult tissues. It stimulates spermatogonial proliferation in germ-Sertoli cell cocultures, Mather JP, et al. (1990) and is a maturation factor for oocytes, Itoh M, et al. (1990). Activin A promotes proliferation of human luteinized preovulation granulose cells (ovarian granulose cells), Rabinovici J, et al. (1990).
In the preimplantation embryo, Activin-A stimulates the rate of morula formation and the velocity of embryo cleavage, Orimo, T, et al. (1996). Recombinant activin A stimulates development of one-cell embryos to morula and blastocyst stages in vitro, Yoshioka K, and Kamonae H. (1996).
Activin-A and -B are both potent vegetizing factors. Activin-A was suspected as a blastula level mesoderm-inducing factor (MIF) when it was identified as a homologue of Xenopus XTF-MIF, Smith JC, et al. (1990); van den Eijnden-Van Raaij AJ, et al. (1990); Albano RM, et al. (1990). Thomsen G, et al. (1990) reported that Activin-B transcripts appear earlier than Activin-A transcripts in Xenopus development. Beta-B was first detected in the blastula, whereas beta-A did not appear until late gastrula stage. They suggested that early MIF and axial patterning was due to Activin-B not A. Asashima M, et al. (1991) reported that activin is present in unfertilized Xenopus eggs and blastulae, but this may be of maternal origin. They agree with Thomsen that newly synthesized A appears in late gastrula stage. Activin-A mRNA was not detected in early post-implantation embryo, but was found in nearby maternal decidual cells, Manova K, et al. (1992) and placental cells, Rabinovici J, et al. (1992). Restricted expression of activin-A appears in the gastrula-stage of embryonic development, but the pre-implantation and post-implantation embryos may be exposed to maternally-derived activin A. Activin-A is expressed in both the oviducts, Ganfolfi F, et al. (1995) and decidual cells in the endometrium, Jones RL, et al. (2002).
Activin-A is a critical factor in germ layer and cell lineage specification during gastrulation. Gastrulation is the time when cells of the epiblast (primitive ectoderm) differentiate from totipotent to specific lineage precursor cells. Activin-A plays an important role in this transition. Embryonic stem cells (ES) derived from the inner cell mass represent a model for epiblast and germ layer differentiation.
Activin is involved in the regulation of both self-renewal and differentiation depending upon cell context. Activin-A induces and/or activates many genes involved with embryonic development including Notch, Delta-1, Delta-2 (mesodermal inducing), Abe T, et al. (2004); Nodal, lefty-A and lefty B (via Smad2/3 activation induced genes), Besser D. (2004); and Oct4, Nanog, Wnt3, bFGF and FGF8, Xiao L, et al. (2006).
Activin/Nodal signaling supports the proliferation (mitosis) of embryonic stem cells in culture; wherein these factors are derived via endogenously active autocrine loops, Ogawa K et al. (2007). In culture, endogenously activated autocrine loops of Activin/Nodal signaling supports propagation of mESC, Ogawa K et al. (2007). Activin-A is necessary and sufficient for the maintenance of self-renewal and pluripotency, Xiao L. et al. (2006). Activin-A maintains pluripotency (stemness) of human embryonic stem cells in culture, Beattie GM, et al. (2005); Valdimarsdottir G and Mummery C. (2005); however, under appropriate conditions it promotes germ layer and tissue differentiation.
ES transition from pluripotent to germ layer depends upon at least two conditions: Activin/Nodal signaling and release from IGF/insulin inhibition generated thru PI3K signaling, McLean AB, et al. (2007). In undifferentiated hESC, TGFbeta/activin/nodal/smad2/3 signaling pathway activity is necessary for maintenance of pluripotency, James D, et al. (2005) and the FGF, Wnt and Hedgehog (Hh) signaling pathways are active, Xiao L et al. (2006). The suppression of induction of differentiation of hESC into mesendoderm and endoderm by Activin-A and Nodal depends upon the activation of the phosphatidylinositol 3-kinase pathway (PI3K) by factors such as IGF or insulin; when this pathway is inactive differentiation proceeds, McLean AB, et al. (2007). BMP and Notch promote differentiation.
The mesendoderm, endoderm and tissue specification activities of Activin-A can be demonstrated in vitro using embryonic stem cells (ESC) and cell lineage precursor cells. Tada S, et al. (2005) demonstrated that activin A could induce a phenotypically defined (Gsc-GFP+E-cadherin(ECD)+PDFGRalpha(alphaR)+ population) mesendoderm from ESC in culture. Activin-A can induce embryonic stem cells within embryoid bodies (EB) to form definitive endoderm (DE) (markers Sox17, CXCR4, Goosecoid and FoxA2) in serum-free culture, Kubo A. et al. (2004); D’Amour KA, et al. (2005). Mouse embryonic stem cells can be induced to form dorsoanterior-like mesoderm by Activin A in chemically defined medium, Johansson BM and Wiles MV. (1995).
Activin or Nodal direct nascent mesoderm towards axial mesoderm and mesendoderm; FGF signaling supports the epithelial-to-mesenchymal transition of new mesoderm (increased expression of axial mesoderm markers). The mesoderm inducing activity of Activin-A requires FGF-2 (basic FGF), Cornell RA and Kimelman D. (1994). FGF-dependent Activin/Nodal signaling also supports anterior visceral endoderm (aVE) formation, Willems E and Leyns L. (2008). Doral mesoderm induction by activin-A is modified by the Wnt family factor Xwnt8, Sokol SY, et al. (1992).
Activin-A is an important regulator of early embryonic organogenesis. Activin-A directs differentiation of definitive endoderm (DE) derivatives towards neuronal- and glial-like cell-types, McKiernan E, et al. (2007). Activin-A inhibits chondrogenic differentiation during chick limb bud formation, Chen P, et al. (1993), but it induces Shh-dependent differentiation of intestinal tissue from embryonic pancreas, van Eyll JM, et al. (2004). Activin-A is an essential early mesenchymal signal in tooth development, Ferguson CA, et al. (1998) and metanephric mesenchyme during kidney development, Maeshima A, et al. (3003). Activin-A, along with FGF-2, BMP4 and VEGF (VegfA) supports differentiation of hematopoeitic progenitor cells from ES cells. BMP4 promotes formation of mesoderm. Basic FGF and activin-A induce differentiation of these mesoderm precursors to hemangioblast fate (Runx1, Scl (Tal1) and Hhex expression is upregulated within 3 hours of stimulation; whereas upregulation of Lmo2 and Fli1 is observed later and VEGF stimulates commitment to hematopoeitic progenitors, Pearson S, et al. (2008).
Activin-A is a regulator of cell differentiation in adult tissues. It is involved with the regulation of differentiation of multiple cell types including: cardiac myogenesis, Sugi Y and Lough J, (1995); cytotrophoblasts, Caniggia I et al. (1997); keratinocytes, Seishima M, et al. (1999); osteoclasts, Fuller K, et al. (2000) and Sakai R, et al. (1993); astrocytes, Satoh M, et al. (2000); and erythroid cells, Boxmeyer HE, et al. (1988). Its ability to induce fibroblasts to differentiate into myofibroblasts Ohga E, et al. (1996) suggests a role in tissue repair processes and potential fibrotic diseases, Hubner G, et al. (1999); Munz B, et al. (2001); Bamberger C, et al. (2005).
Activin-A can induce apoptosis in specific cell lines including B-cells, Nishihara T, et al. (1993, 1995), and murine hepatocytes, Hully JR, et al. (1994). Activin-A induces apoptosis in BAX expressing cell lines. Activin-A apoptosis is suppressed by Bcl-2 and BcL-XL upregulation, Koseki T, et al. (1995, 1998). Smad7, but not Smad6 is an activin A-inducible antagonist of activin A-induced growth arrest and apoptosis in HS-72 cells, Ishisaki A, et al. (1999).
Sigma offers antibodies, shRNAs and other products useful for the study of the Activin beta-A gene.
References:
Abe T, et al. (2004) Activin-like signaling activates Notch signaling during mesodermal induction. Int J Dev Biol. 48: 327-332.
Asashima M, et al. (1991) Presence of activin (erythroid differentiation factor) in unfertilized eggs and blastulae of Xenopus laevis. Proc Natl Acad Sci U S A. 88: 6511-6524.
Albano RM, et al. (1990) A mesoderm-inducing factor produced by WEHI-3 murine myelomonocytic leukemia cells is activin A. Development. 110: 435-443.
Bamberger C, et al. (2005) Activin controls skin morphogenesis and wound repair predominantly via stromal cells and in a concentration-dependent manner via keratinocytes. Am J Pathol. 167: 733-747.
Beattie GM, et al. (2005) Activin A maintains pluripotency of human embryonic stem cells in the absence of feeder layers. Stem Cells. 23: 489-495.
Besser D. (2004) Expression of nodal, lefty-a, and lefty-B in undifferentiated human embryonic stem cells requires activation of Smad2/3. J Biol Chem. 279: 45076-45084.
Broxmeyer HE, et al. (1988) Selective and indirect modulation of human multipotential and erythroid hematopoietic progenitor cell proliferation by recombinant human activin and inhibin. Proc Natl Acad Sci U S A. 85: 9052-9056.
Caniggia I, et al. (1997) Activin is a local regulator of human cytotrophoblast cell differentiation. Endocrinology. 138: 3976-3986.
Chen P, et al. (1993) Chondrogenesis in chick limb bud mesodermal cells: reciprocal modulation by activin and inhibin. Exp Cell Res. 206: 119-127.
Cornell RA and Kimelman D. (1994) Activin-mediated mesoderm induction requires FGF. Development. 120: 453-462.
de Winter JP, et al. (1996) Follistatins neutralize activin bioactivity by inhibition of activin binding to its type II receptors. Mol Cell Endocrinol. 116: 105-114.
D'Amour KA, et al. (2005) Efficient differentiation of human embryonic stem cells to definitive endoderm. Nat Biotechnol. 23: 1534-1541.
Ferguson CA, et al. (1998) Activin is an essential early mesenchymal signal in tooth development that is required for patterning of the murine dentition. Genes Dev. 12: 2636-2649.
Fuller K, et al. (2000) Activin A is an essential cofactor for osteoclast induction. Biochem Biophys Res Commun. 268: 2-7.
Gandolfi F, et al. (1995) Activin beta A subunit is expressed in bovine oviduct. Mol Reprod Dev. 40: 286-291.
Gonzalez-Manchon C and Vale W. (1989) Activin-A, inhibin and transforming growth factor-beta modulate growth of two gonadal cell lines. Endocrinology. 125: 1666-1672.
Gray AM and Mason AJ. (1990) Requirement for activin A and transforming growth factor--beta 1 pro-regions in homodimer assembly. Science. 247: 1328-1330.
Hübner G, et al. (1999) Activin: a novel player in tissue repair processes. Histol Histopathol. 14: 295-304.
Hully JR, et al. (1994) Induction of apoptosis in the murine liver with recombinant human activin A. Hepatology. 20: 854-862.
Ishisaki A, et al. (1999) Differential inhibition of Smad6 and Smad7 on bone morphogenetic protein- and activin-mediated growth arrest and apoptosis in B cells. J Biol Chem. 274: 13637-13642.
Itoh M, et al. (1990) Activin A stimulates meiotic maturation of the rat oocyte in vitro. Biochem Biophys Res Commun. 166: 1479-1484.
James D, et al. (2005) TGFbeta/activin/nodal signaling is necessary for the maintenance of pluripotency in human embryonic stem cells. Development. 132: 1273-1282.
Johansson BM and Wiles MV. (1995) Evidence for involvement of activin A and bone morphogenetic protein 4 in mammalian mesoderm and hematopoietic development. Mol Cell Biol. 15: 141-151.
Jones RL, et al. (2002) Activin A promotes human endometrial stromal cell decidualization in vitro. J Clin Endocrinol Metab. 87: 4001-4004.
Katayama T, et al. (1990) Activin A increases the number of follicle-stimulating hormone cells in anterior pituitary cultures. Mol Cell Endocrinol. 69: 179-185.
Kelber JA, et al. (2008) Cripto is a noncompetitive activin antagonist that forms analogous signaling complexes with activin and nodal. J Biol Chem. 283: 4490-4500.
Kitaoka M, et al. (1988) Activin-A: a modulator of multiple types of anterior pituitary cells. Biochem Biophys Res Commun. 157: 48-54.
Koseki T, et al. (1998) Correlation between Bcl-X expression and B-cell hybridoma apoptosis induced by activin A. Cell Signal. 10: 517-521.
Koseki T, et al. (1995) Activin A-induced apoptosis is suppressed by BCL-2. FEBS Lett. 376: 247-250.
Kubo A, et al. (2004) Development of definitive endoderm from embryonic stem cells in culture. Development. 131: 1651-1662.
Ling N, et al. (1986) A homodimer of the beta-subunits of inhibin A stimulates the secretion of pituitary follicle stimulating hormone. Biochem Biophys Res Commun. 138: 1129-1137.
Maeshima A, et al. (2003) Activin a produced by ureteric bud is a differentiation factor for metanephric mesenchyme. J Am Soc Nephrol. 14: 1523-1534.
Manova K, et al. (1992) Expression of activins and TGF beta 1 and beta 2 RNAs in early postimplantation mouse embryos and uterine decidua. Mech Dev. 36: 141-152.
Mather JP, et al. (1990) Activin stimulates spermatogonial proliferation in germ-Sertoli cell cocultures from immature rat testis. Endocrinology. 127: 3206-3214.
McKiernan E, et al. (2007) Directed differentiation of mouse embryonic stem cells into pancreatic-like or neuronal- and glial-like phenotypes. Tissue Eng. 13: 2419-2430.
McLean AB, et al. (2007) Activin a efficiently specifies definitive endoderm from human embryonic stem cells only when phosphatidylinositol 3-kinase signaling is suppressed. Stem Cells. 25: 29-38.
Mine T, et al. (1989) Stimulation of glucose production by activin-A in isolated rat hepatocytes. Endocrinology. 125: 586-591.
Munz B, et al. (2001) The roles of activins in repair processes of the skin and the brain. Mol Cell Endocrinol. 180: 169-177.
Nishihara T, et al. (1995) Induction of apoptosis in B lineage cells by activin A derived from macrophages. J Interferon Cytokine Res. 15: 509-516.
Nishihara T, et al. (1993) Activin A induces apoptotic cell death. Biochem Biophys Res Commun. 197: 985-991.
Ohga E, et al. 1996) Effects of activin A on proliferation and differentiation of human lung fibroblasts. Biochem Biophys Res Commun. 228: 391-396.
Ogawa K, et al. (2007) Activin-Nodal signaling is involved in propagation of mouse embryonic stem cells. J Cell Sci. 120: 55-65.
Orimo T, et al. (1996) The effect of activin-A on the development of mouse preimplantation embryos in vitro. J Assist Reprod Genet. 13: 669-674.
Pearson S, et al. (2008) The stepwise specification of embryonic stem cells to hematopoietic fate is driven by sequential exposure to Bmp4, activin A, bFGF and VEGF. Development. 135: 1525-1535.
Rabinovici J, et al. (1992) Localization and regulation of the activin-A dimer in human placental cells. J Clin Endocrinol Metab. 75: 571-576.
Sakai R, et al. (1993) Activin enhances osteoclast-like cell formation in vitro. Biochem Biophys Res Commun. 195: 39-46.
Satoh M, et al. (2000) Activin promotes astrocytic differentiation of a multipotent neural stem cell line and an astrocyte progenitor cell line from murine central nervous system. Neurosci Lett. 284: 143-146.
Schwall R, et al. (1989) Multiple actions of recombinant activin-A in vivo. Endocrinology. 125: 1420-1423.
Seishima M, et al. (1999) Activin A induces terminal differentiation of cultured human keratinocytes. J Invest Dermatol. 1999 Apr;112(4):432-6.
Sokol SY and Melton DA. (1992) Interaction of Wnt and activin in dorsal mesoderm induction in Xenopus. Dev Biol. 154: 348-355.
Smith JC, et al. (1990) Identification of a potent Xenopus mesoderm-inducing factor as a homologue of activin A. Nature. 345: 729-731.
Sugi Y and Lough J. (1995) Activin-A and FGF-2 mimic the inductive effects of anterior endoderm on terminal cardiac myogenesis in vitro. Dev Biol. 168: 567-574.
Tada S, et al. (2005) Characterization of mesendoderm: a diverging point of the definitive endoderm and mesoderm in embryonic stem cell differentiation culture. Development. 132: 4363-4374.
Thomsen G, et al. (1990) Activins are expressed early in Xenopus embryogenesis and can induce axial mesoderm and anterior structures. Cell. 63: 485-493.
Totsuka Y, et al. (1988) A novel action of activin A: stimulation of insulin secretion in rat pancreatic islets. Biochem Biophys Res Commun. 156: 335-339.
Valdimarsdottir G and Mummery C. (2005) Functions of the TGFbeta superfamily in human embryonic stem cells. APMIS. 113: 773-789.
van Eyll JM, et al. (2004) Shh-dependent differentiation of intestinal tissue from embryonic pancreas by activin A. J Cell Sci. 117: 2077-2086.
Verspohl EJ, et al. (1993) Activin A: its effects on rat pancreatic islets and the mechanism of action involved. Life Sci. 53: 1069-1078.
Willems E and Leyns L. (2008) Patterning of mouse embryonic stem cell-derived pan-mesoderm by Activin A/Nodal and Bmp4 signaling requires Fibroblast Growth Factor activity. Differentiation. 2008 Jan 2. [Epub ahead of print].
Yoshioka K and Kamomae H. (1996) Recombinant human activin A stimulates development of bovine one-cell embryos matured and fertilized in vitro. Mol Reprod Dev. 45: 151-156.
Xiao L, et al. (2006) Activin A maintains self-renewal and regulates fibroblast growth factor, Wnt, and bone morphogenic protein pathways in human embryonic stem cells. Stem Cells. 24: 1476-1486.
Footnote: Gene Data Sources: HGNC, Entrez Gene, UniProt/Swiss-Prot, UniProt/TrEMBL, GDB, OMIM, GeneLoc, Ensembl.
Copyright © 2008 Sigma-Aldrich Co. Reproduction forbidden without permission.
Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. Best viewed in IE6 or higher.