Your Favorite Gene - Powered By Ingenuity
www.sigmaaldrich.com/stemcell
Sigma-Aldrich
Your Favorite Gene - Powered By Ingenuity
Find Dax1 Products
Gene NROB1; NR0B1_HUMAN
nuclear receptor subfamily 0, group B, member 1
NCBI/Entrez 190
HGNC 7960
UniProt/Swiss-Prot/ UniProt/TrEMBL P51843 Q16392 Q96F69
Ensembl ENSG00000169297
OMIM 300200, 300018
GeneCards GC0XM030232
Synonyms: AHC, AHCH, AHX, DAX1, DAX-1, DSS, DSS-AHC critical region on the X chromosome protein 1, GTD, HHG, NROB1, Nuclear receptor 0B1, Nuclear receptor DAX-1
DAX-1/Nuclear receptor subfamily 0, group B, member 1 (Gene NROB1) Homo sapiens
The X-chromosome-linked NROB1 gene (map locus Entrez: Xp21.3-p21.2; Ensembl: Xp21.2; HGNC: Xp21.3) product, nuclear receptor DAX-1 (dosage-sensitive sex reversal, adrenal hypoplasia critical region, on chromosome X, gene 1), is translated as two splice variants. NROB1A and NROB1 are 400 amino acids long, Ho J, et al. (2004) and 470 amino acids long (51.7 kDa), respectively. NROB1A is the more abundant form in most tissues except testis. It results from the removal of the C-terminal 70 amino acids and changes in the 10 amino acids of the new C-terminal (390 to 400). NROB1A lacks the AF-2 binding motif found in the long form. Both variants contain three LXXLL motifs, ligand-binding domains (LBD) and 4 N-terminal repeats that are believed to bind DNA stem loop structures in lieu of zinc finger DNA-binding domains characteristic of other nuclear receptors. DAX1 variants exist as homodimers and heterodimers with variant partnering (NROB1:NROB1A) and with SHP (NROB1:NROB2); consequently these gene products are complex and will require extensive new studies to fully define their roles, McCabe ER. (2007).
DAX-1 is expressed in tissue of the HPAG-axis including; the hypothalamus, pituitary, adrenal cortex and gonads. DAX-1 expression in a human adrenocortical carcinoma line, NC1-H295, with fetal adrenal cortex-like features suggested that it may have a role in steroidogenesis , Guo W, et al. (1995). It is now known that DAX-1 along with its network proteins (Nanog, SF-1, Wt1) is also expressed in totipotent embryonic stem cells, and preimplantation embryos, Chipsham R, et al. (2004); Niakan KK, et al. (2005).
DAX1, AHCH, was initially discovered as a component of the cascade that regulates development of the hypothalamic-pituitary-adrenal-gonadal axis (HPAG axis). DAX-1 acts as a dominant transcription repressor in hormone-producing tissues of the HPAG axis. It coregulates the embryonic development of these tissues and hormone production in formed tissues. NROB1 along with WT1, SOX9, ATRX and SRY, has been identified as a sex-determining gene, in human sex reversal, XY females and XX males, McElreavey K and Fellous M. (1997); Kojima, Y, et al. (2008).
DAX-1 has emerged as a negative coregulator of genes involved in the HPAG axis and maintenance of pluripotency. The growing list includes estrogen receptor (ER, NR3A1-2); liver receptor homologue-1 (LRH-1); steroidogenesis factor, (NR5A1, SF-1); NR5A2, androgen receptor (AR, NR3C4); progesterone receptor (PR, NR3C3), Iyer AK, et al. (2004) and peroxisome proliferators-activated receptor (PPARgamma), Kim GS, et al. (2008).
DAX-1 represses transcription of genes that are regulated by steroidogenic factor-1 (SF-1, NR5A1), Suzuki T, (2003), an orphan nuclear receptor required for development of the urogenital ridge, adrenal glands and gonads, Ito M, et al. (1997). DAX-1 acts as an adaptor molecule that recruits corepressors such as N-CoR to SF-1, Crawford PA, et al. (1998). Contrary to its repressor function, DAX1 and SF-1 have been reported to function cooperatively to mediate somatic cell differentiation during testis development, Park SY, (2005).
Mutations in NROB1 cause X-linked cytomegalic form of adrenal hypoplasia congenita (AHC). AHC results in adrenal gland structural disorder, congenital adrenal hypoplasia leading to hormone deficiencies and hypogonadotropic hypogonadism (HHG). AHC is lethal if untreated. Duplication of DAX-1 causes dosage sensitive sex reversal (DSS), XY phenotypic females.
An understanding of the role of DAX1 in pluripotent cells is beginning to emerge. DAX1 as a nuclear receptor involved with ES cell pluripotency and differentiation has been recently reviewed, Mullen EM, (2007). DAX1 is a component of the protein interaction network, cellular module, dedicated to pluripotency, Wang J, et al. (2006); Pan G and Thomson JA (2007); Kim J, (2008).
Sigma offers antibodies, shRNAs and other products useful for the study of the Dax1 gene.
References:
Clipsham R, et al. (2004) Nr0b1 and its network partners are expressed early in murine embryos prior to steroidogenic axis organogenesis. Gene Expr Patterns. 4: 3-14.
Crawford PA, et al. (1998) Nuclear receptor DAX-1 recruits nuclear receptor corepressor N-CoR to steroidogenic factor 1. Mol Cell Biol. 18: 2949-56.
Guo W, et al. (1995) Expression of DAX-1, the gene responsible for X-linked adrenal hypoplasia congenita and hypogonadotropic hypogonadism, in the hypothalamic-pituitary-adrenal/gonadal axis. Biochem Mol Med. 56: 8-13.
Ho J, et al. (2004) NR0B1A: an alternatively spliced form of NR0B1. Mol Genet Metab. 83: 330-6.
Ito M, et al. (1997) DAX-1 inhibits SF-1-mediated transactivation via a carboxy-terminal domain that is deleted in adrenal hypoplasia congenita. Mol Cell Biol. 17: 1476-83.
Iyer AK and McCabe ER. (2004) Molecular mechanisms of DAX1 action. Mol Genet Metab. 83: 60-73.
Kim J, et al. (2008) An extended transcriptional network for pluripotency of embryonic stem cells. Cell. 132: 1049-61.
Kim GS, et al. (2008) The orphan nuclear receptor DAX-1 acts as a novel transcriptional corepressor of PPARgamma. Biochem Biophys Res Commun. 370: 264-8.
Kojima Y, et al. (2008) Up-regulation of SOX9 in human sex-determining region on the Y chromosome (SRY)-negative XX males. Clin Endocrinol (Oxf). 68: 791-9.
McElreavey K and Fellous M. (1997) Sex-determining genes. Trends Endocrinol Metab. 8: 342-6.
McCabe ER. (2007) DAX1: Increasing complexity in the roles of this novel nuclear receptor. Mol Cell Endocrinol. 265-266: 179-82.
Mullen EM, et al. (2007) Nuclear Receptors in Regulation of Mouse ES Cell Pluripotency and Differentiation. PPAR Res. 2007: 61563.
Niakan KK and McCabe ER. (2005) DAX1 origin, function, and novel role. Mol Genet Metab. 86: 70-83.
Pan G and Thomson JA. (2007) Nanog and transcriptional networks in embryonic stem cell pluripotency. Cell Res. 17: 42-9.
Park SY, et al. (2005) Nuclear receptors Sf1 and Dax1 function cooperatively to mediate somatic cell differentiation during testis development. Development. 132: 2415-23.
Suzuki T, et al. (2003) LXXLL-related motifs in Dax-1 have target specificity for the orphan nuclear receptors Ad4BP/SF-1 and LRH-1. Mol Cell Biol. 23: 238-49.
Wang J, et al. (2006) A protein interaction network for pluripotency of embryonic stem cells. Nature. 444: 364-8.
Footnote: Gene Data Sources: HGNC, Entrez Gene, UniProt/Swiss-Prot, UniProt/TrEMBL, GDB, OMIM, GeneLoc, Ensembl.
Copyright © 2008 Sigma-Aldrich Co. Reproduction forbidden without permission.
Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. Best viewed in IE6 or higher.