Your Favorite Gene - Powered By Ingenuity
www.sigmaaldrich.com/stemcell
Sigma-Aldrich
Your Favorite Gene - Powered By Ingenuity
Find EN1 Products
Gene EN1: HME1_HUMAN
Homeobox protein engrailed-1
NCBI/Entrez 2019
HGNC 3342
UniProt/Swiss-Prot/ UniProt/TrEMBL Q05925 Q4ZG44
Ensembl ENSG00000163064
OMIM 131290
GeneCards GC02M119315
Synonyms: Homeobox protein engrailed-1, Hu-En-1
Engrailed homolog 1 (Gene En1)Homo sapiens
The EN1 (map locus: Ensembl/HGNC: 2q14.2; Entrez: 2q13-q21, Köhler A, et al. (1993)) gene product, homeobox protein engrailed-1, is a 392 AA (40.1 kDa) homeobox transcription factor that contains a 60 AA (303-362) DNA-binding homeobox, a 20 AA (199 to 218) polyAla region and an 8 AA (224 to 231) polyGly region.
Homeobox transcription factor engrailed-1 is involved in the regionalization of the central nervous system (CNS) and spinal cord and support of neuron function and survival. It is also essential for the patterning of limb development, Loomis CA, et al. (1996), aspects of craniofacial bone development, Deckelbaum RA, et al. (2006) and trunk skeletal muscle development, Ahmed MU, et al. (2006).
Dysfunction of regulation or mutations in engrailed-1 may be linked to major diseases such as Autism, Kuemerle B, et al. (2007) and Parkinson’s diseases, Simon HH, et al. (2001).
En1 is the earliest molecular marker of midbrain tissue, Shamim H. et al. (1999) and it is an essential gene for midbrain development and regionalization. It is expressed in all neuronal groups of the medulla and throughout the cerebellum including cerebellar granule cell layers, white matter of the vermis and flocculus, inferior olive, arcuate nucleus, caudal raphe nuclei, corpus pontobulbare and nucleus ambiguus, Zec N, et al. (1997). The role of En-1 in cerebellar development has been discussed recently by Murcia CL, et al. (2007). En-1 is also involved in regionalization of the optic tectum, Nakamua H, et al. (2001).
En-1 may play an important role in understanding and future therapies for Parkinson’s disease. En-1 is involved in generation and/or survival of mid-hindbrain precursor cells, Wurst W, et al. (1994) and it is highly expressed by essentially all dopaminergic neurons in the substantia nigra compacta (SNc) and ventral tegmentum, Simon HH, et al. (2001). En-1 and En-2 regulate expression of alpha-synuclein, a gene that is genetically linked to Parkinson's disease. Engrailed genes are expressed by midbrain dopaminergic neurons from their generation to adulthood but are not required for their specification. Engrailed genes control the survival of midbrain dopaminergic neurons in a gene dose-dependent manner, Simon HH, et al. (2001). Engrailed genes are cell-autonomously required to prevent apoptosis in mesencephalic dopaminergic neurons, Alberi L, et al. (2004), Simon HH, et al. (2004). It is especially important to note that En-1 expression and survival effects on dopaminergic neurons of the substantia nigra (SN) and ventral tegmental area (VTA) persist in adults, Sonnier L, et al. (2007) and are neural activity dependent, Di Nardo AA, et al. (2007). The loss of En-1 is linked to apoptosis of midbrain dopaminergic neurons, a condition of Parkinson’s disease.
EN-1 is coexpressed with Pax2 within a population of spinal cord ventral interneurons that are directed by Pax6, Burrill JD, et al. (1997), but it is not required for the early specification of ventral interneuron cell types in the embryonic spinal cord, Saueressig H, et al. (1999). En-1 has been identified as a regulator of pathfinding and fasciculation during axon growth, Saueressig H, et al. (1999). En1 expressing spinal interneurons represent a heterogeneous population that connects with both motoneurons (primarily GABAergic) and sensory afferents, Wenner P, et al. (2000). En1 along with Pax6 have essential roles in establishing recurrent inhibitory circuits between motoneurons and Renshaw cells, Sapir T, et al. (2004). Renshaw cells, the interneuronal cell type that mediates recurrent inhibition of motor neurons, are derived from a subset of spinal V1 interneurons.
Engrailed-1 is expressed in embryonic ventral limb ectoderm and is essential for dorsal-ventral and proximal-distal limb patterning, Loomis, et al. (1996) and for the proper formation of the apical ectodermal ridge (AER). En1 plays a role in the compaction of the AER into a narrow rim at the tip of the limb, Loomis CA, et al. (1998). En1 is involved in the coordination of AER formation, wherein it is required for positioning and maintenance of two lineage borders, Kimmel RA, et al. (2000); one border is transient and corresponds to the D/V midline of the AER, and the second border corresponds to the dorsal AER margin.
Sigma offers antibodies and shRNAs useful for the study of EN1 gene products.
References:
Ahmed MU, et al. (2006) Establishment of the epaxial-hypaxial boundary in the avian myotome. Dev Dyn. 235: 1884-1894.
Albéri L, et al. (2004) Engrailed genes are cell-autonomously required to prevent apoptosis in mesencephalic dopaminergic neurons. Development. 131: 3229-3236.
Burrill JD, et al. (1997) PAX2 is expressed in multiple spinal cord interneurons, including a population of EN1+ interneurons that require PAX6 for their development. Development. 124: 4493-4503.
Deckelbaum RA, et al. (2006) The homeoprotein engrailed 1 has pleiotropic functions in calvarial intramembranous bone formation and remodeling. Development. 133: 63-74.
Di Nardo AA, et al. (2007) Dendritic localization and activity-dependent translation of Engrailed1 transcription factor. Mol Cell Neurosci. 35: 230-236.
Köhler A, et al. (1993) Regional assignment of the human homeobox-containing gene EN1 to chromosome 2q13-q21. Genomics. 15: 233-235.
Kuemerle B, et al. (2007) The mouse Engrailed genes: a window into autism. Behav Brain Res. 176: 121-32.
Loomis CA, et al. (1998) Analysis of the genetic pathway leading to formation of ectopic apical ectodermal ridges in mouse Engrailed-1 mutant limbs. Development. 125: 1137-1148.
Loomis CA, et al. (1996) The mouse Engrailed-1 gene and ventral limb patterning. Nature. 382: 360-363.
Murcia CL, et al. (2004) Dissecting complex genetic interactions that influence the Engrailed-1 limb phenotype. Mamm Genome. 15: 352-360.
Nakamura H. (2001) Regionalisation and acquisition of polarity in the optic tectum. Prog Neurobiol. 65: 473-488.
Sapir T, et al. (2004) Pax6 and engrailed 1 regulate two distinct aspects of renshaw cell development. J Neurosci. 2004 Feb 4;24(5): 1255-1264.
Saueressig H, et al. (1999) Engrailed-1 and netrin-1 regulate axon pathfinding by association interneurons that project to motor neurons. Development. 126: 4201-4212.
Shamim H, et al. (1999) Sequential roles for Fgf4, En1 and Fgf8 in specification and regionalisation of the midbrain. Development. 126: 945-959.
Simon HH, et al. (2004) Midbrain dopaminergic neurons: control of their cell fate by the engrailed transcription factors. Cell Tissue Res. 318: 53-61.
Simon HH, et al. (2001) The Fate of midbrain dopaminergic neurons controlled by the engrailed genes. J Neurosci. 21: 3126-3134.
Sonnier L, et al. (2007) Progressive loss of dopaminergic neurons in the ventral midbrain of adult mice heterozygote for Engrailed1. J Neurosci. 27: 1063-1071.
Wenner P, et al. (2000) Topographical and physiological characterization of interneurons that express engrailed-1 in the embryonic chick spinal cord. J Neurophysiol. 84: 2651-2657.
Wurst W, et al. (1994) Multiple developmental defects in Engrailed-1 mutant mice: an early mid-hindbrain deletion and patterning defects in forelimbs and sternum. Development. 120: 2065-2075.
Zec N, et al. (1997) Expression of the homeobox-containing genes EN1 and EN2 in human fetal midgestational medulla and cerebellum. J Neuropathol Exp Neurol. 56: 236-242.
Footnote: Gene Data Sources: HGNC, Entrez Gene, UniProt/Swiss-Prot, UniProt/TrEMBL, GDB, OMIM, GeneLoc, Ensembl.
Copyright © 2008 Sigma-Aldrich Co. Reproduction forbidden without permission.
Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. Best viewed in IE6 or higher.