Your Favorite Gene - Powered By Ingenuity
www.sigmaaldrich.com/stemcell
Sigma-Aldrich
Your Favorite Gene - Powered By Ingenuity
Find FoxD3 Products
Gene FoxD3; FOXD3_HUMAN
Forkhead Box D3
NCBI/Entrez 27022
HGNC 3804
UniProt/Swiss-Prot/ UniProt/TrEMBL Q9UJU5 Q9BYM2 Q9UDD1
Ensembl ENSG00000187140
OMIM 611539
GeneCards GC01P063500
Synonyms: Forkhead box protein D3, Genesis, HFH2, HNF3/FH transcription factor genesis.
Forkhead Box D3 (Gene FoxD3) Homo sapiens
The intronless FoxD3 gene (map locus Entrez and HGNC: 1p32-p31; Ensembl: 1p31.3) product, forkhead box D3/HNF3/Genesis/FoxD3, is a 478 amino acids long (47.6 kDa) transcription factor that contains a fork-head DNA binding domain (141-235) and five poly-A regions (from 6 to 14 AA long) in the C-terminal half of the protein. The DNA consensus binding sequence is 5'-A[AT]T[AG]TTTGTTT-3'.
FoxD3/HFH2 was initially identified as a member of the winged helix (formerly HNF-3/Forkhed (HFH)) transcription family in embryonal carcinoma, Sutton J, et al. (1996). FoxD3 is expressed in embryonic (ES) stem and embryonic cancer (EC) cells; within the Spemann organizer; in the late-stage gastrula inner cell mass (ICM) (epiblast), Harland R and Gerhart J. (1997); Sutton J, et al. (1996); and in the extra-embryonic tissue (trophectoderm), Tompers DM, et al. (2005). Subsequently, FoxD3 is found in premigrating and migrating neural crest cells, Barembaum M, (2005) and motor-neuron progenitors of the developing spinal cord, Labosky PA and Kaestner KH. (1998); Hromas R, et al. (1999); Dottori M, et al. (2001).
Embryonic neural crest cells are the progenitors of the peripheral nervous system and differentiate into peripheral neurons, glia cells, pigment cells and connective tissues of face, neck and heart. They are induced at the junction of the neural plate and embryonic ectoderm to undergo epithelial to mesenchymal transition (EMT) followed by migration into targeted tissues. FoxD3 along with Slug, Zic5 and Soc9 is considered to be a neural crest marker.
FoxD3 can act as an activator or repressor of transcription depending upon its cell context and binding partners. FoxD3 is essential for survival of the late stage blastocysts, embryonic stem (ES) and teratocarcinomas (EC) cells, Hanna LA, et al. (2002); Teng L, et al. (2008). It is involved in dorsal mesodermal development within the gastrula. FoxD3 functions as a transcriptional corepressor along with Grg4 (Groucho-related gene-4), Yaklichkin S, et al. (2007) in the Spemann organizer where it induces dorsal mesoderm via expression of Nodal and Nodal-related genes (TGFbeta superfamily), Steiner AB, et al. (2006). In close association with Nanog and OCT4 it anchors an interdependent network of transcription factors that regulate stem cell pluripotency, Pan G, et al. (2006).
FoxD3 is involved in early embryonic patterning wherein it participates in the lineage differentiation of neural crest cells. It has been reported to prevent terminal quiescence in primitive neural crest cells by inhibiting p21 expression, Hromas R, et al. (1999). Dottori M, et al. (2001) showed that ectopic expression of FoxD3 in neural tube promoted a neural crest-like phenotype and repression of interneuron differentiation by a Slug and RhoB-independent mechanism. FoxD3 is an upstream regulator of neural crest determination that is required for Slug induction by Zic factors, Sasai N, et al. (2001). FoxD3 is involved in the segregation of neural crest lineage from neuroepithelium and in the repression of melanogenesis, Kos R, et al. (2001). Lister JA, et al. (2006) speculate that Foxd3 may not be essential for induction of neural crest cell identity, but that it may regulate differentiation into specific neural crest cell lineages such as jaw cartilage, peripheral neurons, glia and iridopore pigment cells. FoxD3 repression of neural crest to melanocyte differentiation is blocked by colgate/hdac1 to permit mitfa-dependent melanogenesis to occur, Ignatius MS, et al. (2008).
FoxD3 is involved with the delamination of neural crest cells from the neuroepithelium. Sox9 help protect neural crest cells from apoptosis. Slug/Snail, in the presence of Sox 9 induces epithelial mesenchymal transition (EMT) in neural epithelial cells and FoxD3 regulates expression of cell-cell adhesion molecules in support of crest cell migration, Cheung M, et al. (2005).
Neural crest tissues are induced by BMP, Wnt and FGF signaling. Wnt signaling in neural crest cells leads to co-activation by Pax3 and Zic1, Sato T, et al. (2005) and subsequent upregulation of Lrig3, Zhao H, et al. (2008) followed by FoxD3 along with slug, Pax3, Msx1 and cadherin 6B signaling during the "premigratory" stages of neural crest development, Taneyhill LA, et al. (2005). Over expression of FoxD3 leads to defective neural crest development, Pohl BS and Knochel W. (2001). FoxD3 is autoregulatory self inhibiting.
Sigma offers antibodies and shRNAs useful for the study of FoxD3 gene products.
References:
Barembaum M, and Bronner-Fraser M. (2005) Early steps in neural crest specification. Semin Cell Dev Biol. 16: 642-646.
Cheung M, et al. (2005) The winged-helix transcription factor Foxd3 suppresses interneuron differentiation and promotes neural crest cell fate. Development. 128: 4127-4138.
Dottori M, et al. (2001) Muscle disease caused by mutations in the skeletal muscle alpha-actin gene (ACTA1). Neuromuscul Disord. 13: 519-31.
Hanna LA, et al. (2002) Requirement for Foxd3 in maintaining pluripotent cells of the early mouse embryo. Genes Dev. 16: 2650-2661.
Harland R, and Gerhart J. (1997) Formation and function of Spemann's organizer. Annu Rev Cell Dev Biol. 13: 611-667.
Hromas R, et al. (1999) Genesis, a Winged Helix transcriptional repressor, has embryonic expression limited to the neural crest, and stimulates proliferation in vitro in a neural development model. Cell Tissue Res. 297: 371-382.
Ignatius MS, et al. (2008) colgate/hdac1 Repression of foxd3 expression is required to permit mitfa-dependent melanogenesis. Dev Biol. 313: 568-583.
Kos R, et al. (2001) The winged-helix transcription factor FoxD3 is important for establishing the neural crest lineage and repressing melanogenesis in avian embryos. Development. 128: 1467-1479.
Labosky PA, and Kaestner KH. (1998) The winged helix transcription factor Hfh2 is expressed in neural crest and spinal cord during mouse development. Mech Dev. 76: 185-190.
Lister JA, et al. (2006) Zebrafish Foxd3 is required for development of a subset of neural crest derivatives. Dev Biol. 290: 92-104.
Pan G, et al. (2006) A negative feedback loop of transcription factors that controls stem cell pluripotency and self-renewal. FASEB J. 20: 1730-1732.
Pohl BS, and Knöchel W. (2001) Overexpression of the transcriptional repressor FoxD3 prevents neural crest formation in Xenopus embryos. Mech Dev. 103: 93-106
Sasai N, Mizuseki K, Sasai Y. (2001) Requirement of FoxD3-class signaling for neural crest determination in Xenopus. Development. 128: 2525-2536.
Steiner AB, et al. (2006) FoxD3 regulation of Nodal in the Spemann organizer is essential for Xenopus dorsal mesoderm development. Development. 133: 4827-4838.
Sutton J, et al. (1996) Genesis, a winged helix transcriptional repressor with expression restricted to embryonic stem cells. J Biol Chem. 1996 Sep 20;271(38): 23126-23133.
Taneyhill LA, and Bronner-Fraser M. (2005) Dynamic alterations in gene expression after Wnt-mediated induction of avian neural crest. Mol Biol Cell. 16: 5283-5293.
Teng L, et al. (2008) Requirement for Foxd3 in the maintenance of neural crest progenitors. Development. 135: 1615-1624.
Tompers DM, et al. (2005) Foxd3 is required in the trophoblast progenitor cell lineage of the mouse embryo. Dev Biol. 285: 126-137.
Yaklichkin S, et al. (2007) FoxD3 and Grg4 physically interact to repress transcription and induce mesoderm in Xenopus. J Biol Chem. 282: 2548-2557.
Footnote: Gene Data Sources: HGNC, Entrez Gene, UniProt/Swiss-Prot, UniProt/TrEMBL, GDB, OMIM, GeneLoc, Ensembl.
Copyright © 2008 Sigma-Aldrich Co. Reproduction forbidden without permission.
Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. Best viewed in IE6 or higher.