Your Favorite Gene - Powered By Ingenuity
www.sigmaaldrich.com/stemcell
Sigma-Aldrich
YFG search logo
Find ASCL1 (MASH1) Products
Gene ASCL1: ASCL1_HUMAN
Achaete-scute complex-like 1 (Drosophila)
NCBI/Entrez 429
HGNC 738
UniProt/Swiss-Prot/ UniProt/TrEMBL P50553 Q9BQ30
Ensembl ENSG00000139352
OMIM 209880 100790
GeneCards GC12P101856
Synonyms: Achaete-scute homolog 1, ASH1, HASH1, MASH1
Achaete-scute complex-like 1 (Drosophila) (Gene ASCL1) Homo sapiens
The ASCL1 (map locus: Entrez/HGNC 12q22-q23; Ensembl 12q23.2) gene, MASH1/achaete-scute homolog 1, is a 236 AA (25.4 kDa) basic helix-loop-helix (bHLH) transcription factor with a 40 AA helix-loop-helix motif (132 to 171) and a HLH N-terminal adjacent DNA-binding 11 AA basic motif (121-131). The protein also contains closely linked 15 AA polyAla (33-47) and 12 AA polyGln (51-62) regions. BasicHLH transcription factors are heterodimeric.
Mash1 is essential for neuronal differentiation and specification within the nervous system.
MASH1 (ASCL1) is expressed in temporally and spatially restricted domains of the neuroepithelium of developing CNS and PNS where it helps determine regionalization of the future brain and spinal cord. Mash1 and ngn1 are expressed in adjacent, non-overlapping regions of the neuroepithelium, Ma Q, et al. (1997) and Mash1 and ngn2 are expressed in complementary populations of the CNS and PNS, Parras CM. et al. (2002). Ngn2 and Mash1 function together to regulate the zonal distribution of progenitors in the developing neocortex, Britz O, et al. (2006). Mash1 progenitor cells at early stages in each CNS region preferentially become neurons, and at late stages they become oligodendrocytes, Kim EJ, et al. (2008). Mash1 is required for neurogenesis in the ventral telencephalon and specific areas of the diencephalon, Horton S, et al. (1999), Casarosa S, et al. (1999), Yun K and Fischman S, et al. (2002).
Neural crest cells (NCC) are a migratory population that differentiates into neurons and glia of the peripheral nervous system (the autonomic system) and various non-neural mesectodermal and endocrine cells types. Mash1 plays a variety of roles in the development and speciation of NCC cell progeny, Lo LC, et al. (1991). Mash1 and Phox2b are essential for NCC neuron development into sympathetic, parasympathetic, and enteric ganglia components of the peripheral autonomic nervous system, Pattyn A, et al. (1999). MASH1 is expressed (BMP2-induced) initially in the neural committed non-neural precursor cells of the autonomic nervous system, Anderson DJ (1994), Sommer L, et al. (1995), Anderson DJ, (1997). These MASH1+ precursor cells differentiate further into sympathetic, parasympathetic and enteric neuron subtypes.
Mash1 is involved in the development of a variety of cell types throughout the CNS and PNS.
In the forebrain neocortex, Mash1 promotes the differentiation of GABAergic local circuit neurons. Mash1+ progenitor cells located in the neocortical ventricular and subventricular zone of the dorsal forebrain differentiate into a GABAergic lineage (also Dlx1/2 positive) that represents 65% of the neocortical GABAergic neurons in humans, Letinic K, et al. (2002). Conversely, Mash1 suppresses GABAergic neurons in the hindbrain and spinal cord, Jo AY, et al. (2007). Mash1 is expressed in a subset of oligodendrocyte precursors (OPCs) as soon as they are generated in the forebrain ventricular zone and it cooperates with Olig2 in OPC specification, Parras CM, et al. (2007).
Mash1 supports the division of neural progenitor cells (NPC) from the ventral midbrain (VM) while maintaining neurogenic potential. In cooperation with Nurr1, Mash1 contributes to the generation of dopaminergic (DA) neurons, Park CH, et al. (2006), Kim HJ, et al. (2007). Mash1 function is essential for the development of all central and peripheral neurons that express noradrenergic traits transiently or permanently, Hirsh MR, et al. (1998).
In the hindbrain, Mash1 and Math3 promote branchiomotor neuron development and regulate subsequent oligodendrocyte development, Ohsawa R, et al. (2005). Mash1 is coexpressed with Nkx2-2 in the neuroepithelial domain of the hindbrain, which gives rise to 5-HT neurons. Mash1 is essential for the development of 5-HT neurons (serotonergic) as a proneural gene for the production of postmitotic neuronal precursors and as a determinant of the serotonergic phenotype, Pattyn A, et al. (2004).
Mash1 supports the differentiation of specific cell subpopulation within the spinal cord. The ventral spinal cord p2 progenitor domain gives rise to interneurons V2a and V2b. Mash1 acts synergistically with Foxn4 to specify the development of V2b interneurons, Li S, et al. (2005), Del Barrio MG, et al. (2007). The major neuronal subtypes in the dorsal spinal cord are dILA and dILB neurons. A Mash1+ progenitor population gives rise to the dILA and dILB neurons of the dorsal spinal cord; however, Mash1 promotes the final differentiation of dILA, but not dILB, Wildner H, et al. (2006).
Mash 1 supports the differentiation of several neuroendocrine cell types including chromaffin cells that are derived from neural crest derived sympathoadrenal (SA) progenitors, Huber K, et al. (2002), Unsicker K, et al. (2005); hypothalamic propiomelanocortin (POMC+) neurons, McNay DE, et al. (2006) and gastric neuroendocrine cells, Kokubu H, et al. (2008).
Mash1 is required for the development of a variety of other cell types including chemoreceptive glomus cells of the carotid body, Kameda Y. (2005), olfactory neurons, Cau E, et al. (1997, 2002), basal cells and type II cells of taste buds, Seta Y, et al. (2006), Miura H, et al. (2006), and C-cells of the thyroid gland, Kameda Y, et al. (2007).
Studies of the expression profiles, genetic cascades and regulatory roles of Mash1 may enhance an understanding or treatment of several major medical conditions. A recent study has suggested that Mash1 may be useful in the development of motoneurons to treat spinal cord injuries, Hamada M, et al. (2006). Mash1 is involved in a beta-Amyloid (Abeta) induced process that promotes cell death, Uchida Y, et al. (2007) associated with Alzheimer’s disease. Since Mash1 is involved in the development of dopaminergic neurons, it may be important to scientists for understanding and treatment of Parkinson’s disease.
Mash1 has been identified as a positive adjunct marker of pulmonary small cell carcinoma versus Merkel cell carcinoma, Ralston J, et al. (2008).
Sigma offers antibodies and shRNAs useful for the study of ASCL1 (MASH1) gene products.
References:
Anderson DJ, et al. (1997) Cell lineage determination and the control of neuronal identity in the neural crest. Cold Spring Harb Symp Quant Biol. 62: 493-504.
Anderson DJ. (1994) Stem cells and transcription factors in the development of the mammalian neural crest. FASEB J. 8: 707-713.
Britz O, et al. (2006) A role for proneural genes in the maturation of cortical progenitor cells. Cereb Cortex. 16 Suppl 1: i138-51.
Casarosa S, et al. (1999) Mash1 regulates neurogenesis in the ventral telencephalon. Development. 126: 525-534.
Cau E, et al. (2002) Mash1 and Ngn1 control distinct steps of determination and differentiation in the olfactory sensory neuron lineage. Development. 129: 1871-1880.
Cau E, et al. (1997) Mash1 activates a cascade of bHLH regulators in olfactory neuron progenitors.Development. 124: 1611-1621.
Del Barrio MG, et al. (2007) A regulatory network involving Foxn4, Mash1 and delta-like 4/Notch1 generates V2a and V2b spinal interneurons from a common progenitor pool. Development. 134: 3427-3436.
Hamada M, et al. (2006) Introduction of the MASH1 gene into mouse embryonic stem cells leads to differentiation of motoneuron precursors lacking Nogo receptor expression that can be applicable for transplantation to spinal cord injury. Neurobiol Dis. 22: 509-522.
Hirsch MR, et al. (1998) Control of noradrenergic differentiation and Phox2a expression by MASH1 in the central and peripheral nervous system. Development. 125: 599-608.
Horton S, et al. (1999) Correct coordination of neuronal differentiation events in ventral forebrain requires the bHLH factor MASH1. Mol Cell Neurosci. 14: 355-369.
Huber K, et al. (2002) Development of chromaffin cells depends on MASH1 function. Development. 129: 4729-4738.
Jo AY, et al. (2007) Contrasting and brain region-specific roles of neurogenin2 and mash1 in GABAergic neuron differentiation in vitro. Exp Cell Res. 313: 4066-4081.
Kameda Y, et al. (2007) Mash1 regulates the development of C cells in mouse thyroid glands. Dev Dyn. 236: 262-270.
Kameda Y. (2005) Mash1 is required for glomus cell formation in the mouse carotid body. Dev Biol. 283: 128-139.
Kim EJ, et al. (2008) Ascl1 (Mash1) lineage cells contribute to discrete cell populations in CNS architecture. Mol Cell Neurosci. 38: 595-606.
Kim HJ, et al. (2007) Control of neurogenesis and tyrosine hydroxylase expression in neural progenitor cells through bHLH proteins and Nurr1. Exp Neurol. 203: 394-405.
Kokubu H, et al. (2008) Mash1 is required for neuroendocrine cell development in the glandular stomach. Genes Cells. 13: 41-51.
Letinic K, et al. (2002) Origin of GABAergic neurons in the human neocortex. Nature. 417: 645-649.
Li S, et al. (2005) Foxn4 acts synergistically with Mash1 to specify subtype identity of V2 interneurons in the spinal cord. Proc Natl Acad Sci U S A. 102: 10688-10693.
Lo LC, et al. (1991) Mammalian achaete-scute homolog 1 is transiently expressed by spatially restricted subsets of early neuroepithelial and neural crest cells. Genes Dev. 5: 1524-1537.
Ma Q, et al. (1997) Mash1 and neurogenin1 expression patterns define complementary domains of neuroepithelium in the developing CNS and are correlated with regions expressing notch ligands. Neurosci. 17: 3644-3652.
McNay DE, et al. (2006) Mash1 is required for generic and subtype differentiation of hypothalamic neuroendocrine cells. Mol Endocrinol. 20: 1623-1632.
Miura H, et al. (2006) Cell lineage and differentiation in taste buds. Arch Histol Cytol. 69: 209-225.
Ohsawa R, et al. (2005) Mash1 and Math3 are required for development of branchiomotor neurons and maintenance of neural progenitors. J Neurosci. 25: 5857-5865.
Park CH, et al. (2006) Differential actions of the proneural genes encoding Mash1 and neurogenins in Nurr1-induced dopamine neuron differentiation. J Cell Sci. 119: 2310-2320.
Pattyn A, et al. (2004) Ascl1/Mash1 is required for the development of central serotonergic neurons. Nat Neurosci. 7: 589-595.
Pattyn A, et al. (1999) The homeobox gene Phox2b is essential for the development of autonomic neural crest derivatives. Nature. 399: 366-370.
Parras CM, et al. (2007) The proneural gene Mash1 specifies an early population of telencephalic oligodendrocytes. J Neurosci. 27: 4233-4242.
Parras CM, et al. (2002) Divergent functions of the proneural genes Mash1 and Ngn2 in the specification of neuronal subtype identity. Genes Dev. 16: 324-338.
Ralston J, et al. (2008) MASH1: a useful marker in differentiating pulmonary small cell carcinoma from Merkel cell carcinoma. Mod Pathol. 2008 Jun 27. [Epub ahead of print]
Seta Y, et al. (2006) The bHLH transcription factors, Hes6 and Mash1, are expressed in distinct subsets of cells within adult mouse taste buds. Arch Histol Cytol. 69: 189-198.
Sommer L, et al. (1995) The cellular function of MASH1 in autonomic neurogenesis. Neuron. 15: 1245-1258.
Uchida Y, et al. (2007) Differential regulation of basic helix-loop-helix factors Mash1 and Olig2 by beta-amyloid accelerates both differentiation and death of cultured neural stem/progenitor cells. J Biol Chem. 282: 19700-19709.
Unsicker K, et al. (2005) The chromaffin cell and its development. Neurochem Res. 30: 921-925.
Wildner H, et al. (2006) dILA neurons in the dorsal spinal cord are the product of terminal and non-terminal asymmetric progenitor cell divisions, and require Mash1 for their development. Development. 133: 2105-2113.
Yun K, et al. (2002) Modulation of the notch signaling by Mash1 and Dlx1/2 regulates sequential specification and differentiation of progenitor cell types in the subcortical telencephalon. Development. 129: 5029-5040.
Footnote: Gene Data Sources: HGNC, Entrez Gene, UniProt/Swiss-Prot, UniProt/TrEMBL, GDB, OMIM, GeneLoc, Ensembl.
Copyright © 2008 Sigma-Aldrich Co. Reproduction forbidden without permission.
Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. Best viewed in IE6 or higher.