Your Favorite Gene - Powered By Ingenuity
www.sigmaaldrich.com/stemcell
Sigma-Aldrich
Your Favorite Gene - Powered By Ingenuity
Find NKX2-2 Products
Gene NKX2-2; NKX22_HUMAN
Homeobox protein Nkx-2.2
NCBI/Entrez 4821
HGNC 7835
UniProt/Swiss-Prot/ UniProt/TrEMBL O95096 Q9HBZ5
Ensembl ENSG00000125820
OMIM 604612
GeneCards GC20M021439
Synonyms: Homeobox protein NK-2 homolog B, Homeobox protein Nkx-2.2, NKX2.2, NKX2B, NK2 homeobox 2
Homeobox protein Nkx-2.2 (Gene Nkx2.2) Homo Sapiens
The Nkx2-2 (map locus: 20p11.22) gene product, homeobox protein Nkx2.2, is a 273 AA (30.1 kDa) NK2 family transcription factor that contains a 60 AA (128 to 187) DNA binding homeobox. The protein contains two nuclear localization signals (NLS) that function cooperatively; KKRKRR and RYKMKRAR, Hessabi B, et al. (2000).
Nkx2.2, Nkx6.1 and Irx3 are part of a homeodomain protein combinatorial code that specifies progenitor cell identity and neuronal fate in the ventral neural tube, Briscoe J, et al (2000). Nkx2.2 is expressed in the ventral midline domains of neuroectoderm (neural tube), Weiss JB, et al. (1998), during early development where its expression requires sonic hedgehog (Shh) signaling, Pabst O. et al. (2000).
Nkx2-2 is required for the establishment of the dopaminergic progenitor domain in mammalian ventral midbrain, Prakash N and Wurst W, (2007) and serotonergic (5-HT) neurons in the brainstem and ventral spinal cord, Cheng L, et al. (2003). Nkx2.2 is selectively expressed in oligodendrocyte progenitors (OPC) that arise from glial precursors located in the ventral neural tube.
Nkx2.2 has emerged as a key regulator of oligodendrocyte differentiation. It regulates the differentiation and/or maturation, but not the initial specification of oligodendrocyte progenitors, Qi Y, et al. (2001). Oligodendrocytes (OL) arise from Nkx2.2-expressing progenitors by a Shh-dependent mechanism, Soula C, et al. (2001). Coexpression of Olig2 (a bHLH transcription factor) and Nkx2.2 in the spinal cord, Zhou Q, et al. (2001) and ventricular and subventricular zones of the midbrain, Fu H, et al. (2003) promotes oligodendrocyte differentiation, Agius E, et al. (2004), wherein both factors act as repressors, Zhou Q, et al. (2001). In addition to supporting oligodendrocyte differentiation during development, Nkx2.2 supports the generation of new oligodendrocytes and remyelination after injury to adult CNS tissues, Watanabe M, et al. (2004), Fancy SP, et al. (2004).
Nkx2.2 is involved in the differentiation of endocrine cell types within the pancreas. It is required for the differentiation of insulin-producing beta-cells, Sander M, et al. (2000), Wang J, et al. (2004) and most of the glucagon-producing alpha-cells, Prado CL, et al. (2004). In addition, it represses the differentiation of epsilon (ghrelin-secreting) pancreatic cells, Pauls S, et al. (2007). Nkx2.2 is required for mature beta-cell function and islet structure, Doyle MJ and Sussel L. (2007). Nkx2.2 promotes differentiation of ES stem cells into insulin-producing cells, Shiroi A, et al. (2005).
Nkx2.2 is expressed at the onset of intestinal epithelial cell differentiation where it supports hormone-producing enteroendocrine cell differentiation, Desai S, et al. (2008) and taste bud cells, Miura H, et al. (2003).
Nkx2.2 is emerging as a marker for oncogenic transformation of Ewing’s sarcoma where its presence correlates with a poor prognosis, Smith R, et al. (2006), Owen LA, et al. (2008), Cheung IY, et al. (2007).
Sigma offers antibodies and shRNAs useful for the study of Nkx2.2 gene products.
References:
Briscoe J, et al. (2000) A homeodomain protein code specifies progenitor cell identity and neuronal fate in the ventral neural tube. Cell. 101: 435-445.
Cheng L, et al. (2003) Lmx1b, Pet-1, and Nkx2.2 coordinately specify serotonergic neurotransmitter phenotype. J Neurosci. 23: 9961-9967.
Cheung IY, et al. (2007) Novel markers of subclinical disease for Ewing family tumors from gene expression profiling. Clin Cancer Res. 13: 6978-6983.
Doyle MJ and Sussel L. (2007) Nkx2.2 regulates beta-cell function in the mature islet. Diabetes. 56: 1999-2007.
Fancy SP, et al. (2004) Increased expression of Nkx2.2 and Olig2 identifies reactive oligodendrocyte progenitor cells responding to demyelination in the adult CNS. Mol Cell Neurosci. 27: 247-254.
Hessabi B, et al. (2000) The homeodomain of Nkx2.2 carries two cooperatively acting nuclear localization signals. Biochem Biophys Res Commun. 270: 695-700.
Miura H, et al. (2003) Co-expression pattern of Shh with Prox1 and that of Nkx2.2 with Mash1 in mouse taste bud. Gene Expr Patterns. 3: 427-430.
Owen LA, et al. (2008) EWS/FLI mediates transcriptional repression via NKX2.2 during oncogenic transformation in Ewing's sarcoma. PLoS ONE. 3: e1965.
Pabst O, et al. (2000) NKX2 gene expression in neuroectoderm but not in mesendodermally derived structures depends on sonic hedgehog in mouse embryos. Dev Genes Evol. 210: 47-50.
Pauls S, et al. (2007) Function and regulation of zebrafish nkx2.2a during development of pancreatic islet and ducts. Dev Biol. 304: 875-890.
Prado CL, et al. (2004) Ghrelin cells replace insulin-producing beta cells in two mouse models of pancreas development. Proc Natl Acad Sci U S A. 101: 2924-2929.
Prakash N and Wurst W. (2007) A Wnt signal regulates stem cell fate and differentiation in vivo. Neurodegener Dis. 4: 333-338.
Qi Y, et al. (2001) Control of oligodendrocyte differentiation by the Nkx2.2 homeodomain transcription factor. Development. 128: 2723-2733.
Sander M, et al. (2000) Homeobox gene Nkx6.1 lies downstream of Nkx2.2 in the major pathway of beta-cell formation in the pancreas. Development. 127: 5533-5540.
Shiroi A, et al. (2005) Differentiation of embryonic stem cells into insulin-producing cells promoted by Nkx2.2 gene transfer. World J Gastroenterol. 11: 4161-4166.
Smith R, et al. (2006) Expression profiling of EWS/FLI identifies NKX2.2 as a critical target gene in Ewing's sarcoma. Cancer Cell. 9: 405-416.
Soula C, et al. (2001) Distinct sites of origin of oligodendrocytes and somatic motoneurons in the chick spinal cord: oligodendrocytes arise from Nkx2.2-expressing progenitors by a Shh-dependent mechanism. Development. 128: 1369-1379.
Wang J, et al. (2004) The concerted activities of Pax4 and Nkx2.2 are essential to initiate pancreatic beta-cell differentiation. Dev Biol. 266: 178-189.
Watanabe M, et al. (2004) Transient upregulation of Nkx2.2 expression in oligodendrocyte lineage cells during remyelination. Glia. 46: 311-322.
Weiss JB, et al. (1998) Dorsoventral patterning in the Drosophila central nervous system: the intermediate neuroblasts defective homeobox gene specifies intermediate column identity. Genes Dev. 12: 3591-3602.
Zhou Q, et al. (2001) The bHLH transcription factor Olig2 promotes oligodendrocyte differentiation in collaboration with Nkx2.2. Neuron. 31: 791-807.
Footnote: Gene Data Sources: HGNC, Entrez Gene, UniProt/Swiss-Prot, UniProt/TrEMBL, GDB, OMIM, GeneLoc, Ensembl.
Copyright © 2008 Sigma-Aldrich Co. Reproduction forbidden without permission.
Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. Best viewed in IE6 or higher.