Your Favorite Gene - Powered By Ingenuity
www.sigmaaldrich.com/stemcell
Sigma-Aldrich
Your Favorite Gene - Powered By Ingenuity
Find NR4A2 Products
Gene NR4A2: NR4A2_HUMAN
Nuclear receptor subfamily 4, group A, member 2
NCBI/Entrez 4929
HGNC 7981
UniProt/Swiss-Prot/ UniProt/TrEMBL P43354 Q6NXU0 Q53EL4
Ensembl ENSG00000153234
OMIM 168600 601828
GeneCards GC02M156889
Synonyms: HZF-3, Immediate-early response protein NOT, NOT, NURR1, Orphan nuclear receptor NR4A2, Orphan nuclear receptor NURR1, RNR1, TINUR, Transcriptionally-inducible nuclear receptor
Nuclear receptor subfamily 4, group A, member 2 (Gene NR4A2) Homo Sapiens
The NR4A2 (map locus: Ensembl: 2q24.1; Entrez/HGNC: 2q22-q23) gene product, NURR1/NOT/Nuclear receptor subfamily 4 group A member 2, is a 598 AA (66.6 kDa) protein, that contains a 76 AA (260-335) DNA-binding domain, two NR C4-type zinc finger domains, 21 AA (263-283) and 25 AA (299 to 323), and a ligand-binding domain, 51 AA (409 to 459). Nurr1 also contains a phosphorylation site, Ser181. Four distinct isoforms have been identified. The mouse and human NR4A2 genes have been cloned and characterized by Saucedo-Cardenas O, (1997) and Ichinose H, et al. (1999), respectively. Human NR4A2 contains eight exons and the potential regulatory consensus binding sites for NF-kappaB, CREB and Sp1.
Nurr1 is a member of a large gene family comprised of nuclear receptors for steroids, retinoids and thyroid hormones. Nurr1 was identified in 1992 as a brain-specific transcription factor, Law SW, et al. (1992), but its expression occurs in other tissues and organs outside the brain. Nurr1 is also expressed in T-cells, B-cells, fibroblasts, and synovial inflammatory cells. It is involved in T-cell apoptosis, brain development, the hypothalamic-pituitary-adrenal axis and vascular disease.
Nurr1 is selectively expressed in both the developing and the adult brain where it persists in the olfactory bulb, parts of the cortex, the hippocampal formation and especially the substantia nigra. The substantia nigra and ventral tegment are areas that regulate movement and affective behavior. Cells in these areas typically degenerate in Parkinson’s disease (PD). Consequently, Nurr1 has been studied extensively in the area of dopaminergic system development, Jankovic J, et al. (2005).
NURR1 is required for the maintenance of the dopaminergic system. It is expressed in the nigrostriatal dopamine system and is required for maintenance of dopaminergic neurons, Zetterström RH, et al. (1996, 1997); Pennisi E. (1997); and Le W, et al. (1999). Nurr1 functions late in dopamine cell development to drive differentiation of ventral mesencephalic late DA precursor neurons where it is essential for specifying commitment of mesencephalic precursors to the full dopaminergic phenotype, Saucedo-Cardenas O, et al. (1998). Nurr1 supports dopaminergic neurons in the substantia nigra/ventral tegmental area, but is not essential to hypothalamic neurons, Castillo, SO, et al. (1998). Nurr1 can induce cell arrest and a highly differentiated morphology in the dopamine-synthesizing cell line MN9D, Castro DS, et al. (2001). The role of Nurr1 in differentiation of DA neurons precursors and DA neuron survival has been reviewed, Arenas E. (2005) and Alavian KN, et al. (2008).
Nurr1 enhances the transcription of several genes that support the synthesis and storage of dopamine. Nurr1 enhances the transcription of human tyrosine hydroxylase (TH), Sakurada K, et al. (1999), dopamine transporter gene (hDAT), Sacchetti P, et al (2001), vesicular monoamine transporter 2 (VMAT2), and l-aromatic amino acid decarboxylase (AADC), Hermanson E, et al. (2003).
In addition to its importance for the understanding of neurological diseases such as Parkinson’s disease, Nurr1 has been associated with major inflammatory diseases such as rheumatoid (RA) and psoriatic arthritis (PsA) and vascular diseases such as atherosclerosis and vein-graft disease.
Nurr1 may have a role in human inflammatory arthritis, McEvoy AN, et al. (2002), Davies MR, et al. (2005). Nurr1 is induced in rheumatoid arthritis (RA) and psoriatic arthritis (PsA) inflammatory cells by synoviocyte corticotrophin-releasing hormone (CRH), Murphy EP, et al. (2004). Glucocorticoids dramatically suppressed cytokine- and CRH-induced synovial NURR1 mRNA.
The links between Nurr1 and vascular diseases such as atherosclerosis and vein-graft disease has been studied, Pei L, et al. (2005), Martínez-González J, et al (2005) and recently reviewed, Bonta PI, et al. (2007), Pols TW, et al. (2007).
Sigma offers antibodies and shRNAs useful for the study of NR4A2 (NURR1) gene products.
References:
Alavian KN, et al. (2008) Transcriptional regulation of mesencephalic dopaminergic neurons: the full circle of life and death. Mov Disord. 23: 319-328.
Arenas E. (2005) Engineering a dopaminergic phenotype in stem/precursor cells: role of Nurr1, glia-derived signals, and Wnts. Ann N Y Acad Sci. 1049: 51-66.
Bonta PI, et al. (2007) NR4A nuclear receptors in atherosclerosis and vein-graft disease. Trends Cardiovasc Med. 17:105-11.
Castillo SO, et al. (1998) Dopamine biosynthesis is selectively abolished in substantia nigra/ventral tegmental area but not in hypothalamic neurons in mice with targeted disruption of the Nurr1 gene. Mol Cell Neurosci. 11: 36-46.
Castro DS, et al. (2001) Induction of cell cycle arrest and morphological differentiation by Nurr1 and retinoids in dopamine MN9D cells. J Biol Chem. 276: 43277-43284.
Davies MR, et al. (2005) Nurr1 dependent regulation of pro-inflammatory mediators in immortalised synovial fibroblasts. J Inflamm (Lond). 2: 15.
Hermanson E, et al. (2003) Nurr1 regulates dopamine synthesis and storage in MN9D dopamine cells. Exp Cell Res. 288: 324-334.
Ichinose H, et al. (1999) Molecular cloning of the human Nurr1 gene: characterization of the human gene and cDNAs. Gene. 230: 233-239.
Jankovic J, et al. (2005) The role of Nurr1 in the development of dopaminergic neurons and Parkinson's disease. Prog Neurobiol. 77: 128-138.
Law SW, et al. (1992) Identification of a new brain-specific transcription factor, NURR1. Mol Endocrinol. 6: 2129-2135.
Le W, et al. (1999) Selective agenesis of mesencephalic dopaminergic neurons in Nurr1-deficient mice. Exp Neurol. 159: 451-458.
Martínez-González J and Badimon L. (2005) The NR4A subfamily of nuclear receptors: new early genes regulated by growth factors in vascular cells. Cardiovasc Res. 65: 609-618.
McEvoy AN, et al. (2002) Activation of nuclear orphan receptor NURR1 transcription by NF-kappa B and cyclic adenosine 5'-monophosphate response element-binding protein in rheumatoid arthritis synovial tissue. J Immunol. 168: 2979-2987.
Murphy EP, et al. (2004) Involvement of the nuclear orphan receptor NURR1 in the regulation of corticotropin-releasing hormone expression and actions in human inflammatory arthritis. Arthritis Rheum. 44: 782-793.
Pei L, et al. (2000) Induction of NR4A orphan nuclear receptor expression in macrophages in response to inflammatory stimuli. J Biol Chem. 280: 29256-29262.
Pennisi E. (1997) Key protein found for brain's dopamine-producing neurons. J Science. 276: 202.
Pols TW, et al. (2007) NR4A nuclear orphan receptors: protective in vascular disease? Curr Opin Lipidol. 18: 515-520.
Sacchetti P, et al. (2001) Nurr1 enhances transcription of the human dopamine transporter gene through a novel mechanism. J. Neurochem. 76: 1565-1572.
Sakurada K, et al. (1999) Nurr1, an orphan nuclear receptor, is a transcriptional activator of endogenous tyrosine hydroxylase in neural progenitor cells derived from the adult brain. Development. 126: 4017-4026.
Saucedo-Cardenas O, et al. (1998) Nurr1 is essential for the induction of the dopaminergic phenotype and the survival of ventral mesencephalic late dopaminergic precursor neurons. Proc Natl Acad Sci U S A. 95: 4013-4018.
Saucedo-Cardenas O, et al. (1997) Cloning and structural organization of the gene encoding the murine nuclear receptor transcription factor, NURR1. Gene. 187: 135-139.
Zetterström RH, et al. (1997) Dopamine neuron agenesis in Nurr1-deficient mice. Science. 276: 248-250.
Zetterström RH, et al. (1996) Cellular expression of the immediate early transcription factors Nurr1 and NGFI-B suggests a gene regulatory role in several brain regions including the nigrostriatal dopamine system. Brain Res Mol Brain Res. 41: 111-120.
Footnote: Gene Data Sources: HGNC, Entrez Gene, UniProt/Swiss-Prot, UniProt/TrEMBL, GDB, OMIM, GeneLoc, Ensembl.
Copyright © 2008 Sigma-Aldrich Co. Reproduction forbidden without permission.
Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. Best viewed in IE6 or higher.