Your Favorite Gene - Powered By Ingenuity
www.sigmaaldrich.com/stemcell
Sigma-Aldrich
Your Favorite Gene - Powered By Ingenuity
Find PAX6 Products
Gene PAX6: PAX6_HUMAN
Paired box gene 6 (aniridia, keratitis)
NCBI/Entrez 5080
HGNC 8620
UniProt/Swiss-Prot/ UniProt/TrEMBL P26367 Q6F6J5 Q8IVH0
Ensembl ENSG00000007372
OMIM 148190 106210
GeneCards GC11M031768
Synonyms: AN, AN2, Aniridia type II protein, D11S812E, MGC17209, MGDA, Oculorhombin, Paired box protein Pax-6, WAGR
Paired box protein Pax-6 (Gene PAX6) Homo sapiens
Pax family genes characterized by the presence of a highly conserved DNA-binding motif (paired domain) regulate embryonic pattern formation and cell proliferation and differentiation during embryonic organogenesis. The Pax subfamily group 4 composed of Pax4 and Pax6 contains a homeodomain in addition to the paired box domain common to the family, Dahl E, et al. (1997).
The PAX6 (map locus: 11p13) gene product, paired box gene 6/Pax6, exists in several isoforms. Pax6-1 and Pax6,5a are the most extensively studied isoforms. Pax6-1 (Pax6-a) is a 422 AA (46.6 kDa) protein that contains a paired box (paired domain), 127 AA (4 to 130); a DNA-binding homeobox (homeodomain) 60 AA (210 to 269), a Gln/Gly-rich region, 79 AA (131 to 209) and a proline/serine/threonine (P/S/T)-rich C-terminal domain. Pax6,5a, 436 AA, is altered by a 5a insertion (5a exon) within the paired box region; wherein, Gln47 is replaced with a 15 AA sequence. Different aspects of Pax6 gene function are mediated by different isoforms of the Pax6 protein. The distinct roles in brain development of the paired domain, homeodomain and 5a splice have been reviewed, Haubst N et al. (2004). The 5a positive isoform has recently been linked to development of the fovea within the vertebrate retina, Azuma N, et al. (2005).
Pax-6 is widely expressed in the developing CNS during the neural plate stage. It is expressed in the neural tube just after its closure and subsequently it is spatially and temporally restricted within the telencephalon and diencephalon of the forebrain, Mastick, GS, et al. (1997); the optic and nasal placodes, Li HS, et al. (1994); the rhombencephalon (hindbrain); the somites and the spinal cord, Gerard M, et al. (1995), Amirthalingam K, et al. (1995). Pax6 is found in both layers of the optic cup, the optic stalk and prospective corneal epithelium; in the infundibulum and in Rathke’s pouch, Terzic J and Saraga-Babic M. (1999).
In developing human brain, Pax6 is strongly expressed in proliferating radial glia (RG) cells and some neuronal and intermediate progenitors within the dorsal and ventral proliferative zones, Mo Z and Zecevic N. (2008). It is required for development of radial glial and cortical cells and neuronal migration, Warren N, et al. (1999).
Pax6 contributes to both embryonic and adult neurogenesis as a multifunctional regulator, Osumi N, et al. (2008). Pax6 is a patterning and arealization dorsalizing factor that regulates dorso-ventral and anterior-posterior neural cell specification, migration and axonal projections within the forebrain telencephalon, Stoykova A, et al. (2000) and the diencephalon (thalamocortical development), Kawano H, et al. (1999), Pratt T, et al. (2000; the hindbrain (rhombencephalon), Engelkamp D, et al. (1999); Osumi N, et al. (1997) and the spinal cord, Sapir T, et al. (2004). Pax6 is required to establish the diencephalic/mesencephalic (forebrain/midbrain) boundary, Matsunaga E, at al. (2000).
Pax6 supports the regionalization of the telencephalon by limiting the invasion of the cortex by cells originating in the ganglionic eminence, Chapouton P, et al. (1999). Pax6 is critical to the formation of the first two major longitudinal tracts, the tract of the postoptic commissure (TPOC) (a major pathway for forebrain dopaminergic projections) and the stria medullaris, in embryonic mouse forebrain, Nural HF and Mastick GS (2004). Pax-6 is required for thalamocortical pathway formation in fetal rats, Kawano H, et al. (1999). Pax6 is required for the formation of the subcommissural organ (SCO) via development of the diencephalic dorsal midline secretory radial glia, Estivill-Torrús G, et al. (2001).
Pax6 is a master control gene for eye development, Gehring WJ, (1996) that is required for the proper formation of the lens, iris, retina and cornea, Davis J. et al. (2003); Baumer N, et al. (2002). Mutations in Pax6 are linked to Aniridia and small eye. Pax6 is a competence factor that supports FGF10 induction of lacrimal gland development, Makarenkova HP, et al. (2000). Pax6 is present in all cells that form the neural retina and pigment epithelium including the lens, cornea and the neural and pigmented retinas, Macdonald R, et al. (1995), Davis JA and Reed RR, (1996). Pax6 is also expressed in surface epithelia of adult cornea and conjunctiva, Koroma BM, et al. (1997). The relative abundance of Pax6-5a isoform varies between lens and iris of the mature bovine eye, Jaworski C, et al. (1997).
Pax-6 is required for the formation of the pineal gland, Mitchell TN, et al. (2003) and it is dorsalizing factor within the developing pituitary, Kioussi C, et al. (1999).
Pax-6 is expressed in ectoderm derived tissues outside the nervous system. It is required for the differentiation of glucagon-producing alpha-cells within the pancreas, St-Onge, et al. (1997) and differentiation of gastrointestinal endocrine cells, Larsson LI, et al. (1998).
Pax6 is expressed in the majority of pancreatic adenocarcinomas, Lang D, et al. (2008) and in glioblastoma multiforme (GBM), an invasive brain cancer, Zhou YH, et al. (2005). In GBM, Pax6 is reported to be a repressor of invasiveness, Mayes DA, et al. (2006).
There are over 1300 Pax6 citations in Pubmed.
Sigma offers antibodies and shRNAs useful for the study of PAX6 gene products.
References:
Amirthalingam K, et al. (1995) Embryonic expression and DNA-binding properties of zebrafish pax-6. Biochem Biophys Res Commun. 215: 122-128.
Azuma N, et al. (2005) The Pax6 isoform bearing an alternative spliced exon promotes the development of the neural retinal structure. OHum Mol Genet. 14: 735-745.
Bäumer N, et al. (2002) Pax6 is required for establishing naso-temporal and dorsal characteristics of the optic vesicle. Development. 129: 4535-4545.
Chapouton P, et al. (1999) The role of Pax6 in restricting cell migration between developing cortex and basal ganglia. Development. 126: 5569-5579.
Dahl E, et al. (1997) Pax genes and organogenesis. Bioessays. 19: 755-765.
Davis J, et al. (2003) Requirement for Pax6 in corneal morphogenesis: a role in adhesion. J Cell Sci. 116: 2157-2167.
Davis JA and Reed RR. (1996) Role of Olf-1 and Pax-6 transcription factors in neurodevelopment. Neurosci. 16: 5082-5094.
Engelkamp D, et al. (1999) Role of Pax6 in development of the cerebellar system. Development. 126: 3585-3596.
Estivill-Torrús G, et al. (2001) The transcription factor Pax6 is required for development of the diencephalic dorsal midline secretory radial glia that form the subcommissural organ. Mech Dev. 109: 215-224.
Gehring WJ. (1996) The master control gene for morphogenesis and evolution of the eye. Genes Cells. 1: 11-15.
Gérard M, et al. (1995) PAX-genes expression during human embryonic development, a preliminary report. C R Acad Sci III. 318: 57-66.
Haubst N, et al. (2004) Molecular dissection of Pax6 function: the specific roles of the paired domain and homeodomain in brain development. Development. 131: 6131-6140.
Jaworski C, et al. (1997) Alternative splicing of Pax6 in bovine eye and evolutionary conservation of intron sequences. Biochem Biophys Res Commun. 240: 196-202.
Kawano H, et al. (1999) Pax-6 is required for thalamocortical pathway formation in fetal rats. J Comp Neurol. 408: 147-160.
Kioussi C, et al. (1999) Pax6 is essential for establishing ventral-dorsal cell boundaries in pituitary gland development. Proc Natl Acad Sci U S A. 96: 14378-14382.
Koroma BM, et al. (1997) The Pax-6 homeobox gene is expressed throughout the corneal and conjunctival epithelia. Invest Ophthalmol Vis Sci. 38: 108-120.
Lang D, et al. (2008) PAX6 is expressed in pancreatic adenocarcinoma and is downregulated during induction of terminal differentiation. Mol Carcinog. 47: 148-156.
Larsson LI, et al. (1998) Pax 4 and 6 regulate gastrointestinal endocrine cell development. Mech Dev. 79: 153-159.
Li HS, et al. (1994) Pax-6 is first expressed in a region of ectoderm anterior to the early neural plate: implications for stepwise determination of the lens. Dev Biol. 162: 181-194.
Macdonald R, et al. (1995) Midline signalling is required for Pax gene regulation and patterning of the eyes. Development. 121: 3267-3278.
Makarenkova HP, et al. (2000) FGF10 is an inducer and Pax6 a competence factor for lacrimal gland development. Development. 127: 2563-2572.
Mastick GS, et al. (1997) Pax-6 functions in boundary formation and axon guidance in the embryonic mouse forebrain. Development. 124: 1985-1997.
Matsunaga E, et al. (2000) Pax6 defines the di-mesencephalic boundary by repressing En1 and Pax2. Development. 127: 2357-2365.
Mayes DA, et al. (2006) PAX6 suppresses the invasiveness of glioblastoma cells and the expression of the matrix metalloproteinase-2 gene. Cancer Res. 66: 9809-9817.
Mitchell TN, et al. (2003) Polymicrogyria and absence of pineal gland due to PAX6 mutation. Ann Neurol. 53: 658-663.
Mo Z and Zecevic N. (2008) Is Pax6 critical for neurogenesis in the human fetal brain? Cereb Cortex. 18: 1455-1465.
Nural HF and Mastick GS. (2004) Pax6 guides a relay of pioneer longitudinal axons in the embryonic mouse forebrain. J Comp Neurol. 479: 399-409.
Osumi N, et al. (2008) Concise review: Pax6 transcription factor contributes to both embryonic and adult neurogenesis as a multifunctional regulator. Stem Cells. 26: 1663-1672.
Osumi N, et al. (1997) Pax-6 is involved in the specification of hindbrain motor neuron subtype. Development. 124: 2961-2972.
Pratt T, et al. (2000) A role for Pax6 in the normal development of dorsal thalamus and its cortical connections. Development. 127: 5167-5178.
Sapir T, et al. (2004) Pax6 and engrailed 1 regulate two distinct aspects of renshaw cell development. J Neurosci. 24: 1255-1264.
St-Onge L, et al. (1997) Pax6 is required for differentiation of glucagon-producing alpha-cells in mouse pancreas. Nature. 387: 406-409.
Stoykova A, et al. (2000) Pax6 modulates the dorsoventral patterning of the mammalian telencephalon. J Neurosci. 20: 8042-8050.
Terzic J and Saraga-Babic M. (1999) Expression pattern of PAX3 and PAX6 genes during human embryogenesis. Int J Dev Biol. 43: 501-508.
Warren N, et al. (1999) The transcription factor, Pax6, is required for cell proliferation and differentiation in the developing cerebral cortex. Cereb Cortex. 9: 627-635.
Zhou YH, et al. (2005) PAX6 suppresses growth of human glioblastoma cells. J Neurooncol.71: 223-229.
Footnote: Gene Data Sources: HGNC, Entrez Gene, UniProt/Swiss-Prot, UniProt/TrEMBL, GDB, OMIM, GeneLoc, Ensembl.
Copyright © 2008 Sigma-Aldrich Co. Reproduction forbidden without permission.
Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. Best viewed in IE6 or higher.