Your Favorite Gene - Powered By Ingenuity
www.sigmaaldrich.com/stemcell
Sigma-Aldrich
YFG search logo
Find PHOX2A Products
Gene PHOX2A: PHX2A_HUMAN
Paired mesoderm homeobox protein 2A
NCBI/Entrez 401
HGNC 691
UniProt/Swiss-Prot/ UniProt/TrEMBL O14813 Q96KQ2
Ensembl ENSG00000165462
OMIM 602078, 602753
GeneCards GC11M071627
Synonyms: Aristaless homeobox protein homolog, ARIX, ARIX1 homeodomain protein, CFEOM2, FEOM2, MGC52227, NCAM2, Paired-like homeobox 2A, Paired mesoderm homeobox protein 2A, PMX2A
Paired-like Homeobox 2a (Gene PHOX2A)
The human PHOX2A (map locus: Entrez: 11a13.2: Ensembl/HGNC: 11q13.4) gene product, paired-like homeobox 2A, is a 284 AA (29.6 kDa) protein that contains a 60 AA (90-149) DNA-binding helix-turn-helix (HTH) homeobox. Phox2a and phox2b are paralogues with identical homeobox domains. However, the paralogues are not functionally equivalent. Phox2b can replace Phox2a in structures that depend on both genes, but 2a cannot always replace 2b, Coppola E, et al. (2005).
Phox2a plays a critical role in the development of the autonomic nervous system (ANS), wherein it promotes the differentiation of dopaminergic and noradrenergic neurons. Phox2a is expressed in differentiating neurons of the central and peripheral autonomic nervous system and motor nuclei of the hindbrain, Pattyn A et al. (1997). Phox2a is essential for proper development of the locus coeruleus, visceral sensory ganglia (adrenergic), a subset of sympathetic and parasympathetic (noradrenergic) ganglia and the IIIrd, IVth, VIIth, IXth, and Xth cranial sensory ganglia, Morin X, et al. (1997), Pattyn A et al. (1997).
The locus coeruleus (LC) is the major noradrenergic center of the brain. The LC neurons originate in the alar plate of rhombomere 1 and migrate into the basal plate, Aroca P, et al. (2006). Development of the LC requires initial BMP-dependent expression of Phox2a and Phox2b in dorsal rhombomere1 (r1), Vogel-Höpker A and Rohrer H. (2002). The transformation of LC neurons into noradrenergic neurons involves the expression of two catecholaminergic enzymes, tyrosine hydroxylase (TH) and dopamine beta-hydroxylase (DBH), the terminal enzyme for noradrenaline biosynthesis. Phox2a/Arix is expressed in noradrenergic, DBH-positive tissues and regulates the transcription of TH and DBH in neural crest-derived noradrenergic neurons (sympatho-adrenergic cells), Zellmer E, et al. (1995); Yang C, et al. (1998); Lo L, et al. (1999); Seo H, et al. (2002); and Stanke M, et al. (1999).
The induction of DBH expression by Phox2a/Arix is enhanced by direct protein-protein interaction with HAND2, Rychlik JL, et al. (2003) and interaction between HAND2 and CBP (cAMP response element binding protein), Xu H, et al. (2003) in the transcription complex. Hand2 plays a major role in the induction of the noradrenalin phenotype, Morikawa Y, et al. (2007).
Phox2a regulates transcription of human alpha3 nicotinic acetylcholine receptor gene, which encodes the ligand-binding subunit of the ganglionic type nicotinic receptor. The alpha3 subunit is expressed in every terminally differentiated ganglionic cell; this is the first example of a "pan-autonomic" gene whose expression is regulated by PHOX2 proteins, Benfante R, et al. (2007).
Phox2a mutations have been linked to autosomal recessive congenital fibrosis of the extraocular muscles (CFEOM2; MIM: 602078), Wang SM, et al. (1998); Yazdani A, et al. (2003), reviewed by Heidary G, et al. (2008). CFEOM2 is a strabismus syndrome characterized by non-progressive, restrictive ophthalmoplegia of the extra-ocular muscles and congenital blepharoptosis and the dysinnervation of extraocular muscles supplied by the oculomotor (cranial nerve III) and/or trochlear (cranial nerve IV) nerves, Bosley TM et al. (2006).
Sigma offers antibodies, shRNAs and other products useful for the study of the PHOX2A.
References:
  1. Aroca P, et al. (2006) Locus coeruleus neurons originate in alar rhombomere 1 and migrate into the basal plate: Studies in chick and mouse embryos. J Comp Neurol. 496: 802-818.
  2. Benfante R, et al. (2007) Transcription factor PHOX2A regulates the human alpha3 nicotinic receptor subunit gene promoter. J Biol Chem. 282: 13290-13302.
  3. Bosley TM, et al. (2006) Neurological features of congenital fibrosis of the extraocular muscles type 2 with mutations in PHOX2A. Brain. 129: 2363-2374.
  4. Coppola E, et al. (2005) Reciprocal gene replacements reveal unique functions for Phox2 genes during neural differentiation. EMBO J. 24:4392-403.
  5. Heidary G, et al. (2008) Congenital fibrosis of the extraocular muscles. Semin Ophthalmol. 23: 3-8.
  6. Lo L, et al. (1999) Specification of neurotransmitter identity by Phox2 proteins in neural crest stem cells. Neuron. 22: 693-705.
  7. Morikawa Y, et al. (2007) Hand2 determines the noradrenergic phenotype in the mouse sympathetic nervous system. Dev Biol. 307: 114-126.
  8. Morin X, et al. (1997) Defects in sensory and autonomic ganglia and absence of locus coeruleus in mice deficient for the homeobox gene Phox2a. Neuron. 18: 411-423.
  9. Pattyn A, et al. (1997) Expression and interactions of the two closely related homeobox genes Phox2a and Phox2b during neurogenesis. Development. 124: 4065-4075.
  10. Rychlik JL, et al. (2003) The interaction between dHAND and Arix at the dopamine beta-hydroxylase promoter region is independent of direct dHAND binding to DNA. J Biol Chem. 278: 49652-49660.
  11. Seo H, et al. (2002) A direct role of the homeodomain proteins Phox2a/2b in noradrenaline neurotransmitter identity determination. J Neurochem. 2002 Mar;80(5): 905-916.
  12. Stanke M, et al. (1999) The Phox2 homeodomain proteins are sufficient to promote the development of sympathetic neurons. Development. 126: 4087-4094.
  13. Vogel-Höpker A and Rohrer H. (2002) The specification of noradrenergic locus coeruleus (LC) neurones depends on bone morphogenetic proteins (BMPs). Development. 129: 983-991.
  14. Wang SM, et al. (1998) Congenital fibrosis of the extraocular muscles type 2, an inherited exotropic strabismus fixus, maps to distal 11q13. Am J Hum Genet. 63: 517-525.
  15. Xu H, et al. (2003) HAND2 synergistically enhances transcription of dopamine-beta-hydroxylase in the presence of Phox2a. Dev Biol. 262: 183-193.
  16. Yang C, et al. (1998) Paired-like homeodomain proteins, Phox2a and Phox2b, are responsible for noradrenergic cell-specific transcription of the dopamine beta-hydroxylase gene. J Neurochem. 71: 1813-1826.
  17. Yazdani A, et al. (2003) A novel PHOX2A/ARIX mutation in an Iranian family with congenital fibrosis of extraocular muscles type 2 (CFEOM2). Am J Ophthalmol. 136: 861-865.
  18. Zellmer E, et al. (1995) A homeodomain protein selectively expressed in noradrenergic tissue regulates transcription of neurotransmitter biosynthetic genes. J Neurosci. 15: 8109-8120.
Footnote: Gene Data Sources: HGNC, Entrez Gene, UniProt/Swiss-Prot, UniProt/TrEMBL, GDB, OMIM, GeneLoc, Ensembl.
Copyright © 2008 Sigma-Aldrich Co. Reproduction forbidden without permission.
Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. Best viewed in IE6 or higher.