Your Favorite Gene - Powered By Ingenuity
www.sigmaaldrich.com/stemcell
Sigma-Aldrich
Your Favorite Gene - Powered By Ingenuity
Find Sox21 Products
Gene SOX21: SOX21_HUMAN
Transcription factor SOX-21
NCBI/Entrez 11166
HGNC 11197
UniProt/Swiss-Prot/ UniProt/TrEMBL Q9Y651, Q5TBS1, O95129
Ensembl ENSG00000125285
OMIM 604974
GeneCards GC13M094159
Synonyms: SOX25, SOXA, SOX-A, Transcription factor SOX-21
SRY (sex determining region Y)-box 21/Sox-21 (Gene SOX21) Homo sapiens
The SOX21 (map locus: Entrez and HGNC: 13q31-q32; Ensembl 13q32.1, Malas S, (1999)) gene product, SRY (sex determining region Y)-box 21/Sox-21 protein, is a 276 AA (28.6 kDa) high mobility group (HMG box) domain (8 to 76) containing transcription factor. HMG box domains preferentially bind to and distort DNA. Sox21 contains three polyalanine tracts; 137 to 146, 154 to 166 and 208 to 219.
The Sox family members are HMG-box containing transcription factors related to mammalian testis determining factor, SRY. Sox14 and Sox21 are B2-subgroup Sox transcription factors involved primarily in gene repression, especially of neurogenesis. Sox14 and Sox21 are similar to one another but distinct from the B1-subgroup Sox proteins, Sox1, Sox2 and Sox3. Sox14 and Sox21 are viewed as opposing regulators of genes regulated by the B1 Sox factors, Uchikawa M et al. (1999).
Sox21 is generally classified as a repressor gene that promotes neurogenesis, Sandberg M, et al. (2005); Ohba H, et al. (2004). The overexpression of Sox21 leads to substantial repression of NGF-induced neurite outgrowth in PC12 cells, Ohba H, et al. (2004). Sox21 is upregulated by proneural basic helix-loop-helix proteins (pHLH), Sandberg M, et al. (2005). These pHLH proteins are members of pathways that compete with the B1-subgroup stem cell maintenance promoting SOX proteins, Sox1, Sox2 and Sox3. Hence, Sox21 helps to counter the pluripotency maintenance genes and to allow neurogenesis to occur, Wegner M and Stolt CC (2005).
The neurogenesis promoting activity of Sox21 is positionally restricted within the CNS, where it functions primarily as a dorsalizing factor that helps establish dorsoventral patterning, Rex M, et al. (1997). Sox21 is expressed mainly in the developing midbrain-hindbrain boundary at the onset of neurulation. Knock-out of Sox21a in zebrafish caused ventralization of the embryos, Argenton F, et al. (2004).
Sox 21 mRNA is not detected in normal adult tissues, Gure AO, at al. (2000) but has been reported in a couple of cancer models. Sox21 was reported to be expressed in high-metastatic K-1735 murine melanoma cells, Tani M, et al. (1997) and by some small cell lung cancer (SCLC) cell lines, Gure AO, at al. (2000).
Sigma offers antibodies, shRNAs and other products useful for the study of the SOX21 gene products.
References:
Argenton F, et al. (2004) Ectopic expression and knockdown of a zebrafish sox21 reveal its role as a transcriptional repressor in early development. Mech Dev. 121: 131-142.
Güre AO, et al. (2000) Serological identification of embryonic neural proteins as highly immunogenic tumor antigens in small cell lung cancer. Proc Natl Acad Sci U S A. 97: 4198-4203.
Malas S, et al. (1999) The isolation and high-resolution chromosomal mapping of human SOX14 and SOX21; two members of the SOX gene family related to SOX1, SOX2, and SOX3. Mamm Genome. 10: 934-937.
Ohba H, et al. (2004) Sox21 is a repressor of neuronal differentiation and is antagonized by YB-1. Neurosci Lett. 358: 157-160.
Rex M, et al. (1997) cSox21 exhibits a complex and dynamic pattern of transcription during embryonic development of the chick central nervous system. Mech Dev. 66: 39-53.
Sandberg M, et al. (2005) Sox21 promotes the progression of vertebrate neurogenesis. Nat Neurosci. 8: 995-1001.
Tani M, et al. (1997) Isolation of a novel Sry-related gene that is expressed in high-metastatic K-1735 murine melanoma cells. Genomics. 39: 30-37.
Uchikawa M, et al. (1999) Two distinct subgroups of Group B Sox genes for transcriptional activators and repressors: their expression during embryonic organogenesis of the chicken. Mech Dev. 84: 103-120.
Wegner M and Stolt CC. (2005) From stem cells to neurons and glia: a Soxist's view of neural development. Trends Neurosci. 28: 583-588.
Footnote: Gene Data Sources: HGNC, Entrez Gene, UniProt/Swiss-Prot, UniProt/TrEMBL, GDB, OMIM, GeneLoc, Ensembl.
Copyright © 2008 Sigma-Aldrich Co. Reproduction forbidden without permission.
Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. Best viewed in IE6 or higher.