Your Favorite Gene - Powered By Ingenuity
www.sigmaaldrich.com/stemcell
Sigma-Aldrich
Your Favorite Gene - Powered By Ingenuity
Find Sox9 Products
Gene SOX9: SOX9_HUMAN
SRY (sex determining region Y)-box 9 (campomelic dysplasia, autosomal sex-reversal)
NCBI/Entrez 6662
HGNC 11204
UniProt/Swiss-Prot/ UniProt/TrEMBL P48436, Q53Y80
Ensembl ENSG00000125398
OMIM 114290, 608160
GeneCards GC17P067628
Synonyms: CMD1, CMPD1, SRA1, Transcription factor SOX-9
SRY (sex determining region Y)-box 9/Sox-9 (Gene SOX9) Homo sapiens
The SOX9 (map locus: Entrez 17q24.3-q25.1; Ensembl 17q24.3 and HGNC 17q23) gene product, SRY (sex determining region Y)-box 9/Sox-9 protein, is a 509 AA (56.1 kDa) high mobility group (HMG box) domain (105 to 173) containing transcription factor. HMG box domains preferentially bind to and distort DNA. Sox9 contains a 40 AA Gln/Pro-rich region (339 to 378) that includes a 5 AA polyproline stretch (342 to 346).
TSox9, Sox8 and Sox10 comprise subgroup E of the Sox-family transcription factors. This subgroup has been linked to the specification and differentiation of a variety of epithelial tissues and its members appear as early response genes to neural crest induction. Sox-9 promotes neural-crest-like properties in neural tube progenitors rather than CNS neural differentiation and together with the other SoxE genes directs migratory crest cells away from neuronal lineages towards glial cells, precursors of astrocytes and oligodendrocyts, and melanocytes, Cheung M and Briscoe J. (2003), Hong CS, et al. (2005). Sox9 also participates in the switching from neural to glial stem cell lineages in the developing spinal cord, Stolt CC, et al. (2003) and CNS, Kordes U, et al. (2005).
The role of Sox9 as a maintenance factor for progenitor lineage specific stem cell pools is starting to emerge. Sox9 is required for maintenance of the pancreatic progenitor cell pool, Seymour PA, et al. (2007), Lynn FC, et al. (2007); the retinal pool and Muller glial cells, Poche RA, et al. (2008); progenitor cells and the hair stem cell compartment, Vidal VP, et al. (2005), that participates in development of all skin epithelial lineages, Nowak JA et al. (2008) and the mesenchymal stem-cell derived osteochondroprogenitors, Zhou G, et al. (2006).
Sox9 has been linked to an impressive array of differentiation and development processes. These include: chondrogenesis, Healy C, et al. (1996, 1999), Wheatley S, et al. (1996), Lefebvre V and de Crombrugghe B. (1998), Bi W, et al (1999); differentiation of Sertoli cells, Morais da Silva S, et al. (1996), Kobayashi A, et al. (2005); tooth morphogenesis (odontogenesis), Mitsiadis TA et al. (1998); invagination of the otic placode, Barrionuevo F, et al. (2008); pancreas development, Lioubinski O, et al. (2003); valvulogenesis, Akiyama H, et al. (2004), Lincoln J, et al. (2007); specification of pyloric sphincter epithelium, Moniot B, et al. (2004); tracheal cartilage ring formation, Elluru RG and Whitsett JA. (2004); development of the outer root sheath (ORS) of hair follicles and hair stem cell compartment, Vidal VP, et al. (2005); maintenance of structural integrity of the notochord (axial skeletogenesis), Barrionuevo F, et al. (2006); differentiation of intestinal Paneth cells, Mori-Akiyama Y, et al. (2007); Bastide P, et al. (2007); melanocyte differentiation and pigmentation, Cook AL, et al. (2005); Passeron T, et al. (2007); and prostate development, Thomsen MK, et al. (2008).
The most exhaustively studied roles of Sox9 in development involve chondrogenesis and sex-determination. Dysregulations of Sox9 that involve loss of function mutations cause the syndrome of Campomelic Dysplasia (CD), Schafer AJ, et al. (1995); Foster JW, et al. (1996). CD is a semilethal osteochondrodysplasia characterized by skeletal malformation (bone dysmorphology) and frequently associated-autosomal male to female (46, XY females) sex-reversal, Cameron FJ, et al. (1996); conversely, overexpression of Sox9 can induce female to male (46, XX karyotype) sex-reversal, Huang B, et al. (1999).
Sox9 is an important member of the mammalian sex-determination genes that includes: sex determining gene, Y (SRY), anti-Müllerian hormone (AMH), Wilms tumor gene 1 (WT1), steroidogenic factor-1 (SF1), nuclear receptor DAX-1 (DAX1) and DMRT1. Sox9 promotes development of the male phenotype. It is expressed at high levels in male but not female genital ridges and sex cords of developing testis, Kent J et al. (1996). Expression is specific to the Sertolli cell lineage, not fetal germ cells. After the coelomic epithelial cells migrate into the gonad, there is first a decision to become interstitial or supporting cells; transient expression of SRY in the supporting cells determines their fate as Sertoli cells by up-regulating Sox9, Sekido R, et al. (2004). Several researchers have reported that Sox9 expression is sufficient to induce testis formation in mammals, Vidal VP, et al. (2001); Guo JK, et al. (2002); Qin Y, et al. (2005).
Sertoli cells are responsible for creating an immunoprivileged environment in the testis. Recognition of this quality has lead to an interest in using Sertoli cells in heterotopic sites to protect clinically important cells such as insulin producing pancreatic islet cells (to reverse type I diabetes) and dopaminergic (DA) neural cells (to treat Parkinson’s disease) from immune rejection. Sox9 has been proposed as a useful marker of Sertoli cells in these heterotropic transplants, Hemendinger RA, et al. (2002).
Chondrogenic competence results from the cooperative function of the subgroup-E Sox genes in response to BMP signaling; subsequently, Sox9 along with Sox5 and Sox6 execute and maintain the cartilage differentiation program, a multistep process that leads to endochondral bone formation, Lefebvre V, et al. (2001), Chimal-Monroy J, et al. (2003).
Sox9 becomes active in prechondrocytic mesenchymal condensations, and is maintained at high levels in differentiated chondrocytes. The regulation of Sox9 and its role in chondrogenesis is an area of intense research.
Sox9 is a general marker for basal cell carcinoma (BCC) where high expression is linked to enhanced malignancy and tumor invasiveness, Vidal VP, et al. (2008); Wang, H, et al. (2008).
Inappropriate ECM deposition is a hallmark of fibrosis. Since Sox9 induces the formation of extracellular matrix (ECM) components, especially collagen, a role for Sox9 in the regulation of the pathology of fibrosis has been suggested, Hanley KP, et al. (2008).
Sigma offers antibodies, shRNAs and other products useful for the study of the SOX9 gene.
References:
Akiyama H, et al. (2004) Essential role of Sox9 in the pathway that controls formation of cardiac valves and septa. Proc Natl Acad Sci U S A. 101: 6502-6507.
Barrionuevo F, et al. (2008) Sox9 is required for invagination of the otic placode in mice. Dev Biol. 317: 213-224.
Barrionuevo F, et al. (2006) Sox9 is required for notochord maintenance in mice. Dev Biol. 295: 128-140.
Bastide P, et al. (2007) Sox9 regulates cell proliferation and is required for Paneth cell differentiation in the intestinal epithelium. J Cell Biol. 178: 635-648.
Bi W, et al. (1999) Sox9 is required for cartilage formation. Nat Genet. 22: 85-89.
Cameron FJ, et al. (1996) A novel germ line mutation in SOX9 causes familial campomelic dysplasia and sex reversal. Hum Mol Genet. 5: 1625-1630.
Cheung M and Briscoe J. (2003) Neural crest development is regulated by the transcription factor Sox9. Development. 130: 5681-5693.
Chimal-Monroy J, et al. (2003) Analysis of the molecular cascade responsible for mesodermal limb chondrogenesis: Sox genes and BMP signaling. Dev Biol. 257: 292-301.
Cook AL, et al. (2005) Co-expression of SOX9 and SOX10 during melanocytic differentiation in vitro. Exp Cell Res. 308: 222-235.
Elluru RG and Whitsett JA. (2004) Potential role of Sox9 in patterning tracheal cartilage ring formation in an embryonic mouse model. Arch Otolaryngol Head Neck Surg. 130: 732-736.
Foster JW. (1996) Mutations in SOX9 cause both autosomal sex reversal and campomelic dysplasia. Acta Paediatr Jpn. 38: 405-411.
Guo JK, et al. (2002) Early gonadal development: exploring Wt1 and Sox9 function. Novartis Found Symp. 244: 23-31.
Hanley KP, et al. (2008) Ectopic SOX9 mediates extracellular matrix deposition characteristic of organ fibrosis. J Biol Chem. 283: 14063-14071.
Healy C, et al. (1999) Regulation and role of Sox9 in cartilage formation. Dev Dyn. 215: 69-78.
Healy C, et al. (1996) Expression of the chicken Sox9 gene marks the onset of cartilage differentiation. Ann N Y Acad Sci. 785: 261-272.
Hemendinger RA, et al. (2002) Identification of a specific Sertoli cell marker, Sox9, for use in transplantation. Cell Transplant. 2002;11(6): 499-505.
Hong CS, et al. (2005) Sox proteins and neural crest development. Semin Cell Dev Biol. 16: 694-703.
Huang B, et al. (1999) Autosomal XX sex reversal caused by duplication of SOX9. Am J Med Genet. 87: 349-353.
Kent J, et al. (1996) A male-specific role for SOX9 in vertebrate sex determination. Development. 122: 2813-2822.
Kobayashi A, et al. (2005) Sox9 in testis determination. Ann N Y Acad Sci. 1061: 9-17.
Kordes U, et al. (2005) Sox group E gene expression distinguishes different types and maturational stages of glial cells in developing chick and mouse. Brain Res Dev Brain Res. 157: 209-213.
Lefebvre V, et al. (2001) L-Sox5, Sox6 and Sox9 control essential steps of the chondrocyte differentiation pathway. Osteoarthritis Cartilage. 9 Suppl A: S69-75.
Lefebvre V and de Crombrugghe B. (1998) Toward understanding SOX9 function in chondrocyte differentiation. Matrix Biol. 16: 529-540.
Lincoln J, et al. (2007) Sox9 is required for precursor cell expansion and extracellular matrix organization during mouse heart valve development. Dev Biol. 305: 120-132.
Lioubinski O, et al. (2003) Expression of Sox transcription factors in the developing mouse pancreas. Dev Dyn. 227: 402-408.
Lynn FC, et al. (2007) Sox9 coordinates a transcriptional network in pancreatic progenitor cells. Proc Natl Acad Sci U S A. 104: 10500-10505.
Mitsiadis TA, et al. (1998) Expression of the transcription factors Otlx2, Barx1 and Sox9 during mouse odontogenesis. Eur J Oral Sci. 106: 112-116.
Moniot B, et al. (2004) SOX9 specifies the pyloric sphincter epithelium through mesenchymal-epithelial signals. Development. 131: 3795-3804..
Morais da Silva S, et al. (1996) Sox9 expression during gonadal development implies a conserved role for the gene in testis differentiation in mammals and birds. Nat Genet. 14: 62-68.
Mori-Akiyama Y, et al. (2007) Sox9 is required for invagination of the otic placode in mice. Dev Biol. 317: 213-224.
Nowak JA, et al. (2008) Hair follicle stem cells are specified and function in early skin morphogenesis. Cell Stem Cell. 3: 33-43.
Passeron T, et al. (2007) SOX9 is a key player in ultraviolet B-induced melanocyte differentiation and pigmentation. Proc Natl Acad Sci U S A. 104: 13984-13989.
Poché RA, et al. (2008) Sox9 is expressed in mouse multipotent retinal progenitor cells and functions in Müller Glial cell development. J Comp Neurol. 510: 237-250.
Schafer AJ, et al. (1995) The role of SOX9 in autosomal sex reversal and campomelic dysplasia. Philos Trans R Soc Lond B Biol Sci. 350: 271-277.
Sekido R, et al. (2004) SOX9 is up-regulated by the transient expression of SRY specifically in Sertoli cell precursors. Dev Biol. 274: 271-279.
Seymour PA, et al. (2007) SOX9 is required for maintenance of the pancreatic progenitor cell pool. Proc Natl Acad Sci U S A. 104: 1865-1870.
Stolt CC, et al. (2003) The Sox9 transcription factor determines glial fate choice in the developing spinal cord. Genes Dev. 17: 1677-1689.
Thomsen MK, et al. (2008a) Sox9 is required for prostate development. Dev Biol. 316: 302-311.
Vidal VP, et al. (2008) SOX9 expression is a general marker of basal cell carcinoma and adnexal-related neoplasms. J Cutan Pathol. 35: 373-379.
Vidal VP, et al. (2005) Sox9 is essential for outer root sheath differentiation and the formation of the hair stem cell compartment. Curr Biol. 15: 1340-1351.
Vidal VP, et al. (2001) Sox9 induces testis development in XX transgenic mice. Nat Genet. 28: 216-217.
Wang H, et al. (2008) SOX9 is expressed in human fetal prostate epithelium and enhances prostate cancer invasion. Cancer Res. 68: 1625-1630.
Wheatley S, et al. (1996) Aetiology of the skeletal dysmorphology syndrome campomelic dysplasia: expression of the Sox9 gene during chondrogenesis in mouse embryos. Ann N Y Acad Sci. 785: 350-352.
Zhou G, et al. (2006) Dominance of SOX9 function over RUNX2 during skeletogenesis. Proc Natl Acad Sci U S A. 103: 19004-19019.
Footnote: Gene Data Sources: HGNC, Entrez Gene, UniProt/Swiss-Prot, UniProt/TrEMBL, GDB, OMIM, GeneLoc, Ensembl.
Copyright © 2008 Sigma-Aldrich Co. Reproduction forbidden without permission.
Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. Best viewed in IE6 or higher.