Your Favorite Gene - Powered By Ingenuity
Sigma-Aldrich
Your Favorite Gene - Powered By Ingenuity
Find PKC Products
Interaction Network for PKC
PKC Details
Related Pathways
G-Protein Coupled Receptor Signaling
PPAR alpha / RXR alpha Activation
Synaptic Long Term Depression
View All Pathways
Activation of PKC through GPCR
Synonyms: AAG6, AI875142, LOC146784, MGC129900, MGC129901, PKC ALPHA, PKCA, PKRCA, PRKACA, PRKCA
Activation of PKC through GPCR
Protein Kinase C (PKC) refers to a multigene family of kinases classified as conventional: (PKCalpha; PKCbeta1 and PKCbeta2 (splice variants); and PKCgamma; novel: PKCdelta; PKCepsilon; PKCeta; PKCmu; PKCnu; and PKCtheta; and atypical: PKCiota/lamda and PKCzeta isoforms. The conventional PKC isoforms are activated by calcium (Ca2+) and sn-1,2-diacylglycerol (DAG), whereas the novel PKC isoforms are activated by DAG, but not calcium. Both Ca2+ and DAG are signaled by the actions of phospholipaseCbeta (PLCβ). PLCβ is activated by the Gαq (Galphaq) subunit of trimeric G-proteins linked to G-protein coupled receptors (GPCRs). GPCRs provide a wide range of specificity for agonist stimulation of conventional and novel PKC mediated cell signaling.
Activated PLCb or PLCg cleaves the inositol substrate phosphatidyl-inositol 4,5 bisphosphate (PIP2) to yield diacylglycerol and inositol 1,4,5-trisphosphate (IP3). DAG is a direct activator of conventional and novel PKCs. Calcium is released from the endoplasmic reticulum (ER) by IP3 and helps localize conventional PKCs to the cell membrane. PKC mediates CPI phosphorylation, which inhibits myosin phosphatase. The accumulation of phosphorylated myosin supports platelet contraction and secretion. The MEK kinase, Raf1 can be phosphorylated by conventional and novel PKC isotypes alpha and eta, but not PKCzeta. Activation of Raf1 leads to activation of ERK1/2 regulated genes. Various PKCs regulate cell survival through the release of NFkBs
References:
  1. Schonwasser D.C., et. al. (1998) Activation of the mitogen-activated protein kinase/extracellular signal-regulated kinase pathway by conventional, novel, and atypical protein kinase C isotypes. Mol. Cell Biol. 18, 790-798.
  2. Shirai Y., and Saito N. (2002) Activation mechanisms of protein kinase C: maturation, catalytic activation, and targeting. J. Biochem. (Tokyo). 132, 663-668.
  3. Watanabe Y., et. al. (2001) Protein kinase C-catalyzed phosphorylation of an inhibitory phosphoprotein of myosin phosphatase is involved in human platelet secretion. Blood. 97, 3798-3805.
Back to top
Content for this page is provided by Dennis R. Conrad, Ph.D., a Life Science industry consultant with over 25 years of experience in the formulation and optimization of cell culture media. Dr. Conrad's email address is biomediaexpert@earthlink.net