|
Find AKT1 Products
Interaction Network for AKT1
AKT1 Details
|
|
Related Pathways
PI3K / AKT Signaling Fc Epsilon RI Signaling Hypoxia Signaling in the Cardiovascular System View All Pathways
|
|
|
| Synonyms: AKT, AKT1, MGC99656, PKB, PKB-ALPHA, PKB/AKT, PRKBA, Protein kinase B, RAC, RAC-ALPHA, Thymoma viral proto-oncogene 1 |
Akt Signaling
The protein kinase B, PKB, family of proteins, also called Akt or RAC, are serine/threonine protein kinases involved in the regulation of cell survival; cell cycle progression; protein synthesis; glucose/glycogen metabolism; and a growing list of specialized cell function. Akt/PKB exists as three isoforms: AKT1 (PKB); AKT2 (PKBbeta); and AKT3 (PKBgamma). Akt/PKB is activated by phosphorylation. The principle Akt/PKB kinase is 3-phosphoinositide dependent protein kinase-1 (PDK1 (PKPK1)). Akt/PKB is dephosphorylated by protein phosphatase 2A (PP2A). Akt/PKB is maintained in an active phosphorylated state by the ligand occupation of various cytokine and growth factor receptors. Ligand-withdrawal or down-regulation of Akt/PKB activators leads to apoptosis.
Akt/PKB mediates cell survival signaling by phosphorylating several factors involved in apoptosis machinery including: caspase-9, BCL2-antagonist of cell death (BAD), forkhead in rhabdomyosarcoma-like 1 (FKHRL1); IκB kinase, IKKalpha (an NFkappaBp65 activator). Akt/PKB upregulates protein synthesis by turning off constitutively active glycogen synthase kinase-3, GSK3; and turning on mTOR (TORC1). Akt/PKB promotes cell-cycle progression by phosphorylating cyclin-dependent kinase (CDK) inhibitors; p27(Kip1) and p21(CIP1). Akt/PKB attenuates the action of the second messenger cAMP by activating the cAMP phosphodiesterases-3B (PDE-3B). Akt/PKB promotes the uptake of glucose by promoting the translocation of the GLUT-4 glucose transporter to the cell membrane.
|
|
References:
- Downward, J. (1988) Mechanisms and consequences of activation of protein kinase B/Akt. Curr. Opin. Cell Biol. 10, 262-267.
- Jimenez, C. et. al. (2002) The p85 regulatory subunit controls sequential activation of phosphoinositide 3-kinase by Tyr kinases and Ras. J. Biol. Chem. 277(44):41556-41562.
- Kitamura, T. et. al. (1999) Insulin-induced phosphorylation and activation of cyclic nucleotide phosphodiesterase 3B by the serine-threonine kinase Akt. Mol. Cell Biol. 19, 6286-6296.
- Ruggero, D. and Sonenberg, N. (2005) The Akt of translational control. Oncogene. 24, 7426-34.
- Testa, J. R. and Tsichlis, P. N. (2005) AKT signaling in normal and malignant cells. Oncogene. 7391-7393.
- Zhou, X. M. et. al. (2000) Growth factors inactivate the cell death promoter BAD by phosphorylation of its BH3 domain on Ser155. J. Biol. Chem. 275, 25046-25051.
|
| Back to top |
Content for this page is provided by Dennis R. Conrad, Ph.D., a Life Science industry consultant with over 25 years of experience in the formulation and optimization of cell culture media. Dr. Conrad's email address is biomediaexpert@earthlink.net
|