Your Favorite Gene - Powered By Ingenuity
Sigma-Aldrich
Your Favorite Gene - Powered By Ingenuity
Find TNF Products
Interaction Network for TNF
TNF Details
Related Pathways
Apoptosis Signaling
Death Receptor Signaling
Acute Phase Response Signaling
View All Pathways
Apoptosis Through Death Receptors
Synonyms: AT-TNF, DIF, Differentiation-induced Factor, MGC124630, MGC151434, RATTNF, TMTNF, TNF, TNF-ALPHA, TNFA, Tnfsf1a, TNFSF2
Apoptosis Through Death Receptors
Apoptosis is a normal cellular process involving physiologically relevant cell death. Apoptosis is initiated and promoted by a wide variety of intra- and extra-cellular stimuli. Extra-cellular induced apoptosis can be triggered by cell surface “death” receptors that possess cytoplasmic death domains (FADD, TRADD), typified by members of the tumor necrosis factor receptor superfamily: tumor necrosis factor (TNF) receptor 1 (TNFR-1), TNF-related apoptosis-inducing ligand receptor 1 (TRAILR-1), TRAILR-2, death receptor 3 (DR3/Wsl-1/APO-3/TRAMP/LARD), DR6, ectodermal dysplasia receptor (EDAR), p75-nerve growth factor receptor (p75-NGFR) and Fas/Apo-1/CD95 or by the action of the cytotoxic T-cell proteins such as granzyme-B and perforin.
Apoptotic events are propagated by members of a family of regulated proteolytic enzymes called caspases. The caspases most often associated with apoptosis are caspases 2, 3, 6, 7, 8, 9 and 10. Caspase cascades are composed of upstream (initiator) caspases (2, 8, 10), which are activated by death receptor signalosomes (DISC); caspase 9, which is activated by the mitochondrial cytochrome C/Apaf-1 derived apoptosome; and downstream (effector) caspases (3, 6, 7) that cleave proteins involved in programmed cell death events.
Caspase 2, caspase 8 and caspase 10 are activated through interaction with death receptor cytoplasmic death-inducing signaling complexes (DISC)) that contain FAS-associating death domain (FADD) or TNFR1-associated death domain (TRADD). These receptors are activated by ligands that are involved with signaling cell death to support cell selection, tissue homeostasis, morphogenesis, and host defense in multi-cellular organisms. Typical death receptor ligands include Fas ligand (FasL), tumor necrosis factor alpha (TNFα), NF-related apoptosis inducing ligand (TRAIL), TNF-related weak inducer of apoptosis TWEAK, TNF ligand-related molecule 1 (TL1A) and nerve growth factor (NGF).
References:
  1. Golks, A. et. al. (2006) The role of CAP3 in CD95 signaling: new insights into the mechanism of procaspase-8 activation. Cell Death Differ. 13, 489-498.
  2. Gomez-Angelats, M. and Cidlowski, J.A. (2001) Protein kinase C regulates FADD recruitment and death-inducing signaling complex formation in Fas/CD95-induced apoptosis. J. Biol. Chem. 276, 44944-44952.
  3. Imtiyaz, H.Z. et. al. (2005) Structural requirements for signal-induced target binding of FADD determined by functional reconstitution of FADD deficiency. J. Biol. Chem. 280, 31360-31367.
  4. Sandu, C. et. al. (2005) A mechanism for death receptor discrimination by death adaptors. J. Biol. Chem. 280, 31974-31980.
  5. Wajant, H. (2003) Death receptors. Essays Biochem. 39, 53-71.
Back to top
Content for this page is provided by Dennis R. Conrad, Ph.D., a Life Science industry consultant with over 25 years of experience in the formulation and optimization of cell culture media. Dr. Conrad's email address is biomediaexpert@earthlink.net