Your Favorite Gene - Powered By Ingenuity
Sigma-Aldrich
Your Favorite Gene - Powered By Ingenuity
Find ADCY Products
Interaction Network for ADCY
ADCY Details
Related Pathways
cAMP-mediated Signaling
G-Protein Coupled Receptor Signaling
Dopamine Receptor Signaling
View All Pathways
Cyclic AMP Pathway
Synonyms: 3',5'-cyclic AMP synthetase, ADCY, ADENYL CYCLASE, Adenylate Cyclase, Adenylyl Cyclase, Adenylyl Cyclase protein, ATP diphosphate-lyase (cyclizing)
Cyclic AMP Pathway
Cyclic AMP (cAMP) is an important second messenger. Cyclic AMP mediates downstream activity of protein kinase A (PKA), exchange protein directly activated by cAMP (EPAC) and calcium flux through L-type calcium channels. Calcium is an important second messenger in cell signaling.
The level of cAMP is regulated by the action of a family of adenylate (adenylyl) cyclases that respond to G-protein subunits, especially Galphas and Galphai, which increase and decrease AC activity respectively. Specific heterotrimeric G-proteins are associated with a wide range of G-protein coupled receptors. Consequently, the level of cAMP can be modulated up or down depending upon which GPCRs are activated. Adenylyl cyclase activity is sometimes regulated by calcium and/or protein kinase C. The effects of calcium or PKC on adenylyl cyclase are isoform specific. The level of cAMP is regulated by its hydrolysis to 5’-AMP by cAMP phosphodiesterases (PDE). Various growth factor tyrosine kinase receptors activate PDE via a pathway that follows the receptor tyrosine kinase-PI3K-PDK-Akt/PBD-PDE axis. PDE is a direct substrate of AKT.
References:
  1. de Rooij J., et. al. (1998) Epac is a Rap1 guanine-nucleotide-exchange factor directly activated by cyclic AMP. Nature. 396, 474-477.
  2. Kitamura T., et. al. (1999) Insulin-induced phosphorylation and activation of cyclic nucleotide phosphodiesterase 3B by the serine-threonine kinase Akt. Mol Cell Biol. 19, 6286-6296.
  3. Smit M.J., and Iyengar R. (1998) Mammalian adenylyl cyclases. Advances in Second Messenger and Phosphoprotein Res 32, 1-21.
Back to top
Content for this page is provided by Dennis R. Conrad, Ph.D., a Life Science industry consultant with over 25 years of experience in the formulation and optimization of cell culture media. Dr. Conrad's email address is biomediaexpert@earthlink.net