Your Favorite Gene - Powered By Ingenuity
Sigma-Aldrich
Your Favorite Gene - Powered By Ingenuity
Find ERBB2 Products
Interaction Network for ERBB2
ERBB2 Details
Related Pathways
Neuregulin Signaling
Axonal Guidance Signaling
View All Pathways
ErbB2-ErbB3 Heterodimers
Synonyms: C-ERBB2, c-neu, CD340, EGFR2, ERBB2, ERBB2 RECEPTOR, HER-2, HER2/NEU, mKIAA3023, NEU, NGL, p185erbb2, p185HER2, P185NEU, TKR1
ErbB2-ErbB3 Heterodimers
ErbB2 (HER2) and ErbB3 (HER3) along with ErbB1 (EGFR) and ErbB4 (HER4) are members of the EGF receptor family. ErbB3 binds members of the neuregulin (neu differentiation factor (NDF)) family, but lacks intrinsic tyrosine kinase activity. ErbB2 lacks a neuregulin binding domain, but has a potent tyrosine kinase activity and is the most oncopotent of the Erb receptors. Ligand bound ErbB3 dimerizes as part of its activation. Whenever ErbB2 is expressed on the cell surface together with ErbB3, it preferentially forms ErbB2:ErbB3 dimers that become activated via trans-phosphorylation. Phosphorylated ErbB2:ErbB3 dimers provide docking sites for factors involved in the initiation of cell signaling pathways, especially those involved in cell survival, cell growth and cell transformation. These include the Ras/Raf-1/MEK/ERK mitogenic; the phophoinositide 3-kinase (PI3K)/PDK1/Akt(PKB) anti-apoptotic; and JAK/STAT mitogenic pathways.
ErbB2 is the preferred heterodimerization partner of the other erb (HER) receptors, including erbB3. The expression of erbB2 on the cell surface of normal tissue is generally limited. Overexpression of erbB2 on the cell surface along with erbB3 increases the formation of erbB2:erbB3 heterodimers. ErbB2 containing heterodimers are potent signal mediators that promote prolonged survival and mitogenesis signaling because they release agonists slowly and are relatively resistant to down regulation. Prolonged overexpression of ErbB2 has been linked to a number of epithelial cancers, such as breast cancer. The anti-cancer drug Herceptin (trastuzumab) is believed to work by directing Cbl-dependent endocytosis and degradation of ErbB2 containing heterodimers.
References:
  1. Harari, D. and Yarden, Y. (2000) Molecular mechanisms underlying ErbB2/HER2 action in breast cancer. Oncogene. 19, 6102-6114.
  2. Tzahar, E. et. al. (1996) A hierarchical network of interreceptor interactions determines signal transduction by Neu differentiation factor/neuregulin and epidermal growth factor. Mol. Cell Biol. 16, 5276-5287.
  3. Yarden, Y. (2001) Biology of HER2 and its importance in breast cancer. Oncology. 61, 1-13.
Back to top
Content for this page is provided by Dennis R. Conrad, Ph.D., a Life Science industry consultant with over 25 years of experience in the formulation and optimization of cell culture media. Dr. Conrad's email address is biomediaexpert@earthlink.net