Your Favorite Gene - Powered By Ingenuity
Sigma-Aldrich
Your Favorite Gene - Powered By Ingenuity
Find FLT3 Products
Interaction Network for FLT3
FLT3 Details
Related Pathways
None - Check for Updates
FLT3 Signaling
Synonyms:B230315G04, CD135, FLK2, FLT3, Ly72, MGC124377, MGC124378, STK1
FLT3 Signaling
Flt-3/Flk-2 ligand (FL, Flt3-L, fms-like tyrosine-kinase 3 ligand) is a transmembrane 30 kDa glycoprotein that activates the Flt3 (CD135, FLK2) receptor. The Flt-3 receptor is a member of the class III subfamily of receptor tyrosine kinases (RTK) that includes the PDGF receptor; CSFIR (colony stimulating factor 1 receptor) (FMS) and KIT (stem cell factor receptor, steel locus factor receptor, SLFR). Flt-3 is expressed primarily in hematopoietic stem cells (myelomonocytic, monocytic, pre-G and pro-B lineages) and acute leukemia T, B and myeloid blast cells. Tissues that contain Flt-3 expressing cells include bone marrow, thymus, spleen, liver, testis, placenta, CNS and lymph nodes.
FL (Flt3-L), which is structurally related to M-CSF (CSF-1) and SCF (KL), is expressed in a broad range of cell types. FL is biologically active on the cell surface or in a cleaved soluble form. The binding of FL to the Flt-3 receptor typically produces modest growth and differentiation effects. However, in the presence of a variety of cytokines FL binding to Flt-3 promotes strong growth and differentiation effects. Fl synergizes with GM-CSF, IL-1, IL-3, IL-6, SCF (KL), M-CSF, G-CSF, IL-7 and TPO.
Flt-3 ligand, in synergy with other cytokines, stimulates the growth and differentiation of a number of important cell populations. Activation of Flt-3 stimulates the growth and differentiation of very early stem cells involved in hematopoiesis, myelopoiesis, erythropoiesis and megakaryopoiesis. Flt-3 activation mobilizes hematopoietic progenitor cells into peripheral blood and induces proliferation of myeloid and dendritic cells (DC). Dysregulation of Flt-3 has been associated with acute leukemia(s) involving myeloid (AML), T, and B blast cell(s).
References:
  1. Meyer, C. and Drexler, H. G. (1999) FLT3 ligand inhibits apoptosis and promotes survival of myeloid leukemia cell lines. Leuk Lymphoma. 32, 577-581.
  2. Rosenzwajg, M. et. al. (1998) The influence of interleukin (IL)-4, IL-13, and Flt3 ligand on human dendritic cell differentiation from cord blood CD34+ progenitor cells. Exp Hematol. 26, 63-72.
  3. Shurin, M.R. et. al. (1998) FLT3: receptor and ligand. Biology and potential clinical application. Cytokine Growth Factor Rev. 9, 37-48.
  4. Gringel, S. et. al. (2004) Nuclear fibroblast growth factor-2 interacts specifically with splicing factor SF3a66. J. Biol. Chem. 385, 1203-1208.
  5. Zheng, R. and Small, D. (2005) Mutant FLT3 signaling contributes to a block in myeloid differentiation. Leuk Lymphoma. 46, 1679-1687.
Back to top
Content for this page is provided by Dennis R. Conrad, Ph.D., a Life Science industry consultant with over 25 years of experience in the formulation and optimization of cell culture media. Dr. Conrad's email address is biomediaexpert@earthlink.net