Your Favorite Gene - Powered By Ingenuity
Sigma-Aldrich
Your Favorite Gene - Powered By Ingenuity
Find LBP Products
Interaction Network for LBP
LBP Details
Related Pathways
LPS / IL-1 Mediated Inhibition of RXR Function
Actin Cytoskeleton Signaling
Hepatic Fibrosis / Hepatic Stellate Cell Activation
View All Pathways
LPS Stimulated MAPK Signaling
Synonyms: LBP, LIPOPOLYSACCHARIDE-BINDING PROTEIN, Ly88, MGC124626, MGC22233
LPS Stimulated MAPK Signaling
Lipopolysaccharides (LPS) are major cell wall constituents of Gram-negative bacteria that bind to LPS-binding proteins (LBP) in plasma. The LPS-LBP complex interacts with CD14 to form a ternary complex, LPS:LBP:CD14, which transfers LPS to the toll-like receptor 4 (TLR4) accessory protein MD2 complex. This leads to activation of toll-like receptor-4. LPS activated TRL4 activates the NFkappaB pathway and all three MAPK pathways; ERK, JNK/SAPK and p38. Activation of the NFkappB pathway is complex. It requires activation of phosphatidylinositol-3-kinase (PI3K) upstream of IkappaBalpha degradation. A second NFkappaB activation pathway follows the sequence: TLR4, MyD88, TIR, IRAK1, TRAF6, NIK, IKK, IkappaB, NFkappaB.
ERK is activated by LPS activated TLFR4 through Raf1. LPS activated TLFR4 activates the Raf1/MEK/ERK pathway both by p21Ras-GTPase-dependent and Ras-independent mechanisms. Lipopolysaccharides (LPS) are major cell wall constituents of Gram-negative bacteria that bind to LPS-binding proteins (LBP) in plasma. The LPS-LBP complex interacts with CD14 to form a ternary complex, LPS:LBP:CD14, which transfers LPS to the toll-like receptor 4 (TLR4) accessory protein MD2 complex. This leads to activation of toll-like receptor-4. LPS activated TRL4 activates the NFkappaB pathway and all three MAPK pathways; ERK, JNK/SAPK and p38. Activation of the NFkappB pathway is complex. It requires activation of phosphatidylinositol-3-kinase (PI3K) upstream of IkappaBalpha degradation. A second NFkappaB activation pathway follows the sequence: TLR4, MyD88, TIR, IRAK1, TRAF6, NIK, IKK, IkappaB, NFkappaB. ERK is activated by LPS activated TLFR4 through Raf1. LPS activated TLFR4 activates the Raf1/MEK/ERK pathway both by p21Ras-GTPase-dependent and Ras-independent mechanisms.
References:
  1. Bjorkoy, G. et. al. (1995) Evidence for a bifurcation of the mitogenic signaling pathway activated by Ras and phosphatidylcholine-hydrolyzing phospholipase C. J. Biol. Chem. 270, 21299-21306.
  2. Dauphinee, S.M. and Karsan, A. (2006) Lipopolysaccharide signaling in endothelial cells. Lab Invest. 86, 9-22.
  3. Mas, V.M. et. al. (2003) Protein kinase Czeta mediated Raf-1/extracellular-regulated kinase activation by daunorubicin. Blood. 101, 1543-1550.
Back to top
Content for this page is provided by Dennis R. Conrad, Ph.D., a Life Science industry consultant with over 25 years of experience in the formulation and optimization of cell culture media. Dr. Conrad's email address is biomediaexpert@earthlink.net