|
Find MAPK3 Products
Interaction Network for MAPK3
MAPK3 Details
|
|
Related Pathways
ERK / MAPK Signaling SAPK / JNK Signaling p38 MAPK Signaling View All Pathways
|
|
|
| Synonyms: ERK-1, Esrk1, HS44KDAP, HUMKER1A, Mapk p44, MAPK3, Mapkapk3, MGC20180, P44, p44-erk, P44ERK1, P44MAPK, PRKM3 |
MAPK Family Pathway
The MAPK (mitogen-activated protein kinase) super-family is composed of three major sets of kinases: the extracellular-receptor kinases (ERK) include ERK1; ERK2; ERK3/ERK4, ERK5, and two types of MAPK-related kinases that respond to cellular stress and inflammatory signal: the c-Jun N-terminal kinases/stress-activated protein kinases (JNK/SAPK) which include JNK1, JNK2 and JNK3 and the p38 MAPKinases: p38alpha, p38beta2, p38gamma and p38delta. Activation of specific MAPkinases involves highly regulated and modulated cascades of phosphorylation events mediated by sequential and concerted activation of upstream kinases. ERKs are phosphorylated by members of the MEK family; JNK/SAPKs and p38 MAPKs are phosphorylated by SEKs and MKKs. These are MAPK-kinases (MAP2Ks). The MAP2Ks are phosphorylated by MAPK-kinase kinases (MAP3Ks). A wide variety of kinases can provide MAP3K function to the MAPK pathways. MAP3Ks are activated by a wide range of kinase activities modulated by cell receptors, especially through small GTPases such as the p21Ras super-family composed of Ras, Rho and Rab families. Cellular and context specificity is achieved via a wide variety of guanine nucleotide exchange factors (GEFs) that activate specific GTPases in response to closely linked receptor signal inputs.
Upon activation, specific MAPKs activate specific MAPK-activated protein kinases (MAPKAPKs) which depending upon cell phenotype and context phosphorylate cytoplasmic and nuclear substrates that affect cell processes both directly and through gene expression. MAPKs can also directly phosphorylate gene transcription factors such as Elk1, c-Jun and ATF2. The phosphorylation of MAPKAPK substrates and gene transcription factors induces a wide variety of cellular processes involved with proliferation, differentiation, development, cell division, oncogenesis and survival.
|
|
References:
- Brunet, A. and Pouyssegur, J. (1997) Mammalian MAP kinase modules: how to transduce specific signals. Essays Biochem. 32, 1-16.
- Rubinfeld, H. and Seger, R. (2005) The ERK cascade: a prototype of MAPK signaling. Mol. Biotechnol. 31, 151-174.
- Strniskova, M. et. al. (2002) Mitogen-activated protein kinases and their role in regulation of cellular processes. Gen. Physiol. Biophys. 21, 231-255.
- Yoon, S. and Seger, R. (2006) The extracellular signal-regulated kinase: multiple substrates regulate diverse cellular functions. Growth Factors. 24, 21-44.
|
| Back to top |
Content for this page is provided by Dennis R. Conrad, Ph.D., a Life Science industry consultant with over 25 years of experience in the formulation and optimization of cell culture media. Dr. Conrad's email address is biomediaexpert@earthlink.net
|