Your Favorite Gene - Powered By Ingenuity
Sigma-Aldrich
Your Favorite Gene - Powered By Ingenuity
Find NOS1 Products
Interaction Network for NOS1
NOS1 Details
Related Pathways
Synaptic Long Term Depression
Amyotrophic Lateral Sclerosis Signaling
Arginine and Proline Metabolism
View All Pathways
nNOS Signaling at Neuronal Synapses
Synonyms:BNOS, IHPS1, mtNOS, NNOS, nNOS-2, NO, NOS, NOS-I, NOS1
nNOS Signaling at Neuronal Synapses
Nitric oxide (NO) is a gaseous low reactivity free radical with the ability to freely cross cell membranes. In neural tissues it is a mediator of synaptic signaling, regulator of synaptic architecture and neural plasticity, promoter of neurotoxicity and a wide array of other effects that are cell type and context specific. NO can modulate the release of neurotransmitters such as acetylcholine, catecholamines, excitatory and inhibitory amino acids, serotonin, histamine and adenosine. NO-dependent release of neurotransmitters is generally linked to a guanylyl cyclase-coupled NO receptors (NOGCR)) which catalyzes cGMP formation and activation of cGMP-dependent protein kinase (PKG). NO is involved in synaptic plasticity phenomena such as long-term potentiation (LTP) and depression (LTD), and synaptogenesis. Supraphysiological NO levels induced by hyperexcitatory stimulation of N-methyl-D aspartate (NMDA) receptors by glutamate mediate neurotoxicity (excitotoxicity).
Nitric oxide (NO) production in neural cells is coupled to the stimulation of N-methyl-D-aspartate (NMDA) receptors by glutamate through calcium activation of nitric oxide synthase (NOS). Nitric oxide synthases (EC 1.14.13.39) are calmodulin binding enzymes that convert L-arginine and oxygen into citrulline and NO. Neural nitric oxide synthase (nNOS, NOS-1, NOS-I, NOS type I) isoform alpha (nNOSα, nNOSalpha) contains a PDZ domain that facilitates its binding to NMDA receptors through postsynaptic density protein 95 (PSD95). Tethering of nNOSalpha to NMDAR regulated calcium channels facilitates its activation by glutamate induced calcium flow into the cell. Under normal glutamate excitatory conditions, the flow of calcium is controlled and NO mediates intended signaling effects. Pathological conditions often lead to over-stimulation of the NMDA receptors by glutamate followed by elevation of NO to levels that induce neurotoxicity (excitotoxicity). Excitotoxicity is implicated in many neurodegenerative conditions including ischemia, traumatic brain injury, Parkinson's disease, Huntingdon's disease, Alzheimer's disease, and amyotrophic lateral sclerosis (ALS).
References:
  1. Aarts, M.Y. et. al. (2002) Treatment of ischemic brain damage by perturbing NMDA receptor- PSD-95 protein interactions. Science. 298, 846–850.
  2. Cao. J. et. al. (2005) The PSD95-nNOS interface: a target for inhibition of excitotoxic p38 stress-activated protein kinase activation and cell death. J. Cell Biol. 168, 117-126.
  3. Moreno-Lopez, B. and Gonzalez-Forero, D. (2006) Nitric oxide and synaptic dynamics in the adult brain: physiopathological aspects. Rev. Neurosci. 17, 309-357.
  4. Sunico, C.R. et. al. (2005) Nitric-oxide-directed synaptic remodeling in the adult mammal CNS. J. Neurosci. 25, 1448-1458.
Back to top
Content for this page is provided by Dennis R. Conrad, Ph.D., a Life Science industry consultant with over 25 years of experience in the formulation and optimization of cell culture media. Dr. Conrad's email address is biomediaexpert@earthlink.net