Your Favorite Gene - Powered By Ingenuity
Sigma-Aldrich
Your Favorite Gene - Powered By Ingenuity
Find RAN Products
Interaction Network for RAN
RAN Details
 Related Pathways
None - Check for Updates
Ran Pathway
Synonyms: ARA24, Gsp1, GTPase Ran, RAN, RANGTPASE, RAS-like, family 2, locus 9, Rasl2-9, TC4
RAN Pathway
Ran, (ARA24, TC4) is a Ras-like GTPase involved in the formation of mitotic spindles via stimulation of microtubule polymerization and nuclear envelope and nuclear pore assembly during cell mitosis and in nuclear-cytoplasmic import/export of proteins and RNA during cell interphase. Spindle, nuclear envelope and nuclear pore assembly involve RICCI-dependent and RICCI-independent Ran associations with chromatin.
The directionality of facilitated transport of proteins and RNA across the nuclear envelope by importin and exportin molecules through nuclear pore complexes (NPC) is regulated by the GTP-bound to GDP-bound Ran gradient. GTP-bound Ran facilitates the disassembly (unloading of cargo) from transport complexes inside the nucleus. Nuclear GTP-bound Ran complexes with exporter complexes and is transported to the cytoplasm. GTP hydrolysis in the cytoplasm releases the exported cargo (proteins or RNA). The resulting GDP-bound Ran is escorted back into the nucleus by RanGDI, nuclear transport factor 2 (NTF2). In the nucleus, GDP is exchanged for GTP by the chromatin bound RanGEF, RICCI. Consequently, NTF2 and RICCI work together to increase the level of nuclear GTP-bound Ran. NTF2 activity enhances nuclear import. Nuclear export is driven by the activity of cytoplasmic RanGAP, which hydrolyses GTP-bound to Ran to GDP
References:
  1. Clarke, P. R. and Zhang, C. (2001) Ran GTPase: a master regulator of nuclear structure and function during the eukaryotic cell division cycle? Trends Cell Biol. 11, 366-371.     
  2. Minakhina, S. et. al. (2005) Crosstalk between the actin cytoskeleton and Ran-mediated nuclear transport. BMC Cell Biol. 6, 32.    
  3. Sazer, S. (2005) The view from Awaji island: past, present, and future of RCC1 and the Ran GTPase system. Dev. Cell. 9, 729-733. 
Back to top
Content for this page is provided by Dennis R. Conrad, Ph.D., a Life Science industry consultant with over 25 years of experience in the formulation and optimization of cell culture media. Dr. Conrad's email address is biomediaexpert@earthlink.net