Your Favorite Gene - Powered By Ingenuity
Sigma-Aldrich
Your Favorite Gene - Powered By Ingenuity
Find Rap1 Products
Interaction Network for Rap1
Rap1 Details
 Related Pathways
Synaptic Long Term Potentiation
cAMP-mediated Signaling
ERK/MAPK Signaling
View All Pathways
Rap1 Pathway
Synonyms: AI848598, C21KG, EG668838, G-22K, KREV-1, MGC102329, RAP1, RAP1A, SMGP21
Rap1 Pathway
Rap family members (Rap1A, Rap1B, Rap2A, Rap2B, and Rap2C) are small Ras-like GTPases that regulate cell proliferation, differentiation, apoptosis (PDA activities) and adhesion mechanisms. Raps are regulated by specific RapGEF, RapGAP, and RapGDI molecules, and their location relative to specific upstream and downstream effector molecules. These factors integrate to provide specificity to Rap actions. They also mediate cross-talk among signaling pathways.
Rap1 (Ras-proximate-1) regulates PDA activities through interaction with two MAP3K effectors of the MEK/ERK MAP-kinase pathway, Raf-1 and B-Raf. Rap1 activates the MEK/ERK pathway via interaction with BRaf and inhibits MEK/ERK activity by binding Raf-1. As an inhibitor of the Raf-1 pathway Rap-GTPase is an antagonist of Ras-GTPase. Rap1 activity that regulates ERK activity through Raf-1 and B-Raf is associated primarily with the plasma membrane (PM) and it is regulated by GEFs represented by C3G (Crk SH3 domain Guanine nucleotide exchanger) and Smg GDS. The Rap1 ERK signaling pathway responds to Src kinases and cAMP via activation of PKA. Rap1 involved with ERK signaling is linked to cell receptors via adaptor and scaffold proteins such as c-Cbl and CrkL.
Rap1 regulates cell adhesion mechanisms through effector molecules such as the integrins and cadherins. RIAM (Rap1-GTP-interacting adaptor molecule) links Rap1 to integrin activation. The Rap1 regulation of cell adhesion processes involves a family of GEFs that are directly activated by cAMP: Epac 1(cAMP-GEFI) and Epac 2 (cAMP-GEFII). The activation of Rap1 involved with intergrin and cadherins activity is topologically regulated by the localized expression of Epacs and effectors of Epac activation.
Various GPCR receptor signals modulate the activity of Rap through trimeric G-protein subunits. For example, unactivated Galpha(o) stimulates Rap1-dependent ERK activation by sequestering Rap1GAP. Whereas Galpha(s) and Galpha(i) regulate the levels of cAMP through adenylyl cyclase activation and inhibition, respectively.
References:
  1. Caron, E. (2003) Cellular functions of the Rap1 GTP-binding protein: a pattern emerges. J. Cell Sci. 116, 435-40.     
  2. Stork, P. J. and Schmitt, J. M. (2002) Crosstalk between cAMP and MAP kinase signaling in the regulation of cell proliferation. Trends Cell Biol. 12, 258-266.   
  3. Stork, P. J. (2003) Does Rap1 deserve a bad Rap? Trends Cell Biol. 28, 267-275.
  4. Stork, P. J. and Dillon, T. J. (2005) Multiple roles of Rap1 in hematopoietic cells: complementary versus antagonistic functions. Blood. 106, 2952-2961.
  5. Wang, Z. et. al. (2006) Rap1-mediated activation of extracellular signal-regulated kinases by cyclic AMP is dependent on the mode of Rap1 activation. Mol. Cell Biol. 26, 2130-2145.
Back to top
Content for this page is provided by Dennis R. Conrad, Ph.D., a Life Science industry consultant with over 25 years of experience in the formulation and optimization of cell culture media. Dr. Conrad's email address is biomediaexpert@earthlink.net