|
Find ARHGDIA Products
Interaction Network for ARHGDIA
ARHGDIA Details
|
|
Related Pathways
Regulation of Actin-based Motility by Rho View All Pathways
|
|
|
| Synonyms: 5330430M07Rik, ARHGDIA, Gdi-1, GDIA1, MGC101921, MGC117248, MGC7904, MGC94886, RHO GDI ALPHA, RhoDGI, RHOGDI, RHOGDI-1 |
RhoGDI Pathway
RhoGDI (Rho GDP-dissociation inhibitor) is a small family of proteins which includes the ubiquitous GDP dissociation inhibitor 1 (RhoGDIα) (RABGD1A) (Oligophrenin 2); RhoGDI-2 (RhoGDIβ) (D4), found primarily in haematopoietic cells, and RhoGDIgamma (RhoGDI-3), a membrane-anchored factor found in brain, pancreas, lung, kidney and testis. RhoGDIs regulate Rho subfamily GTPases including Rho, Rac and Cdc42. RhoGDIs have two major functions. They extract prenylated GDP:Rho-GTPases from membranes and sequester them as cytosolic complexes or transfer them to other membranes. They prevent the exchange of GTP for GDP on the Rho-GTPases by guanine nucleotide exchange factors, GEF. RhoGDIs control the action and status of the Rho-GTPases when they are not involved in the promotion of specific effector promoting activities.
Rho, Rac and Cdc42 are important molecular switches involved in the regulation of cytoskeletal structures that affect or enable the regulation of cell morphology, movement, attachment, phagocytosis and cytokinesis. Rho, Rac and Cdc42 are known to control these cell processes via the regulation of the formation of stress fibers; membrane ruffles/lamellipodia and filopodia, respectively. They are also involved in the regulation of gene transcription, cell cycle progression; vesicular trafficking and specialized functions such as activation of the NADPH oxidase complex in phagocytes (Rac1).
|
|
References:
- Brunet N. et. al. (2002) RhoGDI-3 regulates RhoG and targets this protein to the Golgi complex through its unique N-terminal domain. Traffic. 3, 342–358.
- Dovas, A. and Couchman, J. R. (2005) RhoGDI: multiple functions in the regulation of Rho family GTPase activities. Biochem. J. 390, 1-9.
- Olofsson, B. (1999) Rho guanine dissociation inhibitors: pivotal molecules in cellular signalling. Cell Signal. 11, 545-554.
- Schmidt, A. and Hall, A. (2002) Guanine nucleotide exchange proteins for Rho GTPases: turning on the switch. Genes Dev. 16, 1587–1609.
|
Back to top |
Content for this page is provided by Dennis R. Conrad, Ph.D., a Life Science industry consultant with over 25 years of experience in the formulation and optimization of cell culture media. Dr. Conrad's email address is biomediaexpert@earthlink.net
|