Your Favorite Gene - Powered By Ingenuity
Sigma-Aldrich
Your Favorite Gene - Powered By Ingenuity
Find SPHK1 Products
Interaction Network for SPHK1
SPHK1 Details
Related Pathways
Sphingolipid Metabolism
Phospholipid Degradation
PDGF Signaling
View All Pathways
S-1P Stimulated Signaling
Synonyms: 1110006G24Rik, HSK, SK1, Sphingosine kinase, SPHK, SPHK1, SPHK1A
S-1P Stimulated Signaling
Sphingosine-1-phosphate (SPP/S-1P/S1P) is a bioactive sphingolipid generated within cells by types 1 and 2 sphingosine kinases (SPHK1, SPHK2). A variety of growth and survival factors enhance the expression and activation of SPHK1/2. The cytoplasmic enzyme, SPHK1, is recruited by ligand-activated platelet derived growth factor receptors (PDGF-R) where it locally enhances the conversion of sphingosine (Sph) to S-1P. Sphingosine and its precursor, ceramide (N-acylsphingosine) (Cer) are pro-apoptotic second messengers, whereas S-1P is anti-apoptotic. The conversion of Cer and Sph into S-1P shifts the ratio of pro-apoptotic to anti-apoptotic sphingolipids towards survival. The balance of the relative amounts of these phospholipids is referred to as the “sphingolipid rheostat”
Sphingosine-1-phosphate is linked to a variety of biological processes that affect cell growth, survival and motility. S-1P signaling involves two independent, but coordinated, mechanisms. S-1P regulates cell functions such as vascular maturation, angiogenesis, growth, survival, cytoskeletal organization and motility as an “inside out” agonist to a family of G-protein coupled receptors (GPCR). In addition, S-1P regulates cell growth, survival and calcium homeostasis as a GPCR receptor-independent “second messenger.
Secreted S-1P is known to bind to five differentially expressed members of the EDG-family of GPCR receptors: EDG-1/S1P1, EDG-5/S1P2, EDG-3/S1P3, EDG-6/S1P4, and EDG-8/S1P5/NRG-1. These receptors mediate their actions primarily through G-protein alpha i/o and alpha 12/13 subunits. These receptors are linked to angiogenesis, vascular maturation, neuronal survival and immunity especially as they affect aspects of cell migration, and chemotaxis. Second messenger (internal) S-1P stimulates cell growth, suppresses apoptosis and caspase activation by a variety of mechanisms. Sph-1-P blocks apoptosis, at least in part, by inhibiting the formation of ceramide from sphingomyelin by acidic sphingomyleinase.
References:
  1. Malek, R.L. et. al. (2001) Nrg-1 belongs to the endothelial differentiation gene family of G protein-coupled sphingosine-1-phosphate receptors. J. Biol. Chem. 276, 5692-5699.
  2. Maceyka, M. (2002) Sphingosine kinase, sphingosine-1-phosphate, and apoptosis. Biochim. Biophys. Acta. 1585, 193-201.
  3. Olivera, A. et. al. (2003) Sphingosine kinase type 1 induces G12/13-mediated stress fiber formation, yet promotes growth and survival independent of G protein-coupled receptors. J. Biol. Chem. 278, 46452-46460.
  4. Spiegel, S. and Milstien, S. (2003) Exogenous and intracellularly generated sphingosine 1-phosphate can regulate cellular processes by divergent pathways. Biochem. Soc. Trans. 31, 1216-1219.
Back to top
Content for this page is provided by Dennis R. Conrad, Ph.D., a Life Science industry consultant with over 25 years of experience in the formulation and optimization of cell culture media. Dr. Conrad's email address is biomediaexpert@earthlink.net