Your Favorite Gene - Powered By Ingenuity
Sigma-Aldrich
Your Favorite Gene - Powered By Ingenuity
Find SOCS1 Products
Interaction Network for SOCS1
SOCS1 Details
Related Pathways
JAK/Stat Signaling
IL-2 Signaling
IL-6 Signaling
View All Pathways
SOCS Pathway
Synonyms: CIS1, CISH1, Cish7, JAB, JBP, SOCS1, Sosc1, SSI-1, STAT INDUCED STAT INHIBITOR-1, TIP3
SOCS Pathway
The suppressor of cytokine signaling (SOCS) family is composed of SOCS-1/SSI-1/JAB-1/TIP3; SOCS-2; SOCS-3 to SOCS-7 and cytokine-inducible SH2-containing protein (CIS). SOCS proteins are cytokine-induced negative feedback-loop regulators of cytokine signal transduction, especially pathways involving JAK/STAT signaling. SOCS proteins are involved in the down regulation of a variety of cytokines including interferon gamma (IFNgamma), growth hormone (GH), erythropoietin (EPO), granulocyte-macrophage colony stimulating factor (GM-CSF), granulocyte-colony stimulating factor (G-CSF), IL-2, IL-3, IL-6, leptin, IL-12. SOCS-1 and SOC-3 have been linked to cytokine induced insulin resistance via ubiquitin proteosome mediated degradation of IRS-1 and IRS-2. SOCS have been linked to the degradation of the guanine nucleotide exchange factor (GEF) Vav, and focal adhesion kinase (FAK). The specificity of SOCS action is dependent upon the SOCS isoform.
SOCS proteins, CIS, SOCS-1, SOCS-2 and SOCS3, inhibit cytokine-activated Janus kinase/signal transducers and activators of transcription (JAK/ STAT) signaling pathways. SOCS proteins contain a central src homology 2 domain (SH2), a C-terminal SOCS box and unique N-termini. SOCS proteins employ three primary mechanisms to inhibit cytokine signaling. SOCS SH2 domains interact with phosphorylated JAK proteins and other non-receptor protein kinases and inactivate them or with cytokine receptors wherein they block STAT protein binding and activation. The SOCS box in the C-terminal targets molecules associated with SOCS proteins to proteosome mediated degradation via a mechanism that involves binding of elongins B and C and the E3 ubiquitin ligase complex.
References:
  1. Alexander, W.S. and Hilton, D.J. (2004) The role of suppressors of cytokine signaling (SOCS) proteins in regulation of the immune response. Annu. Rev. Immunol. 22, 503-529.
  2. Ilangumaran, S. et. al. (2004) Regulation of the immune system by SOCS family adaptor proteins. Semin. Immunol. 16, 351-365.
  3. Sitko, J.C. et. al. (2004) Tyrosine-phosphorylated SOCS3 interacts with the Nck and Crk-L adapter proteins and regulates Nck activation. J. Biol. Chem. 279, 37662-37669.
Back to top
Content for this page is provided by Dennis R. Conrad, Ph.D., a Life Science industry consultant with over 25 years of experience in the formulation and optimization of cell culture media. Dr. Conrad's email address is biomediaexpert@earthlink.net