Your Favorite Gene - Powered By Ingenuity
Sigma-Aldrich
Your Favorite Gene - Powered By Ingenuity
Find VEGFA Products
Interaction Network for VEGFA
VEGFA Details
Related Pathways
VEGF Signaling
Ephrin Receptor Signaling
RAR Activation
View All Pathways
VEGF and S-1P Signaling
Synonyms: Gd-vegf, MGC70609, VEGF, Vegf-3, VEGF-E, VEGF1, VEGF120, Vegf164, Vegf165, Vegf188, VEGFA, Vegfa 188, VPF, VPF/VEGF
VEGF and S-1P Signaling
Angiogenesis requires the detachment and movement of endothelial cells through basement membranes to new locations (chemotaxis), followed by proliferation and tube and vascular barrier formation. Vascular endothelial growth factor (VEGF) and sphingosine-1-phosphate (SPP/S-1P/S1P) are important mediators of angiogenesis. The predominant endothelial cell receptor for VEGF and is VEGFR-2 (Flk1/KDR) and for S-1P the EDG receptors EDG-1/S1P1 and EDG-3/S1P3.
VEGF activation of VEGFR-2 leads to the stimulation of the phosphoinositol-3 kinase (PI3-K) pathway and subsequent activation of the downstream kinase, Akt/PKB. Akt/PKB mediates cell survival signaling by phosphorylating several factors involved in apoptosis machinery including: caspase-9, BCL2-antagonist of cell death (BAD), forkhead in rhabdomyosarcoma-like 1 (FKHRL1); IκB kinase, IKKalpha (an NFkappaBp65 activator). Akt/PKB stimulates protein synthesis by turning off constitutively active glycogen synthase kinase-3 (GSK3) and turning on mTOR (TORC1). Akt/PKB promotes cell-cycle progression by phosphorylating cyclin-dependent kinase (CDK) inhibitors p27(Kip1) and p21(CIP1). Akt/PKB attenuates the action of the second messenger cAMP by activating the cAMP phosphodiesterases-3B (PDE-3B). In endothelial cells, Akt also phosphorylates and activates endothelial nitric oxide synthase (eNOS) which increases nitric oxide (NO) levels. The activation of VEGFR-2 also activates a CrkII-dependent pathway that induces membrane ruffling and cell movement.
Sphingosine-1-phosphate (S-1P) produced by endothelial cells or blood platelets is a potent endothelial cell chemo-attractant and mediator of cell survival and growth. The predominant endothelial receptor EDG-1 is linked to a G(i)-trimeric GTPase. The activation of EDG-1 by S-1P is linked to the promotion of angiogenesis via the elevation of calcium/calmodulin activity and transactivation of the VEGFR-2 receptor. Transactivation of VEGFR-2 via EDG-1 activation involves its phosphorylation by an EDG-1 activated Src kinase. The concerted effects of calmodulin activation and AKT phosphorylation synergistically elevate eNOS activity and promote cell survival, protein synthesis and motility.
References:
  1. Endo, A. et. al. (2002) Sphingosine 1-phosphate induces membrane ruffling and increases motility of human umbilical vein endothelial cells via vascular endothelial growth factor receptor and CrkII. J. Biol. Chem. 277, 23747-23754.
  2. Lee, O.H. et. al. (1999) Sphingosine 1-phosphate induces angiogenesis: its angiogenic action and signaling mechanism in human umbilical vein endothelial cells. Biochem. Biophys. Res. Commun. 264, 743-750.
  3. Liu, F. et. al. (2001) Differential regulation of sphingosine-1-phosphate- and VEGF-induced endothelial cell chemotaxis. Involvement of G(ialpha2)-linked Rho kinase activity. Am. J. Respir. Cell Mol. Bio. 24, 711-719.
  4. Tanimoto, T. et. al. (2002) Transactivation of vascular endothelial growth factor (VEGF) receptor Flk-1/KDR is involved in sphingosine 1-phosphate-stimulated phosphorylation of Akt and endothelial nitric-oxide synthase (eNOS). J. Biol. Chem. 277, 42997-43001.
Back to top
Content for this page is provided by Dennis R. Conrad, Ph.D., a Life Science industry consultant with over 25 years of experience in the formulation and optimization of cell culture media. Dr. Conrad's email address is biomediaexpert@earthlink.net