Sigma-Aldrich
Life Science > Zinc Finger Nuclease (ZFN)
Functional Genomics & RNAi
Zinc Finger Nuclease (ZFN)
CompoZr® Zinc Finger Nuclease Technology
Zinc finger nucleases (ZFNs) are a class of engineered DNA-binding proteins that facilitate targeted editing of the genome by creating double-strand breaks in DNA at user-specified locations.
Double-strand breaks are important for site-specific mutagenesis in that they stimulate the cell’s natural DNA-repair processes, namely homologous recombination and Non-Homologous End Joining (NHEJ).
Using well-established and robust protocols, these cellular processes can be harnessed to generate precisely targeted genomic edits resulting in cell lines, including somatic cell lines, with targeted gene deletions (Knockouts), integrations, or modifications.
Learning Center:
Tool Box:
Applications Center:
Products:
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Your Favorite Gene - Powered by Ingenuity
Your Favorite Gene Search
Refer-a-Friend iPad Banner
ZFN Symposia Banner
Dive into RNAi Screening - sigma.com/shrnascreen