|
||||||||||||||||||
|
For more than a decade, hydrogen as an alternative to traditional (fossil) energy sources has been in the focus of research and development efforts in all technologically advanced countries of the world. It is strongly believed that hydrogen can help to address the growing demand for energy and slow down global climate change.1
Another material of great interest for hydrogen storage is ammonia borane2 (Prod.No 682098), which releases ~ 13% of hydrogen in two steps at temperatures exceeding 100° C (Figure 1).
Further substances evaluated as high-capacity sources of hydrogen include lithium and sodium borohydrides (Prod.No. 686026, 685917, 686018), which are capable of an irreversible hydrogen release in aqueous solutions in the presence of metal catalysts.7
To see our complete listing in the area of Hydrogen Storage please visit sigma-aldrich.com/hydrogen. References
|
|||||||||||||||||
|
|
||||||||||||||||||