Aldrich Materials Science
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Magnetic Material
Superconductivity
Introduction
Magnetic Properties of Materials
Superconductors
A History of Superconductivity
Molecule-Based Magnets
Spintronics
References
A History of Superconductivity
  • 1908 Dutch physicist Heike Kamerling Onnes produces liquid helium (bp 4.22 K).
  • 1911 Onnes discovers that when mercury is cooled below 4 K is has zero electrical resistance. Mercury becomes the first reported superconducting material.12 Two years later he is awarded a Nobel Prize in Physics for his investigations on the effect of very low temperatures on matter.8
  • 1933 W. Meissner and R. Oschsenfeld observe that when a superconducting material is cooled below its critical temperature (TC) magnetic fields are excluded from the material. This is distinct from a perfectly diamagnetic (zero resistant) material in which there would be no deviation in magnetic field. This becomes known as the Meissner Effect (see Figure 4).13
The Meisner effect
Figure 4. The Meisner effect.
  • 1941 Superconductivity is found in niobium nitride below 16 K.
  • 1962 First commercial superconducting wire (niobium-titanium) developed by Westinghouse.
  • 1972 John Bardeen, Leon Cooper, and John Schrieffer win Nobel Prize in Physics for their theory of superconductivity (BCS theory).8
  • 1975 The first polymeric superconductor (SN)x reported.
  • 1980 The first superconducting (at high pressure) molecular compound, (TMTSF)2PF6 (where TMTSF is tetramethyltetraselenafulvalene, Aldrich product 27,440-2) reported.14
TMTSF
  • 1981 First ambient pressure superconducting molecular compound, (TMTSF)2ClO4, reported.4
  • 1986 Alex Müller and Georg Bednorz synthesize a lanthanum barium cuprate (La2-xBaxCuO4) with a TC of 35 K.15
  • 1987 Bednorz and Müller's group report an yttrium barium cuprate (YBa2Cu3O7) with a TC of 93 K. This is the first reported high-temperature superconductor (HTS), that is, a compound which superconducts above the boiling point of nitrogen (77 K).16,17
  • Bednorz and Müller receive a Nobel Prize in Physics that same year for their discovery.8
  • 1988 A superconducting ceramic (Tl2Ba2Ca2Cu3O10) with a TC as high as 125 K is reported.18
  • 1991 Fullerides of the formula AX@C60 (A = K, Rb, Cs) reported to have superconducting character.5
  • 1993 A mercury barium cuprate (HgBa2Ca2Cu3O10) with a TC of 134 K19 (164 K at 30 Gpa20) is reported. This is the highest TC superconductor to date.
  • 2001 Magnesium boride, MgB2 (Aldrich product 55,391-3) is reported to have superconducting properties (TC = 39 K).6
< Previous  Next >