Sigma-Aldrich
Materials Science > Alternative Energy Materials > Gas Sorption Analyzer
Alternative Energy Materials
Gas Sorption Analyzer
Hy-Energy's PCTPro-2000 Gas Sorption Analyzer
The Most versatile and robust Sieverts Apparatus available
  • Designed to perform wide range precision kinetics, PCT-isotherm, and cycling measurments on samples ranging in size from milligrams to kilograms.
  • Constructed to be compatible with essentially all research-grade gases.
  • Five Calibration volumes, eleven high-pressure control valves, multiple high-precision pressure transducers with auto-range switching.
  • Extremely stable temperature regulated manifold, and a PID-controlled internal pressure system able to regulate pressures from 1 mbar to 200 bar with high accuracy.
Materials for Hydrogen Storage Applications
User-friendly LabView-based software package is designed to remove user error and guesswork.
HyData software control and analysis package:
  • Includes thirteen automated processes and measurement routines; allows real-time viewing of results and parameter adjustment during measurements.
  • Data outputed in a standard, easy-to-read ASCII file format.
  • Advanced data analysis supported through HyAnalysis, a powerful analysis software package.
For further information contact us at matsci@sial.com
Download the PCTPro-2000 brochure (2.6 Mb PDF), or for more information, please register below.