|
Alternative Energy Materials
Polymeric Electrolytes
|
Solid polymeric electrolytes (SPEs) are used as polymer electrolyte membranes in PEM fuel cells.
 Schematic of a typical proton exchange membrane (PEM) fuel cell.
|
|
|
Cross-linkable sulfonated SPE formulations
|
|
 |
659401 Poly(styrene-ran-ethylene), sulfonated, cross-linkable, 5 wt.% solution in 1-propanol
- Contains <0.5% vinyl silane cross-linking agent
- Styrene 76 wt.%
- Sulfonated styrene units 32-38%
- Conductivity = 0.1184 S/cm
- The ionic conductivity can increase up to 50% when heating from RT to 67 °C.
- Treat with 0.5 M Sulfuric acid for 20 min to strengthen after cross-linking.
- Material becomes cross-linked w/800-1000 mJ/cm2 UV exposure after drying at 110°C for 10-15 min; or at 140-150°C for 1 min.
|
|
 |
659444 Polystyrene-block-poly(ethylene-ran-butylene)block-polystyrene, sulfonated, cross-linkable, 5 wt.% solution in 1-propanol
- Contains <0.5% vinyl silane cross-linking agent
- Styrene 29 wt.%
- Sulfonated styrene units 55-65%
- Conductivity = 0.1078 S/cm
- The ionic conductivity can increase up to 50% when heating from RT to 67 °C.
- Treat with 0.5 M Sulfuric acid for 20 min to strengthen after cross-linking.
- Material becomes cross-linked w/800-1000 mJ/cm2 UV exposure after drying at 110°C for 10-15 min; or at 140-150°C for 1 min.
|
Other sulfonated SPE formulations
|
|
 |
659592 Poly(styrene-ran-ethylene), sulfonated, 5 wt.% solution in 1-propanol
- Styrene 76 wt.%
- Sulfonated styrene units 32-38%
- Conductivity = 0.1494 S/cm
- The ionic conductivity can increase up to 50% when heating from RT to 67 °C.
|
|
 |
448885 Polystyrene-block-poly(ethylene/butylene)-block-polystyrene, sulfonated solution, 5 wt.% solution in propanol and dichlorethane
- Styrene 29 wt.%
- Sulfonated styrene units 55-65%
- Conductivity ~ 0.07 S/cm
|
A spin coat system is recommended for casting these polymers into films. Click here to view the tech bulletin on the spin coat system. (78.2 KB PDF)
|
|
Spin-Coating Systems
|
| Product Name |
Prod. No. |
| Chemat precision spin-coater, 2-stage, 220V |
Z551589 |
| Chemat precision spin-coater, 2-stage, 115V |
Z551562 |
|
|
|
Manual Film-Casting Instructions
|
|
To cast a film without using a spin coat system, the following procedure is recommended:
- Concentrate solution to 600 centapoise viscosity (typically 6-9 wt. % polymer).
- Preheat the casting surface to 35°C. A clean, flat teflon-coated glass surface is recommended.
- Secure a silicone release liner to the casting surface.
- Place a small drop of the polymer solution on the release liner.
- Draw the polymer solution down the release liner.
- Dry the film for 30 min. For controlled evaporation it is recommended to cover surface with aluminum foil that has been perforated with small pinholes.