Cobalt
 Cobalt
Metal
Salts
Oxides
Nanomaterials
Solution Deposition Precursors
Vapor Deposition Precursors
Atomic Number: 27
Electronic Configuration: [Ar]3d74s2
Atomic Weight: 58.9332
Did you know? Cobalt is alloyed with iron, nickel, and other metals to form Alnico, an alloy with exceptional magnetic strength.