Platinum
Atomic Number: 78
Electronic Configuration: [Xe]4f145d96s1
Atomic Weight: 195.078
Did you know? In photography, Platinum is sometimes used for archival printmaking. Platinum prints display a greater range of tones than other Black and White printing methods.
Return to Periodic Table