Tellurium
 Tellurium
Metals
Vapor Deposition Precursors
Oxides
Atomic Number: 52
Electronic Configuration: [Kr]4d105s2p4
Atomic Weight: 127.6
Did you know? Tellurium is used in the media layer of several types of rewritable optical discs, including ReWritable Compact Discs (CD-RW), ReWritable Digital Video Discs (DVD-RW) and Blu-ray Discs.