Atomic Number: 74
Electronic Configuration: [Xe]4f145d46s2
Atomic Weight: 183.84
Did you know? Because of its ability to produce hardness at high temperatures and its high melting point, elemental tungsten is used in many high-temperature applications, such as nozzles on rocket engines.