Aldrich Materials Science
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Micro & Nanoelectronics
Ferrocenes
6-(Ferrocenyl)hexanethiol for Self-Assembly on Gold Surfaces
The electrochemical behavior of surface-modified electrodes is of interest for both fundamental and applied materials science. Among the different strategies which can be used to prepare such modified electrodes, the gold-alkane thiol self-assembly system has proven to be particularly attractive. It allows the creation of highly organized molecular thin films on gold electrodes. 1,2
Combined processes of molecular-scale recognition and surface self-assembly offer a powerful route to novel sensing systems based on analyzing interactions at nano-scale. Redox active ferrocene moiety in the selective electrochemical sensing has been found to be particularly interesting and methods of surface self-assembly for the fabrication of sensory devices are being developed. 3
Sigma-Aldrich Materials Science is pleased to offer.
ferrocenes_071
6-(Ferrocenyl)hexanethiol for self-assembly on gold surface (682527)
The redox potentials of the title material are provided below to help you in designing novel electrochemical systems.
Redox
For pricing and Certificate of Analysis please click on the product number below.
Product #  Product Name Add to Cart
682527 6-(Ferrocenyl)hexanethiol
We hope that this novel material will help you explore and create new systems by applying the rapidly developing area of self-assembly.
References
  1. Material Matters, 2006, v.1, No.2.
  2. S.E. Creager, G. K. Rowe, J. ElectroanalyticaI Chemistry, 1994, 370, 203.
  3. P. D. Beer, J. J. Davis, D. A. Drillsma-Milgrom, F. Szemes, Chem. Commun. 2002, 1716.