|
Micro & Nanoelectronics
NanoThinks
 |
| NEW NanoThinksTM solutions will help you get results faster by reducing prep time and mixing errors. We feature our most popular thiol inks in high-purity ethanol, just select the chain length and the functionality of the thiol and prepare your film. NanoThinksTM are ideal for use in molecular assembly or dip-pen nanolithography applications. |
 All NanoThinksTM are 5 mM solutions. |
|
NanoThinksTM Product Name and Catalog Number Chart
|
Functional Group (R) |
Number of Carbons in Chain (N)
|
| C 8 |
C 11 |
C 16 |
C 18 |
| -H |
NanoThinks 8
662208
|
- |
- |
NanoThinks 18
662194
|
| -OH |
- |
NanoThinks ALCO11
662224
|
- |
- |
| -SH |
NanoThinks THIO8
662615
|
- |
- |
- |
| -COOH |
- |
NanoThinks ACID11
662925
|
NanoThinks ACID16
662216
|
- |
|
| Additional New Products for Molecular Self-Assembly |
| Product # |
Product Name |
Structure |
Add to Cart |
| 659452 |
1-Adamantanethiol, 95% |
 |
|
| 660493 |
1H,1H,2H,2H-Perfluorodecanethiol, 97% |
 |
|
Nanothinks is a trademark of the Sigma-Aldrich Corporation Sigma-Aldrich is a registered trademark of Sigma-Aldrich Biotechnology, L.P. |