Aldrich Materials Science
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Micro and Nanoelectronics
Silane
Sol-gel processing is beneficial in the formation of ceramic and glass films for a number of reasons.1-4 It is a simple process that does not require exotic materials, catalysts or expensive deposition equipment. Sol-gel reactions do not employ extreme reaction conditions and the properties of the materials prepared using sol-gel approaches are easy to modify by utilizing an organically modified precursors.2,3
Deposition Grade Silanes comprise alkoxysilanes and their predecessors chlorosilanes typically used in a Sol-Gel process. Transformations of alkoxysilanes during the processing proceed through a series of subsiquent hydrolysis, condensation and thermal treatment steps briefly outlined in Figure 1.
Fig_1
Figure 1
Applications of Deposition Grade Silanes include but are not limited to
  • film coatings on glass and other appropriate surfaces1,4
  • reinforcement of polymeric structures1, such as polyesters,16 Nafion®,17 Surlyn®
  • hermetic barriers for electronics and electrochemical devices5
  • and others applications where Si-O-based chain assemblies are desirable.3,4
Extensive analytical data available for Deposition Grade Silanes allow for a better understanding of chemical composition and properties of the materials created using these precursors as compared to those prepared form conventional silanes available commercially.
Purity of our Deposition Grade Silanes exceed 98%. They are fully characterized by chemical analysis and nuclear magnetic resonance (NMR). The impurities present in these materials are detected using a gas-chromatography (GC) and/or a GC-MS technique and reported on the Certificate of Analysis (CA). In addition, trace metals impurities are also reported on the CA.
Product Information
Product # Product Name/Description Structure Add to Cart
679267 Allytrimethoxysilane 6792671
679224 Butyltrichlorosilane 679224
679356
[3-(Diethylamino)propyl]trimethoxysilane
6793561
679216 Ethyltrichlorosilane 679216
679364 Isobutyl(trimethoxy)silane 6793641
679208 Methyltrichlorosilane 679208
679321
N-Propyltriethoxysilane 6793211
679194 Pentyltrichlorosilane 679194
679259 Tetramethyl orthosilicate 679259
679240 Tetrapropyl orthosilicate 679240
679305 Triethoxy(octyl)silane 6793051
679291 Triethoxyphenylsilane 6792911
679275 Triethoxyvinylsilane 6792751
679232 Trimethoxymethylsilane 6792321
679313 Trimethoxyphenylsilane 6793131
References:
  1. S.K.Young, Material Matters, 2006, Vol 1., No.3, 8.
  2. F. Caruso, Colloids and Colloid Assemblies: Synthesis, Modification, Organization, and Utilization of Colloid Particles, 2004, John Wiley & Sons, New York.
  3. Sol-Gel Technology for Thin Films, Fibers, Preforms, Electronics, and Specialty Shapes, L.C. Klein, Ed., 1988, Noyes Publ., Park Ridge, NJ.
  4. A.C.Pierre, Introduction to Sol-Gel Processing, 2002 Kulwer, Academic Publishers, Boston, Dodrecht, London.
  5. A. B. Wojcik, L. C. Klein, J. Sol-Gel Science and Technology, 1995,4, 57.