Sigma-Aldrich
Materials Science > Nanomaterials > Dendrimers
Nanomaterials
Dendrimers
 New Products Product Catalog Technical Notes
Sigma-Aldrich offers a wide selection of dendritic macromolecules, including dendrimers, dendrons, and hyperbranched polymers. Available dendrimer types include: poly(amidoamine), also known as PAMAM, or STARBURST® dendrimers; phosphorous dendrimers; polylysine dendrimers; and polypropylenimine (PPI) dendrimers. Dendrimer kits group smaller quantities of similar PAMAM dendrimers into cost-effective tool-sets that can help you choose the best type of dendrimer for a particular application by systematically changing the normal dendrimer features (e.g. generation or surface group). Dendrons are wedge-shaped sections of dendrimers with reactive focal point functions, useful as macromolecular building blocks. Hyperbranched polymers are inexpensive dendritic macromolecules that can be used for many applications (conjugate scaffolds, macromolecular chemistry, polymer additives) that are tolerant of imperfect branching and polydispersity.
New Products:
Product Number Description Attributes
683493 PEG-Dendrimer Kit,
Generations 3-6
Kit of PAMAM dendrimers with PEG-12 (-COO(CH2CH2O)12CH3) surface groups
683507 Hydrophilic-Dendrimer Kit Kit of gerenartion 4 PAMAM dendrimers with –OH, -NH2, -PEG, and COOH surface groups
686603 Hyperbranched bis-MPA Polymer, gen. 2 16 (average) surface –OH groups
686581 Hyperbranched bis-MPA Polymer, gen. 3 32 (average) surface –OH groups
686573 Hyperbranched bis-MPA Polymer, gen. 4 64 (average) surface –OH groups
Dendrimer Product Catalog: Related Products:
PAMAM Dendrimers Dendrons
Dendrimer Kits Hyperbranched Polymers
Phosphorous Dendrimers Biocompatible/Biodegradable Materials
Polylysine Dendrimers
Polypropylenimine Dendrimers
Technical Notes:
Summary of generation-dependant properties of PAMAM dendrimers. Solubility table of PAMAM dendrimers.
Read recent article about using PAMAM Dendrimers to encapsulate and release drugs in Material Matters 2.1