Sigma-Aldrich
Spacer
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Materials Science > Nanomaterials > Nanotubes and Fullerenes
Nanomaterials
Nanotubes and Fullerenes
Sigma-Aldrich offers a wide selection of carbon nanomaterials, including carbon nanotubes (CNTs) and fullerenes (buckyballs). Available CNTs are classified into multi-wall nanotubes (MWCNTs), double-wall nanotubes (DWCNTs), and single-wall nanotubes (SWCNTs). In addition, SWCNTs are available with chemical functions covalently grafted to the nanotube outer surface. Fullerenes are available in several degrees of purity and with a wide variety of chemical functions. The functionalized fullerenes include a library of all PCBM derivatives shown to be important for research in Display and Optoelectronics.
Carbon Nanotubes
 new_icon_opcCarbon Nanotubes 
SouthWest NanoTechnologies (SWeNT) Logo 704148 - SWCNT, (6,5) chirality, >80% carbon basis, produced by CVD process
704121 - SWCNT, (7,6) chirality, >80% carbon basis, produced by CVD process
704113 - SWCNT, >75% carbon basis, produced by CVD process
698695 - SWCNT, 40-60 wt.% as SWCNT, produced by Arc Method
698849
- MWCNT, >99% MWCNT
698830 - Graphite platelet nanofibers, >99% (trace metal basis) 
Featured Carbon Nanotubes, Unfunctionalized 
Product Name Outer Diameter
(nm)
Inner Diameter
(nm)
Length (µm) Purity
(% CNT)
Product #
Single-wall, as produced 1.2-1.5 NA 2-5 50-70 519308
Single-wall 1.1 NA .5-100 > 50 636797
Double-wall 5 1.3-2.0 .50 50-80 637351
Multi-wall 7-15 3-6 .5-200 > 95 694185
Multi-wall, Arkema CVD 10-15 2-6 .1-10 > 90 677248
Multi-wall 110-170 5-9 > 90 659258
Multi-wall, powdered cylinder cores 2-15 1-10 10-40 406074
Multi-wall, as produced 6-20 1-5 > 7.5 412988
Graphite, nanofibers 80-200 0.5-10 .5-20 > 95 636398
Featured Carbon Nanotubes, Functionalized
Product Name Diameter (nm) Length (µm) Loading Solubility (mg/ml) Product #
Single-wall, carboxylic acid functionalized
4-5 .5-1.5 3-6 atom% H2O 0.1 DMF 1.0   652490
Single-wall, amide functionalized 4-6 .7-1.0 3-6 atom%   DMF > 0.5 alcohols > 0.5 685380
Single-wall, PABS (polyaminobenzene sulfonic acid) functionalized 1.1 .5-1 65 wt.% H2O 5.0 DMF 0.1 EtOH 0.05 639230
Single-wall, PEG (polyethylene glycol functionalized) 4-5 .5-.6 30 wt.% H2O 5.0     652474
Single-wall, ODA (octadecylamine functionalized) 2-8 .5-1.0 30-40 wt.% THF > 1.0 CS2 > 1.0 toluene > 1.0 652482
Featured Carbon Nanotubes, Arrays
Product Name Substrate CNT Diam.(nm) x L (µm) Purity
(% CNT)
Product #
Multi-wall, Vertically aligned Silicon wafer 100 ± 10% x 30 ± 10% >99.9 687804
Multi-wall,Vertically aligned  Copper 100 ± 10% x 30 ± 10% >99.9 687812
Fullerenes
Featured Fullerenes
Product Name Purity (%) Product #
Fullerene C60 98.0 483036
99.5 379646
Fullerene C60, sublimed 99.9 572500
Fullerene C70 95 379654
Fullerene C70 99 482994
Fullerene C84 98 482986
Phenyl-C61-Butyric-Acid-Methyl Ester, [60]PCBM 99 684430
99.5 684449
99.9 684457
Phenyl-C71-Butyric-Acid-Methyl Ester, [70]PCBM 99 684465
Phenyl-C85-Butyric-Acid-Methyl Ester, [84]PCBM 99 684473
Phenyl-C61-Butyric-Acid-Butyl Ester, PCBB, [60]PCB-C4 > 97 685321
Phenyl-C61-Butyric-Acid-Octyl Ester, PCBO, [60]PCB-C8 99 684481
Thienyl-C61-Butyric-Acid-Methyl Ester, [60]ThCBM 99 688215
Pentadeuterophenyl-C61-Butyric-Acid-Methyl Ester, d5-PCBM 99 684503
          PCBM Library 
          Fullerenes (complete listing)
Technical Notes
Read recent article about unique properties and novel applications of nanoparticles in Material Matters Vol. 2 No. 1 (2.2 Mb PDF).