|
Organic and Printed Electronics
High Efficiency Sensors
|
Ru(d8-bipy3)PF6 is known to be an excellent candidate for emissive materials in organic light-emitting diodes (OLEDs) and organic light-emitting electrochemical cells (OLEC) for sensor applications1. Quantum efficiencies of these materials are in the range of 1.2-1.5%. Scientists at Sigma-Aldrich® are aware of the improvement in emission lifetimes of triplet emitting compounds by deuterating the organic ligand. The deuterated material shows a 20-25% increase in emission lifetime compared to the undeuterated material as shown below2. We think that you will find our newest material Ru(d8-bipy3)PF6 will improve your sensor efficiency.
652407
|
Emission Lifetimes3
|
| Matrix |
Temp. |
Ru(bpy3) |
Ru(d8-bpy3) |
| Water |
298K |
610ns |
760ns |
| Crystal |
5K |
64ms |
80ms |
Please Contact Customer Service at 1-800-325-3010 to place orders.
References:
- Rudman, H.; Rubner, M.F. J. Appl. Phys. 2001, 90, 4338.
- Browne, W.R.; Vos, J.G. Coord. Chem. Rev., 2001, 219, 761.
- Krausz, E.; Moran, G.; Riesen, H. Chem. Phys. Lett. 1990, 165, 401.