Aldrich Materials Science
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Materials Science > Organic and Printed Electronics > Oligothiophene Synthesis
Synthetic Strategy for Large Scale Production of Oligothiophenes
Kanth Josyula, Mitesh Patel, Kaushik Patel, Scott Batcheller
Aldrich Materials Science, 6000 N. Teutonia Ave. Milwaukee, WI 53209
Introduction
Oligothiophenes and their functional derivatives are used in a number of high-technology applications including OLEDs, OFETs and OPVs, where the ability to functionalize the oligothiophene backbone allows for the fine-tuning of the electronic properties.1
These functional oligothiophenes can be synthesized from synthetic intermediates via a number of different reactions. Suzuki coupling2 has proven to be particularly advantageous in the large scale production of oligothiophenes. Among the advantages of Suzuki coupling are predictable scaling reactions, less-toxic reagents, simple removal of metallic impurities and the ability to further purify products by sublimation. In addition, coupling building blocks are commercially available, allowing for the simple synthesis of any required oligothiophene.
General Synthetic Strategy for Large Scale Production of Oligothiophenes
Figure 1 illustrates different synthetic pathways to oligothiophene building blocks, and ultimately to functional oligothiophenes via Suzuki Coupling.
Figure 1.
Oligothiophene Constructor
Functional oligiothiophenes can be synthesized by reacting boronic acid and bromide intermediates in the presence of a palladium catalyst and a base as part of a Suzuki Coupling reaction. A general constructor is presented in Figure 2.
Figure 2.
Organic Electronic Materials
Figure 3 illustrates a representation of functional oligothiophenes synthesized via Suzuki Coupling.
Figure 3.
Materials
References
  1. Mishra, A.; Ma, C.-Q.; Bauerle, P. Chem. Rev. 2009, 109, 1141.
  2. Suzuki, A. Pure Appl. Chem. 1991, 63, 419.