Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Photonic/Optical Materials
Plastic Optical Fibers Photonic/Optical Materials Tutorial
  Introduction
Crystal Grade Materials
Organic NLO Materials
References
Plastic Optical Fibers
Polymeric materials in general, and fluoropolymers in particular, are being extensively researched for their use in plastic optical fibers (POFs), waveguides, optical filters, fiber gratings, as well as other optical devices.17-19 They offer several advantages over their inorganic counterparts including relative ease of processing, light weight, insensitivity to vibrational stress, low cost, and a wide versatility with respect to their design and synthesis to obtain properties that suit specific device parameters. Figure 2 is a photograph of an archetypical optical fiber. In the case of POFs, fluoropolymers, which have the lowest refractive index, serve as cladding material to maximize the numerical aperture (NA) of the fiber. Numerical aperture is a measure of the optical fibers ability to trap light more efficiently and is quantified as NA = [(n core2 – n cladding2)]1/2. Other issues that must be considered in the fabrication of POFs include transparency at the wavelengths of transmission, thermal expansion matching of cladding and core materials, and thermal stability in the temperature range of operation.20
Figure 2. A photograph of an archetypical optical fiber.
< Previous  Next >