|
Organic Electronics
Polymer Semiconductors
|
|
|
Organic polymer semiconductors are key materials for research in organic electronics. Efforts to develop field-effect transistors (FETs), plastic solar cells, organic RFIDs, and electrochemical sensors all depend on availability of reliable organic semiconductors with choices of different molecular architectures and consistent quality. Sigma-Aldrich is pleased to offer these new conjugated polymer semiconductors:
1. Electronic grade Poly(3-alkylthiophenes) – “benchmark” high-performance organic semiconductors.
Poly(3-alkylthiophenes) are used as p-channel conductors in organic FETs,1 and p-type materials in heterojunction photovoltaic (PV) devices2 with highest performance achieved to-date. Semiconductor performance of these polymers can be degraded by residual catalyst impurities,3 defects in polymer chain regiospecificity,4,5 and low molecular weight of the polymers. Our new electronic-grade poly(3-alkylthiophenes) are highest quality materials available in the research market, with consistently high purity, regioregularity, and molecular weight. The choice of the hexyl-, octyl-, and dodecyl- side-chains will allow you to explore effects of polymer architecture6 without worrying about differences in quality of the materials.
| Prod. No. |
Product Name |
Structure |
Properties* |
| 669067 |
Poly(3-hexylthiophene), electronic grade (P3HT) |
 |
· Avg. Mw ~ 17,500 · > 99 % head to tail · < 50 ppm metals |
| 682799 |
Poly(3-octylthiophene), electronic grade (P3OT) |
 |
· Avg. Mw ~ 25,000 · > 98 % head to tail · < 50 ppm metals |
| 682780 |
Poly(3-dodecylthiophene), electronic grade (P3DDT) |
 |
· Avg. Mw ~ 27,000 · > 97% head to tail · < 60 ppm metals |
| *Properties: Molecular Weight, Regioregularity, Metal Content |
2. BBL – n-type conjugated polymer.
Availability of n-type (electron-mobile) organic semiconductors is one of the main challenges to fabricating n-channel FETs and heterojunctions PVs. We are pleased to offer the ladder polymer poly(benzobisimidazobenzophenanthroline (BBL), one of the few conjugated polymers showing n-channel behavior. This material, processed from solutions in methanesulfonic acid, shows high electron mobility in polymer FETs.7 Organic solar cells with promising performance were fabricated from BBL with MEH-PPV 8 and poly(3-alkylthiophene) 9 p-type polymers.
| Prod.No. |
Product Name |
Structure |
Properties* |
| 667846 |
Poly(benzimidazobenzophenanthroline) (BBL) |
 |
· -5.9 eV 9 · - 4.0 eV 9 · μe = 0.1 cm2/(V s) 7 |
| 669067 |
Regioregular poly(3-hexylthiophene) (P3HT) |
 |
· - 5.1 eV · -2.8 eV · μh = 0.1 cm2/(V s) |
| *Properties: HOMO, LUMO, Carrier Mobility |
References:
- 1. Zaumseil, J.; Sirringhaus, H. Chem. Rev. 2007, 107, 1296;
- Li, G.; Shrotriya, V.; Huang, J.; Yao, Y.; Moriarty, T.; Emery, K.; Yang, Y. Nature Materials 2005, 4, 864.
- Cugola, R.; Giovanella, U.; Di Giancincenzo, P.; Bertini, F.; Catellani, M.; Luzzati, S. Thin Solid Films 2006, 511-512, 489.
- Sirringhaus, H.; Brown, P.; Friend, R.; Nielsen, M.; Bechgaard, K.; Langeveld-Voss, B.; Spiering, A.; Janssen, R.; Meijer, E.; Herwig, P.; de Leeuw, D. Nature 1999, 401, 685.
- Kim, Y.; Cook, S.; Tuladhar, S.; Choulis, S.; Nelson, J.; Durrant, J.; Bradley, D.; Giles, M.; McCulloch, I.; Ha, C.; Ree, M. Nature Materials 2006, 5, 197.
- Al-Ibrahim, M.; Roth, H.; Schroedner, M.; Konkin, A.; Zhokhavets, U.; Gobsch, G.; Scharff, P.; Sensfuss, S. Organic Electronics 2005, 6, 65.
- Babel. A.; Jenekhe, S. J. Am. Chem. Soc. 2003, 125, 13656.
- Alam, M.; Jenekhe, S. Chem. Mater. 2004, 16, 4647. 9. Manoj, A.; Narayan, K. Optical Materials 2002, 21, 417.
|