[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Applications](https://www.sigmaaldrich.com/US/en/applications)[Materials Science and Engineering](https://www.sigmaaldrich.com/US/en/applications/materials-science-and-engineering) Organic Electronics
# Organic Electronics
Organic electronics utilize organic polymers or small molecules to create electronic components for many new application areas. Organic electronic materials are lighter, more flexible, and less expensive than conventional silicon-based inorganic materials. Organic electronics are more energy-efficient and resource-friendly in production, use, and disposal.
Small molecule organic electronics are typically fabricated using vacuum-based deposition methods to transfer thin films of organic materials onto the substrate surface. Organic electronics can be made from conductive polymers using low-cost solution processing methods. Semiconductor polymers can be made soluble and turned into an ink, making it possible to print electronic circuits directly into large plastic sheets. These materials are compatible with large-area, roll-to-roll manufacturing processes, which can easily be scaled-up for rapid production at lower cost.
* * *
## Related Products
Slide 1 of 20
1 of 5
[
\
Sigma-Aldrich
\
698423
\
Pentacene](https://www.sigmaaldrich.com/US/en/product/aldrich/698423)
Quick View
[
\
Sigma-Aldrich
\
716006
\
6,13-Bis(triisopropylsilylethynyl)pentacene](https://www.sigmaaldrich.com/US/en/product/aldrich/716006)
Quick View
[
\
Sigma-Aldrich
\
739278
\
6,13-Bis((triethylsilyl)ethynyl)pentacene](https://www.sigmaaldrich.com/US/en/product/aldrich/739278)
Quick View
[
\
Sigma-Aldrich
\
911135
\
TU-3](https://www.sigmaaldrich.com/US/en/product/aldrich/911135)
Quick View
[
\
Sigma-Aldrich
\
900550
\
Poly(3-hexylthiophene-2,5-diyl)](https://www.sigmaaldrich.com/US/en/product/aldrich/900550)
Quick View
[![Tris\[2-phenylpyridinato-C2,N\]iridium(III) sublimed grade](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/structures/167/234/658d0b76-d31d-4fd5-8041-e04e207227c9/640/658d0b76-d31d-4fd5-8041-e04e207227c9.png)
\
Sigma-Aldrich
\
694924
\
Tris\[2-phenylpyridinato-C2,*N*\]iridium(III)](https://www.sigmaaldrich.com/US/en/product/aldrich/694924)
Quick View
[
\
Sigma-Aldrich
\
444561
\
Tris-(8-hydroxyquinoline)aluminum](https://www.sigmaaldrich.com/US/en/product/aldrich/444561)
Quick View
[
\
Sigma-Aldrich
\
900438
\
Super yellow light-emitting PPV copolymer](https://www.sigmaaldrich.com/US/en/product/aldrich/900438)
Quick View
[
\
Sigma-Aldrich
\
900441
\
Green light-emitting spiro-copolymer](https://www.sigmaaldrich.com/US/en/product/aldrich/900441)
Quick View
[
\
Sigma-Aldrich
\
900965
\
CzSi](https://www.sigmaaldrich.com/US/en/product/aldrich/900965)
Quick View
[
\
Sigma-Aldrich
\
901102
\
PPT](https://www.sigmaaldrich.com/US/en/product/aldrich/901102)
Quick View
[
\
Sigma-Aldrich
\
806781
\
TPBi](https://www.sigmaaldrich.com/US/en/product/aldrich/806781)
Quick View
[
\
Sigma-Aldrich
\
900940
\
DMAC-DPS](https://www.sigmaaldrich.com/US/en/product/aldrich/900940)
Quick View
[
\
Sigma-Aldrich
\
808350
\
TokeOni](https://www.sigmaaldrich.com/US/en/product/aldrich/808350)
Quick View
[
\
Sigma-Aldrich
\
902268
\
seMpai](https://www.sigmaaldrich.com/US/en/product/aldrich/902268)
Quick View
[![2-\[(1E,3E)-6-\[3-(Dimethylamino)pyridine\]-1,3-butadien-1-yl\]-4,5-dihydro-4-thiazolecarboxylic acid](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/structures/275/363/b1837e7e-7c9f-403d-bb3b-c80cf622c5f0/640/b1837e7e-7c9f-403d-bb3b-c80cf622c5f0.png)
\
Sigma-Aldrich
\
900952
\
2-\[(1*E*,3*E*)-6-\[3-(Dimethylamino)pyridine\]-1,3-butadien-1-yl\]-4,5-dihydro-4-thiazolecarboxylic acid](https://www.sigmaaldrich.com/US/en/product/aldrich/900952)
Quick View
[
\
Sigma-Aldrich
\
908509
\
Y6](https://www.sigmaaldrich.com/US/en/product/aldrich/908509)
Quick View
[
\
Sigma-Aldrich
\
913251
\
Y7](https://www.sigmaaldrich.com/US/en/product/aldrich/913251)
Quick View
[
\
Sigma-Aldrich
\
792071
\
Spiro-MeOTAD](https://www.sigmaaldrich.com/US/en/product/aldrich/792071)
Quick View
[
\
Sigma-Aldrich
\
902500
\
SHT-263 Solarpur®](https://www.sigmaaldrich.com/US/en/product/aldrich/902500)
Quick View
* * *
## Featured Categories
[](https://www.sigmaaldrich.com/US/en/products/materials-science/electronic-materials/carbon-nanomaterials)
[Carbon Nanomaterials](https://www.sigmaaldrich.com/US/en/products/materials-science/electronic-materials/carbon-nanomaterials)
A comprehensive offering of carbon nanomaterials from fullerene, to carbon nanotubes, graphene, carbon quantum dots and nanodiamonds to empower your research and development in energy storage, high performing electronics, smart composites, and novel nano therapeutics.
[Shop Products](https://www.sigmaaldrich.com/US/en/products/materials-science/electronic-materials/carbon-nanomaterials)
[](https://www.sigmaaldrich.com/US/en/products/materials-science/electronic-materials/liquid-crystals)
[Liquid Crystals](https://www.sigmaaldrich.com/US/en/products/materials-science/electronic-materials/liquid-crystals)
We provide well-defined liquid crystal materials spanning nematic, cholesteric, smectic, and discotic phases, as well as liquid crystal polymers and mesogens to facilitate the design of ordered, smart, stimuli-responsive molecular systems.
[Shop Products](https://www.sigmaaldrich.com/US/en/products/materials-science/electronic-materials/liquid-crystals)
[](https://www.sigmaaldrich.com/US/en/products/materials-science/electronic-materials/oled-and-pled-materials)
[OLED & PLED Materials](https://www.sigmaaldrich.com/US/en/products/materials-science/electronic-materials/oled-and-pled-materials)
We offer a selection of high purity, state-of-art dopant and emitter materials, host materials, and charge transport materials to facilitate your OLED and PLED research
[Shop Products](https://www.sigmaaldrich.com/US/en/products/materials-science/electronic-materials/oled-and-pled-materials)
[](https://www.sigmaaldrich.com/US/en/products/materials-science/electronic-materials/photonic-and-optical-materials)
[Photonic & Optical Materials](https://www.sigmaaldrich.com/US/en/products/materials-science/electronic-materials/photonic-and-optical-materials)
Light up your research with our comprehensive portfolio of photonic and optical materials, including laser, infrared, near-infrared, chemiluminescent, photochromic, thermochromic, phthalocyanine, and porphyrin dyes, as well as photoluminescent, non-linear optical (NLO) and waveguide materials.
[Shop Products](https://www.sigmaaldrich.com/US/en/products/materials-science/electronic-materials/photonic-and-optical-materials)
* * *
Overview
Related Articles & Protocols
Support
## [](https://www.sigmaaldrich.com)Organic light-emitting diodes (OLEDs)
Organic light-emitting diodes (OLEDs) are electroluminescent devices consisting of an organic semiconductor emissive layer sandwiched between an anode, which is positively charged to inject holes, and a cathode, which is negatively charged to inject electrons, and transport layers that permit different electrical charge carriers to flow into the semiconducting emissive layer. Light is generated directly when charges injected at the electrodes recombine in the organic layer. OLEDs produce vibrant colors and brighter light, providing better contrast in displays. Due to the thinner, more flexible properties of organic materials, OLEDs can be used in curved monitor screens, foldable or rollable mobile devices, and wearable devices.
## [](https://www.sigmaaldrich.com)Organic field-effect transistors (OFETs) & Organic thin-film transistors (OTFTs)
Organic transistors are the basic building blocks for flexible integrated circuits and displays in high-performance electronics. Transistors switch power on and off. Source and drain electrodes have direct contact with an organic semiconductor. The gate electrode is isolated from the semiconductor by the dielectric insulator. When voltage is applied to the gate, it makes the semiconductor more or less conductive, either allowing or preventing electrical current to flow between the source and drain. All components, ranging from conductors (for electrodes) and semiconductors (for active channel materials) to insulators (for gate dielectric layers), can be composed of organic materials. Thin-film transistors are a special type of field-effect transistors, in which the semiconductor, electrode and dielectric layers are deposited as thin films onto a supporting substrate. Common electronic applications include RFID tags or electronic paper.
## [](https://www.sigmaaldrich.com)Organic photovoltaics (OPVs)
Organic electronic materials can also be used as donor and acceptor materials to convert light into electricity in solar panels. In OPVs, photoactive layers of semiconducting organic materials are sandwiched between two electrodes to generate photocurrents. Since the donor absorbs the solar photon flux, donor materials must have wide optical absorption to match the solar spectrum. Organic hole-transport materials (HTMs) used in perovskite solar cells have proven to be particularly efficient in maximizing charge transport and harvesting solar energy.
[](https://www.sigmaaldrich.com/documents-search)
[Looking for More Specific Information?](https://www.sigmaaldrich.com/documents-search)
Visit our document search for data sheets, certificates and technical documentation.
[Find Documents](https://www.sigmaaldrich.com/documents-search)
## Related Articles
- [Light-emitting Polymers](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/organic-electronics/light-emitting-polymers)
LEPs enable a wide range of important applications including sensors, flexible LED displays and lighting devices, optical pump lasers, and potentially polymer diode lasers.
- [Aggregation-induced Emission Luminogens for Non-doped Organic Light-emitting Diodes](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/organic-electronics/aggregation-induced-emission-luminogens)
Organic Light-emitting Diodes (OLEDs) are solid-state devices that transform electrical energy into light. OLEDs are considered the next generation technology for high-resolution flexible displays and solid state lighting, attracting intense scientific and industrial interest.
- [3D/4D Printing Technologies](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/3d-printing/3d-and-4d-printing-technologies)
Three-dimensional printing technology applications range from personal tools to aerospace equipment.
- [Inkjet Printing for Printed Electronics](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/3d-printing/inkjet-printing)
Inkjet printing is one of the key enabling technologies of printed electronics. Inkjet printing technology classification, aspects of materials (inks, substrates) and respective pre-and post-processing steps are discussed.
- [See All (63)](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
## Related Protocols
- [Preparing Self-Assembled Monolayers](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/materials-science-and-engineering/organic-electronics/preparing-self-assembled)
SAMs are versatile surface coatings for chemical resistance, bio-compatibility, sensitization, and molecular recognition for sensors and nano fabrication.
- [Thioacetate Deprotection Procedure](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/materials-science-and-engineering/organic-electronics/thioacetate-deprotection)
Hydrolyzing agents deprotect protected thiol derivatives, obtaining unstable free thiols during complex compound synthesis for self-assembled monolayers.
- [Kit for Creating Hydrophilic PDMS Surfaces](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/materials-science-and-engineering/organic-electronics/kit-for-creating-hydrophilic)
Create durable hydrophilic PDMS surface appropriate for multiple printing applications using hydrophilic inks.
- [See All (4)](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
### Find More Articles and Protocols
Enter Keywords
Search
__Related Highlights__
[WEBINAR - Light Energy Harvesting with Semiconductor Nanostructures](https://www.sigmaaldrich.com/US/en/collections/webinars/w435139627)
## How Can We Help
In case of any questions, please submit a [customer support request](https://www.sigmaaldrich.com/US/en/support/customer-support)
or talk to our customer service team:
Email [custserv@sial.com](mailto:custserv@sial.com)
or call +1 (800) 244-1173
## Additional Support
- [Chromatogram Search](https://www.sigmaaldrich.com/chromatogram-search)
Use the Chromatogram Search to identify unknown compounds in your sample.
- [Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)
Web Toolbox - science research tools and resources for analytical chemistry, life science, chemical synthesis and materials science.
- [Customer Support Request](https://www.sigmaaldrich.com/US/en/support/customer-support)
Customer support including help with orders, products, accounts, and website technical issues.
- [FAQ](https://maestro.my.site.com/knowledgeportal/s/)
Explore our Frequently Asked Questions for answers to commonly asked questions about our products and services.
* * *
__Featured Articles__
[MXenes: A Tunable Family of 2D Carbides and Nitrides](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/organic-electronics/mxenes)
Professor Gogotsi and Dr. Shuck introduce MXenes: a promising family of two-dimensional materials with a unique combination of high conductivity, hydrophilicity, and extensive tunability.
[Light-emitting Polymers](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/organic-electronics/light-emitting-polymers)
LEPs enable a wide range of important applications including sensors, flexible LED displays and lighting devices, optical pump lasers, and potentially polymer diode lasers.
[Aggregation-induced Emission Luminogens for Non-doped Organic Light-emitting Diodes](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/organic-electronics/aggregation-induced-emission-luminogens)
Organic Light-emitting Diodes (OLEDs) are solid-state devices that transform electrical energy into light. OLEDs are considered the next generation technology for high-resolution flexible displays and solid state lighting, attracting intense scientific and industrial interest.
[Preparing Self-Assembled Monolayers](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/materials-science-and-engineering/organic-electronics/preparing-self-assembled)
SAMs are versatile surface coatings for chemical resistance, bio-compatibility, sensitization, and molecular recognition for sensors and nano fabrication.
* * *
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)
An unknown error has occured.