| Lithium Ion Batteries |
| Cat. No. |
Description |
Linear Formula |
Purity |
Form/Particle Size |
Add to Cart |
| Battery Electrode Materials |
| 725145-25G |
NEW Lithium cobalt phosphate |
LiCoPO4 |
99% |
powder, <0.5 µm particle size(TEM) |
|
| 725137-25G |
NEW Lithium manganese dioxide |
LiMnO2 |
>99% trace metals basis |
powder, <1 µm particle size |
|
| 725110-25G |
NEW Lithium manganese nickel oxide, spinel |
Li2Mn3NiO8 |
>99% |
powder, <0.5 µm particle size (TEM) |
|
| 725129-25G |
NEW Lithium manganese oxide, spinel |
LiMn2O4 |
>99% |
powder, <0.5 µm particle size (BET) |
|
| 685933-10G |
Lanthanum-nickel alloy |
LaNi5 |
hydrogen-storage grade |
solid |
|
| 442704-100G-A |
Lithium cobalt(III) oxide |
LiCoO2 |
99.8% trace metals basis |
powder |
|
| 266000-25G |
Lithium ribbon |
Li |
99.9% trace metals basis |
thickness × W 1.5 mm × 100 mm |
|
| 265993-25G |
Lithium ribbon |
Li |
99.9% trace metals basis |
thickness × W 0.75 mm × 45 mm |
|
| 400939-100G |
Lithium titanate |
Li2TiO3 |
- |
325 mesh |
|
| 702277-25G |
Lithium titanate, spinel |
Li4Ti5O12 |
>99% |
nanopowder, <100 nm particle size (BET) |
|
| 693804-1G |
Zirconium-scandium-iron alloy |
Zr4Sc1Fe10 |
hydrogen-storage grade |
solid |
|
| Battery Electrolyte Materials |
| 718319-5G |
NEW 3-(Methylsulfonyl)-1-propyne |
C4H6O2S |
95% |
liquid |
|
| 718203-5G |
NEW Allyl methyl sulfone |
C4H8O2S |
96% |
liquid |
|
| 517135-100ML |
Diethyl carbonate anhydrous |
(C2H5O)2CO |
≥99% |
liquid |
|
| 517127-100ML |
Dimethyl carbonate anhydrous |
(CH3O)2CO |
≥99% |
liquid |
|
| 308315-10G |
Lithium hexafluoroarsenate(V) |
LiAsF6 |
98% |
powder |
|
| 450227-5G |
Lithium hexafluorophosphate battery grade |
LiPF6 |
≥99.99% trace metals basis |
powder |
|
| 634565-10G |
Lithium perchlorate battery grade, dry |
LiClO4 |
99.99% trace metals basis |
powder and chunks |
|
| 451142-5G |
Lithium tetrachloroaluminate anhydrous |
LiAlCl4 |
99.99% trace metals basis |
beads, 10 mesh |
|
| 451622-5G |
Lithium tetrafluoroborate anhydrous |
LiBF4 |
99.998% trace metals basis |
powder |
|
| 481548-5G |
Lithium trifluoromethanesulfonate |
CF3SO3Li |
99.995% trace metals basis |
powder |
|
| |
| Hydrogen Storage Materials |
| Cat. No. |
Description |
Linear Formula |
Purity |
Add to Cart |
| 710199-1G |
NEW Lithium ammonia borane |
LiNH2BH3 |
>90% |
|
| 715247-1G |
NEW Magnesium borohydride |
Mg(BH4)2 |
95% |
|
| 731226-5G |
NEW Magnesium borohydride dimethylsulfide complex |
Mg(BH4)2 · x(CH3)2S |
- |
|
| 682098-1G |
Borane-ammonia complex |
H3N-BH3 |
97% |
|
| 686034-10G |
Lithium aluminum hydride |
LiAlH4 |
hydrogen-storage grade |
|
| 686026-10G |
Lithium borohydride |
LiBH4 |
hydrogen-storage grade, ≥90% |
|
| 683043-10G |
Magnesium hydride |
MgH2 |
hydrogen-storage grade |
|
| 685984-10G |
Sodium aluminum hydride |
NaAlH4 |
hydrogen-storage grade |
|
| 686018-10G |
Sodium borohydride |
NaBH4 |
hydrogen-storage grade, 98% |
|
| 686069-1G |
Titanium(II) hydride |
TiH2 |
hydrogen-storage grade |
|
| |
| Fuel Cell Materials |
| Cat. No. |
Description |
Linear Formula |
Purity |
Form/Particle Size |
Add to Cart |
| SOFC Electrode Materials |
| 729175-10G |
NEW Lanthanum nickelate |
La2NiO4 |
<100 nm particle size (BET) |
nanopowder |
|
| 729183-10G |
NEW Lanthanum nickelate, strontium doped |
La1.6Sr0.4NiO4 |
<100 nm particle size (BET) |
nanopowder |
|
| 734632-10G |
NEW Lanthanum strontium cobalt ferrite |
La0.6Sr0.4Co0.2Fe0.8O3 |
<0.5 µm particle size |
powder |
|
| 734640-10G |
NEW Lanthanum strontium ferrite |
La0.6Sr0.4FeO3 |
<0.5 µm particle size |
powder |
|
| 734659-10G |
NEW Manganese cobalt oxide spinel |
Mn1.5Co1.5O4 |
<0.5 µm particle size |
powder |
|
| 704253-10G |
Lanthanum strontium cobalt ferrite composite, LSCF/GDC |
Ce0.9Gd0.102, 50 wt. % La0.6Sr0.4Co0.2Fe0.8O3, 50 wt. % |
- |
powder |
|
| 704288-10G |
Lanthanum strontium cobalt ferrite LSCF 6428 |
La0.6Sr0.4Co0.2Fe0.8O3 |
0.3-0.6 µm particle size |
powder |
|
| 704237-10G |
Lanthanum strontium manganite composite, LSM-20/GDC10 |
Ce0.9Gd0.1O2, 50 wt. % La0.8Sr0.2MnO3, 50 wt. % |
- |
powder |
|
| 704245-10G |
Lanthanum strontium manganite composite, LSM-20/YSZ |
(Y2O3)0.08(ZrO2)0.92, 50 wt. % La0.8Sr0.2MnO3, 50 wt. % |
≥99% |
powder |
|
| 704296-10G |
Lanthanum strontium manganite LSM-20 |
La0.8Sr0.2MnO3 |
≥99% purity |
powder |
|
| 704261-10G |
Lanthanum strontium manganite LSM-35 |
La0.65Sr0.35MnO3 |
0.3-0.5 µm particle size |
powder |
|
| 704210-10G |
Nickel oxide - Cerium samarium oxide for coatings, NiO/SDC |
(CeO2-Sm2O3), 40 wt. % NiO, 60 wt. % |
≥99% trace metals basis |
solid |
|
| 704202-10G |
Nickel oxide - Yttria-stabilized zirconia for coatings, NiO/YSZ |
(Y2O3-ZrO2), 34 wt. % NiO, 66 wt. % |
0.7-1.2 µm particle size(d50) |
powder |
|
| 704229-10G |
Nickel oxide - Yttria-stabilized zirconia, NiO/YSZ |
NiO, 60 wt. % Y2O3-ZrO2, 40 wt. % |
≥99% |
solid |
|
| SOFC Electrolyte Materials |
| 729191-10G |
NEW Cerium oxide, praseodymium doped |
Ce0.8Pr0.2O2 |
<100 nm particle size |
nanopowder |
|
| 734519-10G |
NEW Cerium(IV) oxide-gadolinium doped, contains 10 mol % gadolinium as dopant |
Gd0.1Ce0.9O1.95 |
<0.5 µm particle size |
powder |
|
| 734667-10G |
NEW Cerium(IV) oxide-gadolinium doped pellets (for sputtering), contains 20 mol % gadolinium as dopant |
Gd0.2Ce0.8O1.9 |
Diam x H = 2-3 mm x 3-5 mm |
pellets (for sputtering) |
|
| 734675-10G |
NEW Cerium(IV) oxide-samarium doped pellets (for sputtering), contains 15 mol % samarium as dopant |
Sm0.15Ce0.85O2 |
Diam x H = 2-3 mm x 3-5 mm |
pellets (for sputtering) |
|
| 734624-10G |
NEW Cerium(IV) oxide-samarium doped, contains 20 mol % samarium as dopant |
Sm0.2Ce0.8O1.9 |
<0.5 µm particle size |
powder |
|
| 734683-10G |
NEW Zirconium(IV) oxide-yttria stabilized pellets (for sputtering) |
ZrO2/Y2O3 |
Diam x H = 2-3 mm x 3-5 mm |
pellets (for sputtering) |
|
| 572330-25G |
Cerium(IV) oxide-gadolinium doped, contains 10 mol % gadolinium as dopant |
Ce0.9Gd0.1O2 |
5-10 nm (crystallite size), <500 nm (secondary) |
nanopowder |
|
| 572357-25G |
Cerium(IV) oxide-gadolinium doped, contains 20 mol % gadolinium as dopant |
Ce0.8Gd0.2O2 |
<100 nm particle size |
nanopowder |
|
| 572365-25G |
Cerium(IV) oxide-samarium doped, contains 15 mol % samarium as dopant |
Ce0.85Sm0.15O2 |
<100 nm particle size |
nanopowder |
|
| 572349-25G |
Zirconium(IV) oxide-yttria stabilized, YSZ-8 |
(ZrO2)0.92(Y2O3)0.08 |
≤100 nm particle size |
nanopowder |
|
| Back to Top |