| SAFCB Item Number |
Description |
Sigma Item Number |
Comments |
| 50120C |
BME W/ EARLE'S BAL. SALTS (10X)W/O L-GLUT., W/O NAHCO3 |
B1522 |
Sigma product contains Sodium Bicarbonate. (1X only) |
| 51440C |
DME HG W/O L-GLUTW/OMETH |
D0422 |
Sigma product is also w/out L-cystine |
| 51202C |
L-15 (LEIBOVITZ) MED.W/ 2.05 MM L-GLUT. |
L1518 |
Identical |
| 56509C |
RPMI-1640 MEDIUM, WITH L-GLUTAMINE |
R6504 |
Identical |
| 51312C |
MEDIUM.199 MOD. (M199), W/ EARLE'S BAL. SALTSW/ 0.68 MM L-GLUT. |
M4530 |
Identical |
| 51411C |
MEM W/ EARLE'S BAL. SALTS (EMEM)W/ 2.0 MM L-GLUT. |
M4655 |
Identical |
| 51412C |
MEM MOD., W/ EARLE'S BAL. SALTS (EMEM)W/O L-GLUT. |
M2279 |
Identical |
| 51415C |
MEM MOD., W/ EARLE'S BAL. SALTS (EMEM)W/O L-GLUT., W/ HEPES |
M7278 |
Identical |
| 51416C |
MEM W/ EARLE'S BAL. SALTS (EMEM/NEAA)W/ NONESSENTIAL AMINO ACIDS, W/O L-GLUT. |
M5650 |
Identical |
| 51441C |
DULBECCO'S MOD. EAGLE'S MED.DME/HIGH, W/ 4500 MG/L GLUCOSE |
D5796 |
Identical |
| 51442C |
DULBECCO'S MOD. EAGLE'S MED.DME/HIGH MOD., W/ 4500 MG/L GLUCOSE |
D5671 |
Identical |
| 51443C |
DULBECCO'S MOD. EAGLE'S MED.DME/HIGH MOD., W/ 4500 MG/L GLUCOSE |
D6429 |
Identical |
| 51444C |
DULBECCO'S MOD. EAGLE'S MED.W/ 4500 MG/L GLUCOSE, W/O L-GLUT. |
D6546 |
Identical |
| 51447C |
DULBECCO'S MOD. EAGLE'S MED.DME/LOW, W/ 1000 MG/L GLUCOSE |
D6046 |
Identical |
| 51450C |
DULBECCO'S MOD. EAGLE'S MED.WITH HEPES |
D6171 |
Identical |
| 51451C |
MEM, ALPHA MOD. (AMEM)W/ 2.0 MM L-GLUT. |
M4526 |
Sigma product is without L - Glutamine |
| 51452C |
MEM, ALPHA MOD. (AMEM)W/ 2.0 MM L-GLUT., WITH NUCLEOSIDES |
M8042 |
Sigma product is without L - Glutamine |
| 51471C |
ISCOVE'S MOD. DULBECCO'S MED. (IMDM)W/ 25 MM HEPES, W/ 4.0 MM L-GLUT. |
I3390 |
Sigma product is without L - Glutamine |
| 51501C |
RPMI 1641W/ 2.05 MM L-GLUT. |
R8758 |
Identical |
| 51502C |
RPMI 1640 MOD.W/O L-GLUT. |
R0883 |
Identical |
| 51503C |
RPMI 1640W/O L-GLUT., W/O PHENOL RED |
R7509 |
Identical |
| 51535C |
RPMI 1640 MOD.W/ 25 MM HEPES, W/O L-GLUT. |
R5886 |
Identical |
| 51536C |
RPMI 1640 MOD.W/ 25 MM HEPES, W/ 2.05 MM L-GLUT. |
R7388 |
Sigma product does not contain Sodium Bicarbonate. |
| 51631C |
HAM'S F10W/ 1.0 MM L-GLUT. |
N6013 |
Sigma product is without L - Glutamine |
| 51651C |
HAM'S F12W/ 1.0 MM L-GLUT. |
N4888 |
Sigma product is without L - Glutamine |
| 51671C |
MC COY'S 5A MED., IWAKATA & GRACE MOD.W/ 1.5 MM L-GLUT. |
M8403 |
Sigma product is without L - Glutamine |
| 51942C |
HINK'S TNM-FH INSECT MED |
T3285 |
Identical |
| 56312C |
MEDIUM 199/EBSS 10L |
M5017 |
Identical |
| 56419C |
EMEM |
M0268 |
Identical |
| 56459C |
MEM ALPHA MODIFICATION |
M0894 |
Identical |
| 56469C |
DME LG W/L-GLUTAMINE |
D5523 |
Identical |
| 58672C |
GS SUPPLEMENT (50X) |
G9785 |
Identical |
| 59226C |
NO-ZYME(TM) (1X) |
C5914 |
Identical |
| 55021C |
HANKS' BAL. SALT SOL. (HBSS MOD.)W/O CALCIUM, W/O MAGNESIUM |
H9394 |
Identical |
| 55024C |
HANKS' BAL. SALT SOL. (HBSS)W/ CALCIUM, W/ MAGNESIUM |
H9269 |
Identical |
| 55025C |
HANKS' BAL. SALT SOL. (HBSS MOD.)W/O CALCIUM, W/O MAGNESIUM |
H6648 |
Identical |
| 55026C |
TYRODE'S SALT SOLUTION |
T2397 |
Identical |
| 55037C |
HANKS' BAL. SALT SOL. (HBSS MOD.)W/ CALCIUM, W/ MAGNESIUM |
H8264 |
Identical |
| 55114C |
EARLE'S BAL. SALT SOL. (EBSS) |
E2888 |
Identical |
| 55225C |
HANKS' BAL. SALT SOL. (HBSS MOD.) (10X)W/ PHENOL RED |
H1641 |
Identical |
| 58572C |
MEM NONESSENTIAL AMINO ACIDS (100X) |
M7145 |
Identical |
| 58970C |
MEM VITAMINS SOL. (100X) |
M6895 |
Identical |
| 59203C |
SODIUM PYRUVATE SOL. (100 MM)11.0 G/L NA PYRUVATE |
S8636 |
Identical |
| 59205C |
HEPES (1M SOL.) |
H0887 |
Identical |
| 59221C |
SODIUM BICARBONATE (7.5% SOL.) |
S8761 |
Identical |
| 59223C |
SODIUM HYDROXIDE 1N |
S2770 |
Identical |
| 59300C |
DULBECCO'S PBS (DPBS)W/ CALCIUM, W/ MAGNESIUM |
D8662 |
Identical |
| 59321C |
DULBECCO'S PBS (DPBS MOD.)W/O CALCIUM, W/O MAGNESIUM |
D8537 |
Identical |
| 59331C |
DULBECCO'S PBS (10X DPBS MOD.) |
D1408 |
Identical |
| 59915C |
PLURONIC(R) F-68 (10% SOL.) |
P5556 |
Identical |
| 50444C |
DULBECCO'S MOD. EAGLE'S MED. (5X)DME/HIGH, W/ 4500 MG/L GLUCOSE, W/O L-GLUT., W/O NAHCO3 |
D6546 |
D6546 is 1X ONLY and contains Sodium Bicarbonate. |
| 56064C |
DULBECCO'S PBS (DPBS MOD.)W/O CALCIUM, W/O MAGNESIUM |
D5652 |
Identical |
| 56679C |
MC COY'S 5A MED., IWAKATA & GRACE MOD.W/ 1.5 MM L-GLUT., W/O NAHCO3 |
M4892 |
Identical |
| 56902C |
GRACE'S INSECT MED., ORIG. FORMULAW/ 4.1 MM L-GLUT., W/O CALCIUM CHLORIDE |
G9771 |
Identical |
| 56923C |
IPL-41 INSECT MED., MOD.W/O NAHCO3 |
I7760 |
Sigma product only available in liquid formulation containing Sodium Bicarbonate. Powder not available. |
| 56439C |
DME HG W/L-GLUTAMINE |
D5648 |
Identical |
| 56941C |
TC-100 INSECT MED.W/O NAHCO3 |
T3160 |
Sigma product only available in liquid formulation containing Sodium Bicarbonate. Powder not available. |
| 51445C |
DULBECCO'S MOD. EAGLE'S MED./HAM'S F12W/ 15 MM HEPES, W/ 2.5 MM L-GLUT. |
D8437 |
Identical |
| 51448C |
DULBECCO'S MOD. EAGLE'S MED./HAM'S F12W/ 3151 MG/L GLUCOSE, W/ 2.5 MM L-GLUT. |
D8062 |
Identical |
| 51449C |
DULBECCO'S MOD. EAGLE'S MED.DME/HIGH, W/ 4500 MG/L GLUCOSE, W/O L-GLUT., W/O PHENOL RED |
D1145 |
Sigma product is without Sodium Pyruvate |
| 14400C |
EX-CELL(TM)400 W/L-GLUT 1000ML |
No Equivalent |
No Equivalent |
| 14401C |
EX-CELL(TM)401 W/L-GLUT 1000ML |
No Equivalent |
No Equivalent |
| 14421C |
EX-CELL(TM)420 W/OL-GLUT1000ML |
No Equivalent |
No Equivalent |
| 14620C |
EX-CELL(TM) 620-HSF 1000ML |
No Equivalent |
No Equivalent |
| 58902C |
YEASTOLATE UF (50X) |
Y4375 |
|
| 51653C |
HAM'S F12W/O L-GLUT., W/O HYPOXANTHINE |
No equivalent |
No Equivalent |
| 51417C |
MEM W/ EARLE'S BAL. SALTS (EMEM/NEAA)W/ NONESSENTIAL AMINO ACIDS, W/ 20 MM HEPES |
No Equivalent |
No Equivalent |