| SAFCB Item Number | Description | Sigma Item Number | Comments |
| 50120C | BME W/ EARLE'S BAL. SALTS (10X)W/O L-GLUT., W/O NAHCO3 | B1522 | Sigma product contains Sodium Bicarbonate. (1X only) |
| 51440C | DME HG W/O L-GLUTW/OMETH | D0422 | Sigma product is also w/out L-cystine |
| 51202C | L-15 (LEIBOVITZ) MED.W/ 2.05 MM L-GLUT. | L1518 | Identical |
| 56509C | RPMI-1640 MEDIUM, WITH L-GLUTAMINE | R6504 | Identical |
| 51312C | MEDIUM.199 MOD. (M199), W/ EARLE'S BAL. SALTSW/ 0.68 MM L-GLUT. | M4530 | Identical |
| 51411C | MEM W/ EARLE'S BAL. SALTS (EMEM)W/ 2.0 MM L-GLUT. | M4655 | Identical |
| 51412C | MEM MOD., W/ EARLE'S BAL. SALTS (EMEM)W/O L-GLUT. | M2279 | Identical |
| 51415C | MEM MOD., W/ EARLE'S BAL. SALTS (EMEM)W/O L-GLUT., W/ HEPES | M7278 | Identical |
| 51416C | MEM W/ EARLE'S BAL. SALTS (EMEM/NEAA)W/ NONESSENTIAL AMINO ACIDS, W/O L-GLUT. | M5650 | Identical |
| 51441C | DULBECCO'S MOD. EAGLE'S MED.DME/HIGH, W/ 4500 MG/L GLUCOSE | D5796 | Identical |
| 51442C | DULBECCO'S MOD. EAGLE'S MED.DME/HIGH MOD., W/ 4500 MG/L GLUCOSE | D5671 | Identical |
| 51443C | DULBECCO'S MOD. EAGLE'S MED.DME/HIGH MOD., W/ 4500 MG/L GLUCOSE | D6429 | Identical |
| 51444C | DULBECCO'S MOD. EAGLE'S MED.W/ 4500 MG/L GLUCOSE, W/O L-GLUT. | D6546 | Identical |
| 51447C | DULBECCO'S MOD. EAGLE'S MED.DME/LOW, W/ 1000 MG/L GLUCOSE | D6046 | Identical |
| 51450C | DULBECCO'S MOD. EAGLE'S MED.WITH HEPES | D6171 | Identical |
| 51451C | MEM, ALPHA MOD. (AMEM)W/ 2.0 MM L-GLUT. | M4526 | Sigma product is without L - Glutamine |
| 51452C | MEM, ALPHA MOD. (AMEM)W/ 2.0 MM L-GLUT., WITH NUCLEOSIDES | M8042 | Sigma product is without L - Glutamine |
| 51471C | ISCOVE'S MOD. DULBECCO'S MED. (IMDM)W/ 25 MM HEPES, W/ 4.0 MM L-GLUT. | I3390 | Sigma product is without L - Glutamine |
| 51501C | RPMI 1641W/ 2.05 MM L-GLUT. | R8758 | Identical |
| 51502C | RPMI 1640 MOD.W/O L-GLUT. | R0883 | Identical |
| 51503C | RPMI 1640W/O L-GLUT., W/O PHENOL RED | R7509 | Identical |
| 51535C | RPMI 1640 MOD.W/ 25 MM HEPES, W/O L-GLUT. | R5886 | Identical |
| 51536C | RPMI 1640 MOD.W/ 25 MM HEPES, W/ 2.05 MM L-GLUT. | R7388 | Sigma product does not contain Sodium Bicarbonate. |
| 51631C | HAM'S F10W/ 1.0 MM L-GLUT. | N6013 | Sigma product is without L - Glutamine |
| 51651C | HAM'S F12W/ 1.0 MM L-GLUT. | N4888 | Sigma product is without L - Glutamine |
| 51671C | MC COY'S 5A MED., IWAKATA & GRACE MOD.W/ 1.5 MM L-GLUT. | M8403 | Sigma product is without L - Glutamine |
| 51942C | HINK'S TNM-FH INSECT MED | T3285 | Identical |
| 56312C | MEDIUM 199/EBSS 10L | M5017 | Identical |
| 56419C | EMEM | M0268 | Identical |
| 56459C | MEM ALPHA MODIFICATION | M0894 | Identical |
| 56469C | DME LG W/L-GLUTAMINE | D5523 | Identical |
| 58672C | GS SUPPLEMENT (50X) | G9785 | Identical |
| 59226C | NO-ZYME(TM) (1X) | C5914 | Identical |
| 55021C | HANKS' BAL. SALT SOL. (HBSS MOD.)W/O CALCIUM, W/O MAGNESIUM | H9394 | Identical |
| 55024C | HANKS' BAL. SALT SOL. (HBSS)W/ CALCIUM, W/ MAGNESIUM | H9269 | Identical |
| 55025C | HANKS' BAL. SALT SOL. (HBSS MOD.)W/O CALCIUM, W/O MAGNESIUM | H6648 | Identical |
| 55026C | TYRODE'S SALT SOLUTION | T2397 | Identical |
| 55037C | HANKS' BAL. SALT SOL. (HBSS MOD.)W/ CALCIUM, W/ MAGNESIUM | H8264 | Identical |
| 55114C | EARLE'S BAL. SALT SOL. (EBSS) | E2888 | Identical |
| 55225C | HANKS' BAL. SALT SOL. (HBSS MOD.) (10X)W/ PHENOL RED | H1641 | Identical |
| 58572C | MEM NONESSENTIAL AMINO ACIDS (100X) | M7145 | Identical |
| 58970C | MEM VITAMINS SOL. (100X) | M6895 | Identical |
| 59203C | SODIUM PYRUVATE SOL. (100 MM)11.0 G/L NA PYRUVATE | S8636 | Identical |
| 59205C | HEPES (1M SOL.) | H0887 | Identical |
| 59221C | SODIUM BICARBONATE (7.5% SOL.) | S8761 | Identical |
| 59223C | SODIUM HYDROXIDE 1N | S2770 | Identical |
| 59300C | DULBECCO'S PBS (DPBS)W/ CALCIUM, W/ MAGNESIUM | D8662 | Identical |
| 59321C | DULBECCO'S PBS (DPBS MOD.)W/O CALCIUM, W/O MAGNESIUM | D8537 | Identical |
| 59331C | DULBECCO'S PBS (10X DPBS MOD.) | D1408 | Identical |
| 59915C | PLURONIC(R) F-68 (10% SOL.) | P5556 | Identical |
| 50444C | DULBECCO'S MOD. EAGLE'S MED. (5X)DME/HIGH, W/ 4500 MG/L GLUCOSE, W/O L-GLUT., W/O NAHCO3 | D6546 | D6546 is 1X ONLY and contains Sodium Bicarbonate. |
| 56064C | DULBECCO'S PBS (DPBS MOD.)W/O CALCIUM, W/O MAGNESIUM | D5652 | Identical |
| 56679C | MC COY'S 5A MED., IWAKATA & GRACE MOD.W/ 1.5 MM L-GLUT., W/O NAHCO3 | M4892 | Identical |
| 56902C | GRACE'S INSECT MED., ORIG. FORMULAW/ 4.1 MM L-GLUT., W/O CALCIUM CHLORIDE | G9771 | Identical |
| 56923C | IPL-41 INSECT MED., MOD.W/O NAHCO3 | I7760 | Sigma product only available in liquid formulation containing Sodium Bicarbonate. Powder not available. |
| 56439C | DME HG W/L-GLUTAMINE | D5648 | Identical |
| 56941C | TC-100 INSECT MED.W/O NAHCO3 | T3160 | Sigma product only available in liquid formulation containing Sodium Bicarbonate. Powder not available. |
| 51445C | DULBECCO'S MOD. EAGLE'S MED./HAM'S F12W/ 15 MM HEPES, W/ 2.5 MM L-GLUT. | D8437 | Identical |
| 51448C | DULBECCO'S MOD. EAGLE'S MED./HAM'S F12W/ 3151 MG/L GLUCOSE, W/ 2.5 MM L-GLUT. | D8062 | Identical |
| 51449C | DULBECCO'S MOD. EAGLE'S MED.DME/HIGH, W/ 4500 MG/L GLUCOSE, W/O L-GLUT., W/O PHENOL RED | D1145 | Sigma product is without Sodium Pyruvate |
| 14400C | EX-CELL(TM)400 W/L-GLUT 1000ML | No Equivalent | No Equivalent |
| 14401C | EX-CELL(TM)401 W/L-GLUT 1000ML | No Equivalent | No Equivalent |
| 14421C | EX-CELL(TM)420 W/OL-GLUT1000ML | No Equivalent | No Equivalent |
| 14620C | EX-CELL(TM) 620-HSF 1000ML | No Equivalent | No Equivalent |
| 58902C | YEASTOLATE UF (50X) | Y4375 | |
| 51653C | HAM'S F12W/O L-GLUT., W/O HYPOXANTHINE | No equivalent | No Equivalent |
| 51417C | MEM W/ EARLE'S BAL. SALTS (EMEM/NEAA)W/ NONESSENTIAL AMINO ACIDS, W/ 20 MM HEPES | No Equivalent | No Equivalent |